1 - 2 of 2 products displayed
  • Dynorphin A Antibody
  • Host Rabbit
  • Target Species Human, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA184557
  • Size 100 µL
Applications
Immunohistochemistry (Frozen) (1/100-1/1500), Immunohistochemistry (Paraffin) (Assay Dependent), Radioimmune Assays (1/10,000)
Alternate names:
Preprodynorphin; Beta-neoendorphin-dynorphin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184557 475.00
Check your price
  • Prodynorphin Antibody
  • Host Guinea Pig
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA127974
  • Size 50 µL
Applications
Immunohistochemistry (Frozen) (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127974 349.00
Check your price