1 - 30 of 30 products displayed
  • Biotin Monoclonal Antibody, HRP conjugate for ICC, IHC (P) and ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate HRP
  • Catalog # 033720
  • Size 1mL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
033720 334.00
Check your price
  • Biotin Polyclonal Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Chemical
  • Conjugate HRP
  • Catalog # PA130595
  • Size 1mg
Applications
ELISA (1:10,000-1:40,000), Immunohistochemistry (Assay-Dependent), Western Blot (1:1000-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130595 275.00
Check your price
  • Goat anti-Mouse IgG / IgA / IgM (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot and IF
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A10676
  • Size 250µL
Applications
Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A10676 126.00
Check your price
  • Mouse anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # 31824
  • Size 1mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31824 168.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # 31822
  • Size 1.5mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31822 143.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # 31802
  • Size 1.5mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31802 157.00
Check your price
  • Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, DSB-X Biotin conjugate for IF and ICC
  • Host Goat
  • Target Species Mouse
  • Conjugate DSB-X Biotin
  • Catalog # D20691
  • Size 500µL
Applications
Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
D20691 179.00
Check your price
  • Chicken anti-Mouse IgG Fab Secondary Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Chicken
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # SA172024
  • Size 500µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172024 225.00
Check your price
  • Chicken anti-Mouse IgG Secondary Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Chicken
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # SA172020
  • Size 500µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172020 134.00
Check your price
  • Donkey anti-Mouse IgG Secondary Antibody, Biotin conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Donkey
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # PA128627
  • Size 1mg
Applications
Dot blot (Assay dependent), ELISA (1:20,000-1:100,000), Immunohistochemistry (Frozen) (1:1000-1:5000), Immunohistochemistry (Paraffin) (1:1000-1:5000), Immunomicroscopy (Assay-dependent), Western Blot (1:2000-1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128627 209.00
Check your price
  • Rabbit anti-Mouse IgG Secondary Antibody, Biotin conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # PA128567
  • Size 1mg
Applications
ELISA (1:300,000), Immunohistochemistry (Frozen) (1:1000-1:5000), Immunohistochemistry (Paraffin) (1:1000-1:5000), Immunomicroscopy (Assay-dependent), Western Blot (1:2000-1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128567 194.00
Check your price
  • Bovine anti-Mouse IgG Secondary Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Bovine
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # SA19507
  • Size 500µg
Applications
ELISA (1:5000-1:20000), Western Blot (1:5000-1:40000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA19507 154.00
Check your price
  • Goat anti-Rabbit IgG F(ab')2 Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # 31823
  • Size 2mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31823 157.00
Check your price
  • Goat anti-Mouse IgG F(ab')2 Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # 31803
  • Size 2mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31803 180.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # A24535
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24535 106.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # 31820
  • Size 1.5mg
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31820 195.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # A16100
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16100 84.25
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # A16108
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16108 136.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # A16114
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16114 157.00
Check your price
  • F(ab')2-Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Donkey
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A24510
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24510 225.00
Check your price
  • F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A24516
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24516 106.00
Check your price
  • F(ab')2-Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A24522
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24522 225.00
Check your price
  • F(ab')2-Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A24528
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24528 137.00
Check your price
  • Chicken anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Chicken
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A15985
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15985 139.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Donkey
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A16015
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16015 157.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Donkey
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A16021
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16021 151.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A16170
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16170 179.00
Check your price
  • Horse anti-Mouse IgG (H+L) Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Horse
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # 31806
  • Size 1.5mg
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31806 195.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # 31800
  • Size 2mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31800 139.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A16070
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16070 106.00
Check your price