1 - 30 of 30 products displayed
  • p300 Monoclonal Antibody, Biotin conjugate for IP and ChIP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # 730044
  • Size 100µg
Applications
Immunoprecipitation (Assay Dependent)
Alternate names:
E1A-associated protein p300
Catalog # Size Price (AUD) Your Price (AUD) Qty
730044 309.00
Check your price
  • Actin Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Human, Mouse, Non-human primate, Plant, Pig, Protozoa, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA511866
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (2 ug), Western Blot (1:5000-1:20,000)
Alternate names:
Smooth muscle gamma-actin; Gamma-2-actin; Alpha-actin-3; Beta-actin; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1; Alpha-cardiac actin; Alpha-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511866 399.00
Check your price
  • GAPDH Loading Control Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast
  • Conjugate Biotin
  • Catalog # MA515738BTIN
  • Size 50µL
Applications
ELISA (Assay Dependent), Flow Cytometry (2 ug/test), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (1:500-1:1000)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738BTIN 159.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody, Biotin conjugate for Western Blot and Flow
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA515739BTIN
  • Size 50µL
Applications
Flow Cytometry (2 ug/test), Western Blot (1:500-1:1000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739BTIN 159.00
Check your price
  • Streptavidin Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA517283
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunoprecipitation (1:3000), Western Blot (1:1000-1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517283 349.00
Check your price
  • Streptavidin Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA517282
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517282 349.00
Check your price
  • Streptavidin Monoclonal Antibody for Western Blot, IHC (F), IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA120010
  • Size 500µg
Applications
ELISA (Assay dependent), Flow Cytometry (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120010 399.00
Check your price
  • Streptavidin Monoclonal Antibody for Western Blot, IHC, Flow and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA120011
  • Size 500µg
Applications
ELISA (Assay dependent), Flow Cytometry (Assay dependent), Immunohistochemistry (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120011 399.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody, Biotin conjugate for Western Blot, IP, ChIP and GS
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Sheep, Pig, Rabbit, Rat, Bovine, Dog, Chicken
  • Conjugate Biotin
  • Catalog # MA513062
  • Size 500µL
Applications
ChIP assay (Assay Dependent), Gel Shift (1mg/mL), Immunoprecipitation (2 µg/ml), Western Blot (1:1000-1:5000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513062 399.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody, Biotin conjugate for IF, ICC, IP, GS and AP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA512325
  • Size 500µL
Applications
Affinity Purification (Assay Dependent), Gel Shift (1mg/mL), Immunocytochemistry (1:500-1:2000), Immunofluorescence (1:500-1:2000), Immunoprecipitation (2 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512325 387.00
Check your price
  • HuC/HuD Monoclonal Antibody, Biotin conjugate for Western Blot and IF
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # A21272
  • Size 100µg
Applications
Immunofluorescence (Assay Dependent), Western Blot (1-5 ug/ml)
Alternate names:
Paraneoplastic limbic encephalitis antigen 21; Paraneoplastic cerebellar degeneration-associated antigen; Hu-antigen C; Paraneoplastic encephalomyelitis antigen HuD; Hu-antigen D
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21272 387.00
Check your price
  • c-Myc Tag Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Biotin
  • Catalog # MA121316BTIN
  • Size 50µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:500-1:2000), Immunofluorescence (1:500-1:2000), Immunoprecipitation (Assay Dependent), Western Blot (1:500-1:1000)
Alternate names:
Transcription factor p64; Proto-oncogene c-Myc; Class E basic helix-loop-helix protein 39
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121316BTIN 159.00
Check your price
  • IL-6 Monoclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # M621B
  • Size 250ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Interferon beta-2; Hybridoma growth factor; CTL differentiation factor; B-cell stimulatory factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
M621B 399.00
Check your price
  • IL-5 Monoclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Rat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # M550B
  • Size 250ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
T-cell replacing factor; Eosinophil differentiation factor; B-cell differentiation factor I
Catalog # Size Price (AUD) Your Price (AUD) Qty
M550B 349.00
Check your price
  • Androgen Receptor Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513423
  • Size 500µL
Applications
Immunocytochemistry (1:25-1:50), Immunofluorescence (1:25-1:50), Immunohistochemistry (Paraffin) (1:25), Immunoprecipitation (2 ug), Western Blot (1:100-1:200)
Alternate names:
Nuclear receptor subfamily 3 group C member 4; Dihydrotestosterone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513423 420.00
Check your price
  • TNF alpha Monoclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # M302B
  • Size 250µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tumor necrosis factor ligand superfamily member 2; TNF-alpha; Cachectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
M302B 349.00
Check your price
  • TNF alpha Monoclonal Antibody, Biotin conjugate for Western Blot, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MM350DB
  • Size 250ug
Applications
ELISA (Assay Dependent), Neutralization (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tumor necrosis factor ligand superfamily member 2; TNF-alpha; Cachectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MM350DB 349.00
Check your price
  • HP1 alpha Monoclonal Antibody, Biotin conjugate for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # 730046
  • Size 100µg
Applications
Immunoprecipitation (Assay Dependent)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
730046 309.00
Check your price
  • IL-6 Monoclonal Antibody, Biotin conjugate for Western Blot, IHC (P), IP, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MM600CB
  • Size 250ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Neutralization (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Interleukin HP-1; B-cell hybridoma growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MM600CB 349.00
Check your price
  • PCNA Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Dog, Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA511355
  • Size 500µL
Applications
Flow Cytometry (1:200-1:1000), Immunocytochemistry (1:20-1:50), Immunofluorescence (1:20-1:50), Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (2 µg/ml), Western Blot (1:50-1:1000)
Alternate names:
Cyclin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511355 375.00
Check your price
  • CXCL8 Monoclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # M802B
  • Size 250ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
T-cell chemotactic factor; Protein 3-10C; Neutrophil-activating protein 1; Monocyte-derived neutrophil-activating peptide; Monocyte-derived neutrophil chemotactic factor; Granulocyte chemotactic protein 1; Emoctakin; Chemokine (C-X-C motif) ligand 8; C-X-C motif chemokine 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
M802B 425.00
Check your price
  • GP340 Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB2130102
  • Size 200µl
Applications
ELISA (1:60,000), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:200-1:10,000)
Alternate names:
Surfactant pulmonary-associated D-binding protein; Salivary agglutinin; Hensin; Glycoprotein 340
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2130102 399.00
Check your price
  • IL18R1 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA120257
  • Size 50µg
Applications
Flow Cytometry (Neat-1:10 (10 µl for 10^6 cells in 100 µl))
Alternate names:
IL1 receptor-related protein; CDw218a; CD218 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120257 349.00
Check your price
  • TAP Tag Monoclonal Antibody, Biotin conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate Biotin
  • Catalog # MA1108BTIN
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
Tobacco Etch Virus, Tandem affinity purfication tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1108BTIN 159.00
Check your price
  • GM-CSF Monoclonal Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # M501B
  • Size 250ug
Applications
ELISA (Assay Dependent)
Alternate names:
Sargramostim; Molgramostin; Colony-stimulating factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
M501B 375.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA516308BTIN
  • Size 50µL
Applications
ELISA (Assay Dependent), Flow Cytometry (1 ug/test), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308BTIN 159.00
Check your price
  • IL-10 Monoclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Rat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # M011B
  • Size 250µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Cytokine synthesis inhibitory factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
M011B 375.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # 31822
  • Size 1.5mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31822 143.00
Check your price
  • Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, DSB-X Biotin conjugate for IF and ICC
  • Host Goat
  • Target Species Mouse
  • Conjugate DSB-X Biotin
  • Catalog # D20691
  • Size 500µL
Applications
Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
D20691 179.00
Check your price
  • Chicken anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Chicken
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # A15997
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15997 136.00
Check your price