1 - 19 of 19 products displayed
  • BrdU Monoclonal Antibody for ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA119213
  • Size 100µg
Applications
Flow Cytometry (1-2 µg/ml), Immunocytochemistry (2 ug/ml), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119213 199.00
Check your price
  • BrdU Monoclonal Antibody for IF, IHC (P) and Flow
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA511285
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:500)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511285 309.00
Check your price
  • BrdU Monoclonal Antibody, Biotin conjugate for IHC
  • Host Mouse
  • Target Species Chemical
  • Conjugate Biotin
  • Catalog # 033940
  • Size 1mL
Applications
Immunohistochemistry (Assay Dependent)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
033940 324.00
Check your price
  • BrdU Monoclonal Antibody for IF and IHC
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # 033900
  • Size 400µg
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
033900 304.00
Check your price
  • BrdU Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA512502
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50-100)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512502 349.00
Check your price
  • BrdU Monoclonal Antibody, Alexa Fluor® 647 conjugate for IF and IHC
  • Host Mouse
  • Target Species Chemical
  • Conjugate Alexa Fluor® 647
  • Catalog # B35133
  • Size 350µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
B35133 351.00
Check your price
  • BrdU Monoclonal Antibody, Alexa Fluor® 594 conjugate for IF and IHC
  • Host Mouse
  • Target Species Chemical
  • Conjugate Alexa Fluor® 594
  • Catalog # B35132
  • Size 350µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
B35132 348.00
Check your price
  • BrdU Monoclonal Antibody, Alexa Fluor® 488 conjugate for IF and IHC
  • Host Mouse
  • Target Species Chemical
  • Conjugate Alexa Fluor® 488
  • Catalog # B35130
  • Size 350µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
B35130 355.00
Check your price
  • BrdU Monoclonal Antibody, Biotin conjugate for IF and IHC
  • Host Mouse
  • Target Species Chemical
  • Conjugate Biotin
  • Catalog # B35138
  • Size 350µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
B35138 327.00
Check your price
  • BrdU Monoclonal Antibody for IF and IHC
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # B35128
  • Size 350µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
B35128 286.00
Check your price
  • BrdU Monoclonal Antibody for ICC, IHC (P, F) and Flow
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # OMA106200
  • Size 100µg
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:100-1:200)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
OMA106200 375.00
Check your price
  • BrdU Monoclonal Antibody for IF, IHC (P) and Flow
  • Host Rat
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA182088
  • Size 200µg
Applications
Flow Cytometry (1/25 -1/200), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1/25 -1/200)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182088 499.00
Check your price
  • BrdU Monoclonal Antibody, DyLight® 488 conjugate for Flow
  • Host Mouse
  • Target Species Chemical
  • Conjugate DyLight® 488
  • Catalog # MA3071D488
  • Size 100µg
Applications
Flow Cytometry (1:100-1:200)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3071D488 216.00
Check your price
  • BrdU Monoclonal Antibody, FITC conjugate for IHC and Flow
  • Host Mouse
  • Target Species Chemical
  • Conjugate FITC
  • Catalog # MA112686
  • Size 100µg
Applications
Flow Cytometry (1-5 µg/10^6 cells), Immunohistochemistry (Assay Dependent)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112686 309.00
Check your price
  • BrdU Monoclonal Antibody for IHC (P, F) and Flow
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA181890
  • Size 200µg
Applications
Flow Cytometry (1/25 -1/100), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181890 349.00
Check your price
  • BrdU Monoclonal Antibody for IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA3071
  • Size 100µL
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:1000-1:2500), Immunohistochemistry (1:1000-1:2500)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3071 249.00
Check your price
  • Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, Alexa Fluor® 488 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488
  • Catalog # A21121
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21121 149.00
Check your price
  • BrdU Monoclonal Antibody for IF, IHC (P) and Flow
  • Host Rat
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA182718
  • Size 20µg
Applications
Flow Cytometry (1/25-1/200), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin) (1/25-1/200)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182718 149.00
Check your price
  • Fluorescein/Oregon Green Polyclonal Antibody, HRP conjugate for ICC, IHC and ELISA
  • Host Rabbit
  • Target Species Chemical
  • Conjugate HRP
  • Catalog # A21253
  • Size 500µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21253 336.00
Check your price