1 - 30 of 30 products displayed
  • pro-Brain Natriuretic Peptide Antibody (Monoclonal, 13G12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120886
  • Size 200 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Gamma-brain natriuretic peptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120886 349.00
Check your price
  • pro-Brain Natriuretic Peptide Antibody (Monoclonal, 24E11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120631
  • Size 200 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Gamma-brain natriuretic peptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120631 349.00
Check your price
  • pro-Brain Natriuretic Peptide Antibody (Monoclonal, 15F11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120626
  • Size 200 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Gamma-brain natriuretic peptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120626 375.00
Check your price
  • pro-Brain Natriuretic Peptide Antibody (Monoclonal, 5B6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120622
  • Size 200 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Gamma-brain natriuretic peptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120622 349.00
Check your price
  • Glucagon-Like Peptide 2 Antibody (Monoclonal, 11E5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB0060202
  • Size 200 µl
Applications
ELISA (1:15,000)
Alternate names:
Glucagon-like peptide-2 receptor, GLP-2 receptor, GLP-2-R, GLP-2R , GLP-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0060202 425.00
Check your price
  • Peptide Histidine-Methionine Antibody (Monoclonal)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0200302
  • Size 200 µL
Applications
ELISA (1:2,000)
Alternate names:
MGC13587, PHM27
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0200302 475.00
Check your price
  • PYY Antibody, Biotin
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # PA19590
  • Size 100 µL
Applications
ELISA (1:2000-1:10000)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19590 375.00
Check your price
  • PYY Antibody, Biotin
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # PA19589
  • Size 100 µL
Applications
ELISA (1:2000-1:10000)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19589 375.00
Check your price
  • PYY Antibody
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19582
  • Size 100 µL
Applications
ELISA (1:2000-1:5000)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19582 375.00
Check your price
  • PYY Antibody
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19581
  • Size 100 µL
Applications
ELISA (1:2000-1:5000)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19581 375.00
Check your price
  • Human GnRH1 Synthetic Peptide
  • Host N/A
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PEP10933
  • Size 10 mg
Applications
Control (Assay Dependent)
Alternate names:
LHRH, GnRH, Lutropin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP10933 473.00
Check your price
  • Human GnRH1 Synthetic Peptide
  • Host N/A
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PEP10934
  • Size 25 mg
Applications
Control (Assay Dependent)
Alternate names:
LHRH, GnRH, Lutropin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP10934 550.00
Check your price
  • Procollagen I C-Peptide Antibody (Monoclonal, 18)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # BTE0021802
  • Size 200 µL
Applications
ELISA (1:3,000), Western Blot (Assay Dependent)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
BTE0021802 600.00
Check your price
  • Procollagen I C-Peptide Antibody (Monoclonal, 8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # BTE0020802
  • Size 200 µl
Applications
ELISA (1:6,000)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
BTE0020802 600.00
Check your price
  • Procollagen I C-Peptide Antibody (Monoclonal)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # BTE0020402
  • Size 200 µl
Applications
ELISA (1/10,000)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
BTE0020402 600.00
Check your price
  • LAP (Latency Associated Peptide) Antibody (Monoclonal, 7F6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517186
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517186 309.00
Check your price
  • LAP (Latency Associated Peptide) Antibody (Monoclonal, 9005)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523701
  • Size 500 µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture))
Alternate names:
TGFB, TGFb-1, DPD1, Transforming Growth Factor, Transforming Growth Factor Beta-1, TGF-beta-1, CED
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523701 349.00
Check your price
  • GLP-1 Antibody (Monoclonal, 8G9)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB1470602
  • Size 200 µl
Applications
ELISA (1:8000), Immunofluorescence (1:500), Immunohistochemistry (1:200)
Alternate names:
Glucagon-like peptide-1, GLP-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1470602 600.00
Check your price
  • GLP-1 Antibody (Monoclonal, 10)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0331002
  • Size 200 µL
Applications
ELISA (1:20,000)
Alternate names:
Glucagon-like peptide-1, GLP-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0331002 650.00
Check your price
  • TAP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131487
  • Size 100 µg
Applications
ELISA (Assay-dependent), Flow Cytometry (1:10,000), Immunoprecipitation (1:10,000), Western Blot (1:1000)
Alternate names:
Really interesting new gene 4 protein; Peptide transporter involved in antigen processing 1; Peptide transporter TAP1; Peptide transporter PSF1; Peptide supply factor 1; ATP-binding cassette sub-family B member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131487 550.00
Check your price
  • GLP1R Antibody, Biotin (Monoclonal, 4F3)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # HYB14712B005
  • Size 50 µl
Applications
ELISA (1:8,000)
Alternate names:
Glucagon-like peptide-1 receptor, GLP-1 receptor, GLP-1-R, GLP-1R, GLP-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB14712B005 475.00
Check your price
  • GLP1R Antibody (Monoclonal, 4F3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB1471202
  • Size 200 µl
Applications
ELISA (1:8,000), Immunohistochemistry (Assay dependent), Western Blot (1:2,000)
Alternate names:
Glucagon-like peptide-1, GLP-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1471202 600.00
Check your price
  • BNP Antibody, HRP (Monoclonal, 24C5)
  • Host Mouse
  • Target Species Human
  • Conjugate HRP
  • Catalog # MA191673
  • Size 200 µg
Applications
ELISA (Assay-Dependent), Western Blot (Assay dependent)
Alternate names:
Gamma-brain natriuretic peptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191673 399.00
Check your price
  • C-Peptide Antibody (Monoclonal, C-PEP-01)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119159
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (2-5 µg/ml), Radioimmune Assays (Assay Dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119159 309.00
Check your price
  • TAP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA186830
  • Size 50 µg
Applications
ELISA (1:10000), Flow Cytometry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
Really interesting new gene 4 protein; Peptide transporter involved in antigen processing 1; Peptide transporter TAP1; Peptide transporter PSF1; Peptide supply factor 1; ATP-binding cassette sub-family B member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186830 375.00
Check your price
  • CGRP Antibody (Monoclonal, 5)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # ABS0260502
  • Size 200 µL
Applications
ELISA (1:2000), Immunohistochemistry (Paraffin) (1:400)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0260502 550.00
Check your price
  • CGRP Antibody (Monoclonal, 4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0260402
  • Size 200 µL
Applications
ELISA (1:8000), Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0260402 550.00
Check your price
  • CGRP Antibody (Monoclonal, 4901)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA120209
  • Size 50 µL
Applications
ELISA (0.5 µg/ml), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Neutralization (Assay dependent), Radioimmune Assays (Assay dependent)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120209 475.00
Check your price
  • GLP1R Antibody (Monoclonal, 11E2)
  • Host Mouse
  • Target Species Human, Mammal
  • Conjugate Unconjugated
  • Catalog # HYB1470802
  • Size 200 µL
Applications
ELISA (1:20,000), Western Blot (1:1,000)
Alternate names:
Glucagon-like peptide-1, GLP-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1470802 600.00
Check your price
  • PYY Antibody (Monoclonal, 1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0290102
  • Size 200 µL
Applications
ELISA (1:16,000), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0290102 600.00
Check your price