1 - 30 of 30 products displayed
  • CaMKII alpha Monoclonal Antibody for Western Blot, IF, IHC and IP
  • Host Mouse
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1048
  • Size 100 µL
Applications
Immunofluorescence (1:100), Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (0.5-1 µg/mL)
Alternate names:
Calcium/calmodulin-dependent protein kinase type 2, CaMKII
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1048 375.00
Check your price
  • Phospho-CaMKII alpha (Thr286) Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1047
  • Size 100 µL
Applications
ELISA (Assay dependent), Flow Cytometry (1µg per 10^6 cells), Immunocytochemistry (1:100), Immunofluorescence (1:50), Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type 2, Phospho-CaMKI
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1047 375.00
Check your price
  • Phospho-CaMKII alpha (Thr286) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA126727
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
CAMKA, KIAA0968
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126727 399.00
Check your price
  • CaMKII alpha Monoclonal Antibody for Western Blot, IHC, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Fish, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # 137300
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, calcium/calmodulin-dependent protein kinase type II alpha chain, Calci
Catalog # Size Price (AUD) Your Price (AUD) Qty
137300 304.00
Check your price
  • CaMKII alpha Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532289
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
CAMKA, CaM kinase II alpha subunit, CaM-kinase II alpha chain, CaMK-II alpha subunit, CaMKIINalpha, caM kinase II subunit alpha, caMK-II subunit alpha, calcium/calmodulin-dependent protein kinase CaM kinase II alpha, calcium/calmodulin-dependent protein kinase II alp
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532289 349.00
Check your price
  • CaMKII beta Monoclonal Antibody for Western Blot, IHC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Fish, Human, Mouse, Sheep, Rat, Sea urchin
  • Conjugate Unconjugated
  • Catalog # 139800
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II delta, calcium/calmodulin-dependent protein kinase II delta, calcium/calmodulin-dependent protein kinase type II delta chain, Calcium/calmodulin-dependent protein kinase type II subunit delta, Ca
Catalog # Size Price (AUD) Your Price (AUD) Qty
139800 304.00
Check your price
  • CaMKII alpha Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137226
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
CAMKA, KIAA0968
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137226 425.00
Check your price
  • CaMKII beta/gamma/delta Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536562
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II subunit beta, CaMK-II subunit beta, CAMK2B, CAM2, CAMK2, CAMKB, Calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CaMK-II subunit gamma, CAMK2G, CAMK, CAMK-II, CAMKG, Calcium/calmodulin-dependent protein kinase type II subunit delta, CaM kinase II subunit delta, CaMK-II subunit delta, CAMK2D, CAMKD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536562 309.00
Check your price
  • Phospho-CaMKII alpha/beta/delta (Thr305) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538238
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit alpha, CaM kinase II subunit alpha, CaMK-II subunit alpha, CAMK2A, CAMKA, KIAA0968, CaMK2-a, CaMK2a, CaMK-2a, Calcium/calmodulin-dependent protein kinase type II subunit delta, CaM kinase II subunit delta, CaMK-II subunit delta, CAMK2D, CAMKD, CaMK2- delta, CaMK2 delta, CaMK-2 delta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538238 309.00
Check your price
  • Phospho-CaMKII alpha/delta (Thr286) Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536838
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit alpha, CaM kinase II subunit alpha, CaMK-II subunit alpha, CAMK2A, CAMKA, KIAA0968, CaMK2-a, CaMK2a, CaMK-2a, Calcium/calmodulin-dependent protein kinase type II subunit delta, CaM kinase II subunit delta, CaMK-II subunit delta, CAMK2D, CAMKD, CaMK2- delta, CaMK2 delta, CaMK-2 delta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536838 349.00
Check your price
  • CaMKII alpha/beta/delta Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Rat, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538239
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit alpha, CaM kinase II subunit alpha, CaMK-II subunit alpha, CAMK2A, CAMKA, KIAA0968, CaMK2-a, CaMK2a, CaMK-2a, Calcium/calmodulin-dependent protein kinase type II subunit delta, CaM kinase II subunit delta, CaMK-II subunit delta, CAMK2D, CAMKD, CaMK2- delta, CaMK2 delta, CaMK-2 delta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538239 309.00
Check your price
  • Phospho-CaMKII beta/gamma/delta (Thr287) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539731
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II subunit beta, CaMK-II subunit beta, CAMK2B, CAM2, CAMK2, CAMKB, Calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CaMK-II subunit gamma, CAMK2G, CAMK, CAMK-II, CAMKG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539731 309.00
Check your price
  • CaMKII beta/gamma/delta Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539732
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II subunit beta, CaMK-II subunit beta, CAMK2B, CAM2, CAMK2, CAMKB, Calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CaMK-II subunit gamma, CAMK2G, CAMK, CAMK-II, CAMKG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539732 309.00
Check your price
  • CaMKII beta/gamma Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536195
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II subunit beta, CaMK-II subunit beta, CAMK2B, CAM2, CAMK2, CAMKB, Calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CaMK-II subunit gamma, CAMK2G, CAMK, CAMK-II, CAMKG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536195 309.00
Check your price
  • CaMKII gamma Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514035
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II gamma chain; CaM-kinase II gamma chain; CaM kinase II gamma subunit; CaMK-II gamma subunit; CAMK2G
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514035 349.00
Check your price
  • Phospho-CaMKII alpha/beta/delta (Thr305) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537832
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, calcium/calmodulin-dependent protein kinase type II alpha chain, Calci, calcium/calmodulin-dependent protein kinase (CaM kinase) II delta, calcium/calmodulin-dependent protein kinase II delta, calcium/calmodulin-dependent protein kinase type II delta chain, Calcium/calmodulin-dependent protein kinase type II subunit delta, Ca, CAMKD, CaM kinase II delta subunit, CaM-kinase II delta chain, CaMK-II delta subunit, caM kinase II subunit delta, caMK-II subunit delta, calcium/calmodulin-dependent protein kinase (CaM kinase) II delta, calcium/calmodulin-dependent protein kinase type II delta chain, calcium/calmodulin-dependent protein kinase type II subunit delta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537832 309.00
Check your price
  • Phospho-CaMKII beta/gamma/delta (Thr287) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537833
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II delta, calcium/calmodulin-dependent protein kinase II delta, calcium/calmodulin-dependent protein kinase type II delta chain, Calcium/calmodulin-dependent protein kinase type II subunit delta, Ca, CAMKD, CaM kinase II delta subunit, CaM-kinase II delta chain, CaMK-II delta subunit, caM kinase II subunit delta, caMK-II subunit delta, calcium/calmodulin-dependent protein kinase (CaM kinase) II delta, calcium/calmodulin-dependent protein kinase type II delta chain, calcium/calmodulin-dependent protein kinase type II subunit delta, CAMK, CAMK-II, CAMKG, caMK-II subunit gamma, calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma, calcium/calmodulin-dependent protein kinase type II subunit gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537833 309.00
Check your price
  • CaMKII gamma Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529648
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
CAMK, CAMK-II, CAMKG, caMK-II subunit gamma, calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma, calcium/calmodulin-dependent protein kinase type II subunit gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529648 349.00
Check your price
  • CaMKII gamma Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528789
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
CAMK, CAMK-II, CAMKG, caMK-II subunit gamma, calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma, calcium/calmodulin-dependent protein kinase type II subunit gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528789 349.00
Check your price
  • Phospho-CaMKII alpha/beta (Thr286) Polyclonal Antibody for Western Blot and DB
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14614
  • Size 100 µL
Applications
Dot blot (1:1000), Western Blot (1:1000)
Alternate names:
phospho-Ca2+/Calmodulin-Dependent Protein Kinase II (Thr286)
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14614 375.00
Check your price
  • Phospho-CaMKII alpha (Thr286) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # 44674G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, calcium/calmodulin-dependent protein kinase type II alpha chain, Calci
Catalog # Size Price (AUD) Your Price (AUD) Qty
44674G 309.00
Check your price
  • CaMKII alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517095
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
CaM kinase II alpha subunit; CaM-kinase II alpha chain; CaMK-II alpha subunit; CaMKIINalpha; caM kinase II subunit alpha; CaMK-II subunit alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517095 275.00
Check your price
  • CaMKII delta Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA514036
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
CaM-kinase II delta chain; CaM kinase II delta subunit; CaMK-II delta subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514036 349.00
Check your price
  • CaMKII alpha Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A16650
  • Size 100 µL
Applications
Western Blot (Assay Dependent)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, calcium/calmodulin-dependent protein kinase type II alpha chain, Calci
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16650 309.00
Check your price
  • CaMKII alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # A14012
  • Size 100 µL
Applications
Western Blot (1:1,000)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, calcium/calmodulin-dependent protein kinase type II alpha chain, Calci
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14012 309.00
Check your price
  • CaMKII alpha Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514033
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II alpha chain; CaM-kinase II alpha chain; CaM kinase II alpha subunit; CaMK-II alpha subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514033 349.00
Check your price
  • CaMKII delta Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518532
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
CAMK2B, CAMK2D, CAMK2G, Calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II subunit beta, CaMK-II subunit beta, CAMK2B, CAM2, CAMK2, CAMKB, Calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CaMK-II subunit gamma, CAMK2G, CAMKG, Calcium/calmodulin-dependent protein kinase type II subunitdelta, CaM kinase II subunit delta, CaMK-II subunit delta, CAMK2D, CAMKD
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518532 309.00
Check your price
  • CaMKII beta Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141150
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (Assay-Dependent)
Alternate names:
CAM2, CAMK2, CAMKB, MGC29528
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141150 375.00
Check your price
  • CaMKII alpha Polyclonal Antibody for Western Blot, IHC (P) and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519128
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
CAMK2A; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; CAMKA; KIAA0968; CaM kinase II alpha subunit; CaM-kinase II alpha chain; CaMK-II alpha subunit; OTTHUMP00000165787; OTTHUMP00000165788; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase IIA; calcium/calmodulin-dependent protein kinase type II alpha chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519128 349.00
Check your price
  • CaMKII beta Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529327
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
CAM2, CAMK2, CAMKB, CaM kinase II beta subunit, CaM-kinase II beta chain, caMK-II subunit beta, calcium/calmodulin-dependent protein kinase type II subunit beta, proline rich calmodulin-dependent protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529327 349.00
Check your price