1 - 30 of 30 products displayed
  • Caspase 3 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA516335
  • Size 500µL
Applications
Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (5-10 ug/mL), Immunoprecipitation (10 ug/mg protein lysate), Western Blot (2.5-5 ug/ml)
Alternate names:
Cysteine protease CPP32; Apopain; SREBP cleavage activity 1; Protein Yama; LICE; IRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516335 349.00
Check your price
  • Caspase 3 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Horse, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # 437800
  • Size 100µg
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay Dependent), Western Blot (2mg/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; LICE; IRP; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
437800 304.00
Check your price
  • Caspase 3 Polyclonal Antibody, Biotin conjugate for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # PA516332
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/mL), Immunoprecipitation (10 µg/ml), Western Blot (2.5-5.0 µg/ml)
Alternate names:
Cysteine protease CPP32; Apopain; SREBP cleavage activity 1; Protein Yama; LICE; IRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516332 399.00
Check your price
  • pro-Caspase 3 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA191636
  • Size 100µg
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (2 µg/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain; LICE
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191636 449.00
Check your price
  • Phospho-Caspase 3 (Ser150) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536746
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
SREBP cleavage activity 1; Protein Yama; LICE; IRP; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536746 349.00
Check your price
  • Phospho-Caspase 3 (Ser150) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538438
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
SREBP cleavage activity 1; Protein Yama; LICE; IRP; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538438 309.00
Check your price
  • Caspase 3 Monoclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA116843
  • Size 200µL
Applications
Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
SREBP cleavage activity 1; Protein Yama; LICE; IRP; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116843 349.00
Check your price
  • Caspase 3 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA129157
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:1000), Immunoprecipitation (1:2000), Western Blot (1:500)
Alternate names:
Cysteine protease CPP32; Apopain; SREBP cleavage activity 1; Protein Yama; LICE; IRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129157 399.00
Check your price
  • Caspase 3 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 700182
  • Size 100µg
Applications
Flow Cytometry (0.5-1 ug/test), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (0.1-0.2 ug/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain; LICE
Catalog # Size Price (AUD) Your Price (AUD) Qty
700182 309.00
Check your price
  • Caspase 3 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Ferret, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA126426
  • Size 50µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (10-20 µg/ml), Immunohistochemistry (Paraffin) (10-20 µg/ml), Western Blot (1 µg/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; LICE; IRP; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126426 475.00
Check your price
  • Caspase 3 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511521
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (2µg/mg protein lysate), Western Blot (1 µg/mL)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511521 349.00
Check your price
  • Caspase 3 (active/pro) Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191637
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (2-3 µg/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191637 425.00
Check your price
  • Caspase 3 Monoclonal Antibody for Western Blot, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511516
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunohistochemistry (Paraffin) (1:50-1:100), Immunoprecipitation (1:25-1:50), Western Blot (1:25-1:50)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511516 349.00
Check your price
  • Caspase 3 Polyclonal Antibody for IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534413
  • Size 100ug
Applications
Immunohistochemistry (5 ug/mL)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534413 349.00
Check your price
  • Caspase 3 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 351600Z
  • Size 100µg
Applications
Immunohistochemistry (Assay Dependent), Western Blot (1-2 µg/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
351600Z 305.00
Check your price
  • pro-Caspase 3 Monoclonal Antibody for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA141163
  • Size 100µg
Applications
Western Blot (2 µg/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA141163 309.00
Check your price
  • Caspase 3 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514504
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514504 349.00
Check your price
  • Phospho-Caspase 3 (Ser150) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540260
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540260 309.00
Check your price
  • Caspase 3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Rabbit
  • Conjugate Unconjugated
  • Catalog # PA133035
  • Size 50µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:500), Western Blot (1:1000)
Alternate names:
Cysteine protease CPP32; Apopain; SREBP cleavage activity 1; Protein Yama
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA133035 449.00
Check your price
  • Caspase 3 Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710431
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:100-1:1000)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
710431 309.00
Check your price
  • Caspase 3 Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126415
  • Size 500µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:25-1:75), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126415 475.00
Check your price
  • Caspase 3 Monoclonal Antibody for Western Blot, IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125975
  • Size 250µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (2 µg/mg of protein lysate), Western Blot (1 µg/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125975 375.00
Check your price
  • Caspase 3 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523921
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523921 349.00
Check your price
  • Caspase 7 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA141281
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Cysteine protease LICE2; Apoptotic protease Mch-3; ICE-like apoptotic protease 3; CMH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA141281 309.00
Check your price
  • Caspase 7 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520114
  • Size 100ug
Applications
Immunohistochemistry (2 ug/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
Cysteine protease LICE2; Apoptotic protease Mch-3; ICE-like apoptotic protease 3; CMH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520114 349.00
Check your price
  • Caspase 7 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520115
  • Size 100ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
Cysteine protease LICE2; Apoptotic protease Mch-3; ICE-like apoptotic protease 3; CMH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520115 349.00
Check your price
  • Caspase 9 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519904
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (20 ug/ml), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519904 349.00
Check your price
  • Caspase 9 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519903
  • Size 100ug
Applications
Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519903 349.00
Check your price
  • Caspase 7 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA191896
  • Size 100µL
Applications
Western Blot (Assay dependent)
Alternate names:
Cysteine protease LICE2; Apoptotic protease Mch-3; ICE-like apoptotic protease 3; CMH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191896 399.00
Check your price
  • Caspase 9 Polyclonal Antibody, Biotin conjugate for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Sheep, Rat
  • Conjugate Biotin
  • Catalog # PA516355
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/mL), Immunoprecipitation (10 µg/ml), Western Blot (2.5-5.0 µg/ml)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516355 399.00
Check your price