1 - 30 of 30 products displayed
  • Apoptosis-Enhancing Nuclease Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522208
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Interferon-stimulated 20 kDa exonuclease-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522208 349.00
Check your price
  • PDCD5 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520684
  • Size 100ug
Applications
Immunocytochemistry (5 µg/ml), Western Blot (2.5 µg/ml)
Alternate names:
TF-1 cell apoptosis-related protein 19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520684 349.00
Check your price
  • PDCD5 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520682
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1 ug/ml)
Alternate names:
TF-1 cell apoptosis-related protein 19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520682 349.00
Check your price
  • PDCD5 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528998
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
TF-1 cell apoptosis-related protein 19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528998 349.00
Check your price
  • PDCD5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA526638
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
TF-1 cell apoptosis-related protein 19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526638 349.00
Check your price
  • CD137 Polyclonal Antibody for Western Blot, IF, ICC, Flow, ELISA and FN
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547037
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Functional Assay (Assay-Dependent), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
T-cell antigen ILA; T-cell antigen 4-1BB homolog; CDw137; 4-1BB ligand receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547037 399.00
Check your price
  • DR3 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA523838
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Protein WSL-1; Protein WSL; Lymphocyte-associated receptor of death; Death receptor 3; Apoptosis-mediating receptor TRAMP; Apoptosis-mediating receptor DR3; Apoptosis-inducing receptor AIR; Apo-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523838 275.00
Check your price
  • CD27 Polyclonal Antibody for Western Blot, Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124876
  • Size 25µg
Applications
ELISA (0.5-1 µg/ml), Flow Cytometry (0.5-1 µg for 10^6 cells), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Tumor necrosis factor receptor superfamily member 7; T14; T-cell activation antigen CD27; CD27L receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124876 349.00
Check your price
  • FLT3 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA523600
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Stem cell tyrosine kinase 1; Fms-like tyrosine kinase 3; Fetal liver kinase-2; FL cytokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523600 289.00
Check your price
  • FLT3 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523562
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Stem cell tyrosine kinase 1; Fms-like tyrosine kinase 3; Fetal liver kinase-2; FL cytokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523562 249.00
Check your price
  • TNFRSF13C Polyclonal Antibody for Western Blot, Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546904
  • Size 100µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Neutralization (3-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
BLyS receptor 3; BAFF receptor; B-cell-activating factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546904 375.00
Check your price
  • FAS Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA110313
  • Size 100Tests
Applications
Flow Cytometry (10 ul/10^6 cells)
Alternate names:
FASLG receptor; Apoptosis-mediating surface antigen FAS; Apo-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110313 275.00
Check your price
  • CD99 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA110315
  • Size 100Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
T-cell surface glycoprotein E2; Protein MIC2; E2 antigen; 120000000
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110315 309.00
Check your price
  • c-Kit Monoclonal Antibody for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA170079
  • Size 250ug
Applications
Flow Cytometry (0.1-0.2 µg/10^6 cells)
Alternate names:
Tyrosine-protein kinase Kit; Proto-oncogene c-Kit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170079 309.00
Check your price
  • CD137 Polyclonal Antibody for Western Blot, Flow and FN
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547967
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Functional Assay (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
T-cell antigen 4-1BB; 4-1BB ligand receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547967 349.00
Check your price
  • Ku80 Monoclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA512933
  • Size 500µL
Applications
Flow Cytometry (0.5 ug/test), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunoprecipitation (2 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Nuclear factor IV; Ku80; Ku autoantigen protein p86 homolog; DNA repair protein XRCC5; CTC box-binding factor 85 kDa subunit; ATP-dependent DNA helicase II 80 kDa subunit; ATP-dependent DNA helicase 2 subunit 2; X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining); Thyroid-lupus autoantigen; Lupus Ku autoantigen protein p86; Ku86; 86 kDa subunit of Ku antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512933 309.00
Check your price
  • Ku80 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, Flow and IP
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA512930
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining); Thyroid-lupus autoantigen; Nuclear factor IV; Lupus Ku autoantigen protein p86; Ku86; Ku80; DNA repair protein XRCC5; CTC box-binding factor 85 kDa subunit; ATP-dependent DNA helicase II 80 kDa subunit; ATP-dependent DNA helicase 2 subunit 2; 86 kDa subunit of Ku antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512930 375.00
Check your price
  • FLT3 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA110090
  • Size 100Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Stem cell tyrosine kinase 1; Fms-like tyrosine kinase 3; Fetal liver kinase-2; FL cytokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110090 275.00
Check your price
  • PAR4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532564
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Prostate apoptosis response 4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532564 275.00
Check your price
  • BCMA Polyclonal Antibody for Western Blot, Flow, ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547224
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
B-cell maturation protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547224 349.00
Check your price
  • HEF1 Monoclonal Antibody for Western Blot, IF, IHC, IHC (PFA) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA15785
  • Size 100µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunohistochemistry (PFA fixed) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p105; Renal carcinoma antigen NY-REN-12; Neural precursor cell expressed developmentally down-regulated protein 9; Cas scaffolding protein family member 2; CRK-associated substrate-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15785 550.00
Check your price
  • Granzyme B Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA110338
  • Size 100ug
Applications
Flow Cytometry (2-8 ug/ml)
Alternate names:
T-cell serine protease 1-3E; SECT; Human lymphocyte protein; Granzyme-2; Fragmentin-2; Cytotoxic T-lymphocyte proteinase 2; Cathepsin G-like 1; CTLA-1; C11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110338 225.00
Check your price
  • IL-3 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523797
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.05-0.15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
P-cell-stimulating factor; Multipotential colony-stimulating factor; Mast cell growth factor; Hematopoietic growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523797 349.00
Check your price
  • FLT3 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA523648
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Tyrosine-protein kinase receptor flk-2; Fms-like tyrosine kinase 3; Fetal liver kinase 2; FL cytokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523648 339.00
Check your price
  • FLT3 Monoclonal Antibody, APC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # MA523631
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Tyrosine-protein kinase receptor flk-2; Fms-like tyrosine kinase 3; Fetal liver kinase 2; FL cytokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523631 309.00
Check your price
  • FLT3 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA128018
  • Size 25µg
Applications
ELISA (0.5-1 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Stem cell tyrosine kinase 1; Fms-like tyrosine kinase 3; Fetal liver kinase-2; FL cytokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128018 349.00
Check your price
  • OX40 Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA523610
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
TAX transcriptionally-activated glycoprotein 1 receptor; OX40L receptor; ACT35 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523610 289.00
Check your price
  • OX40 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523676
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
TAX transcriptionally-activated glycoprotein 1 receptor; OX40L receptor; ACT35 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523676 349.00
Check your price
  • OX40 Polyclonal Antibody for Western Blot and Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547483
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
TAX transcriptionally-activated glycoprotein 1 receptor; OX40L receptor; ACT35 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547483 349.00
Check your price
  • PD-1 Monoclonal Antibody for IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516934
  • Size 250ug
Applications
Flow Cytometry (1:10-1:25), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
CD279, PD-1, PD1, SLEB2, hPD-1, hPD-l, programmed cell death protein 1, protein PD-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516934 375.00
Check your price