1 - 30 of 30 products displayed
  • EpCAM Antibody (Monoclonal, VU-1D9)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516973
  • Size 500 ug
Applications
Flow Cytometry (1:10-1:200), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516973 499.00
Check your price
  • NFkB p50 Antibody (Monoclonal, 5D10D11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515870
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1; EBP-1; DNA-binding factor KBF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515870 349.00
Check your price
  • DC-SIGN Antibody (Monoclonal, MR-1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516866
  • Size 500 ug
Applications
Flow Cytometry (Assay Dependent), Functional Assay (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Dendritic cell-specific ICAM-3-grabbing non-integrin 1; C-type lectin domain family 4 member L
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516866 550.00
Check your price
  • Dectin 1 (soluble) Antibody (Monoclonal, 2A11)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516478
  • Size 500 ug
Applications
Flow Cytometry (1:10 -1:50), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
BGR, Clecsf12, beta-GR,C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 12; C-type lectin domain family 7 member A; C-type lectin superfamily member 12; DC-associated C-type lectin 1; beta-glucan receptor; dectin-1; dendritic cell-associated C-type lectin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516478 600.00
Check your price
  • Dectin 1 Antibody (Monoclonal, GE2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516692
  • Size 500 ug
Applications
Flow Cytometry (Neat-1:25), Functional Assay (Assay Dependent)
Alternate names:
Dendritic cell-associated C-type lectin 1; C-type lectin superfamily member 12; Beta-glucan receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516692 499.00
Check your price
  • NFkB p50 Antibody (Monoclonal, 5D10)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515860
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1; EBP-1; DNA-binding factor KBF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515860 349.00
Check your price
  • CD4 Antibody (Monoclonal, YTS177.9)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA517007
  • Size 500 ug
Applications
Flow Cytometry (1:50-1:200), Functional Assay (Assay Dependent)
Alternate names:
T-cell surface antigen T4/Leu-3; T-cell differentiation antigen L3T4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517007 449.00
Check your price
  • eNOS Antibody (Monoclonal, 6H2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515559
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000)
Alternate names:
NOS type III; Endothelial NOS; EC-NOS; Constitutive NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515559 349.00
Check your price
  • SIGN-R1 Antibody (Monoclonal, ER-TR9)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516668
  • Size 500 ug
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (1:25-1:50)
Alternate names:
OtB7; DC-SIGN-related protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516668 650.00
Check your price
  • PD-L1 Antibody (Monoclonal, MIH6)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516847
  • Size 500 ug
Applications
Flow Cytometry (Neat-1:200), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
B7 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516847 499.00
Check your price
  • PCNA Antibody (Monoclonal, 7H4F8)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA517145
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Cyclin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517145 309.00
Check your price
  • CD28 Antibody (Monoclonal, YTH913.12)
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517005
  • Size 500 ug
Applications
Flow Cytometry (1:50-1:100), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
TP44
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517005 550.00
Check your price
  • TLR4 Antibody (Monoclonal, HTA125)
  • Host Mouse
  • Target Species Dog, Guinea pig, Human, Non-human primate, Pig
  • Conjugate Unconjugated
  • Catalog # MA180122
  • Size 500 µg
Applications
Flow Cytometry (1:10-1:50), Functional Assay (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
hToll
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180122 499.00
Check your price
  • NFkB p65 Antibody (Monoclonal, 4-2H22L23)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701079
  • Size 100 µg
Applications
ChIP assay (1-5 µg x 10^6 cells), Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1-5 ug/ml)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; Nuclear factor NF-kappa-B p65 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
701079 309.00
Check your price
  • PRDX1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA3750
  • Size 100 µL
Applications
Immunofluorescence (1:100), Immunohistochemistry (Assay-dependent), Western Blot (1:1,000)
Alternate names:
Thioredoxin-dependent peroxide reductase 2; Thioredoxin peroxidase 2; Proliferation-associated gene protein; Natural killer cell-enhancing factor A; Heme-binding 23 kDa protein; HBP23; Osteoblast-specific factor 3; Macrophage 23 kDa stress protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3750 375.00
Check your price
  • Phospho-NFkB p52 (Ser866) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537664
  • Size 100 µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2; DNA-binding factor KBF2; Oncogene Lyt-10; Lymphocyte translocation chromosome 10 protein; H2TF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537664 309.00
Check your price
  • Phospho-NFkB p52 (Ser870) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537663
  • Size 100 µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2; DNA-binding factor KBF2; Oncogene Lyt-10; Lymphocyte translocation chromosome 10 protein; H2TF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537663 309.00
Check your price
  • PRDX2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA3751
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunohistochemistry (Assay-dependent), Western Blot (1:10,000)
Alternate names:
Thioredoxin-dependent peroxide reductase 1; Thioredoxin peroxidase 1; Thiol-specific antioxidant protein; PRP; Natural killer cell-enhancing factor B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3751 349.00
Check your price
  • CD244 Antibody (Monoclonal, 2B4.69)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516486
  • Size 500 ug
Applications
Flow Cytometry (1:50-1:200), Immunoprecipitation (Assay Dependent)
Alternate names:
Signaling lymphocytic activation molecule 4; SLAM family member 4; NK cell type I receptor protein 2B4; NK cell activation-inducing ligand
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516486 475.00
Check your price
  • NFkB p65 Antibody (Oligoclonal, 4-2HCLC)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710048
  • Size 100 µg
Applications
ChIP assay (3µg), ELISA (Assay-Dependent), Immunocytochemistry (1µg/ml), Immunofluorescence (1µg/ml), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (0.5-1µg/ml)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; Nuclear factor NF-kappa-B p65 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
710048 305.00
Check your price
  • CD3e Antibody (Monoclonal, UCHT1)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA180537
  • Size 500 µg
Applications
Flow Cytometry (1/50-1/100), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180537 499.00
Check your price
  • NFkB p52 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527340
  • Size 100 µL
Applications
Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2; DNA-binding factor KBF2; Oncogene Lyt-10; Lymphocyte translocation chromosome 10 protein; H2TF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527340 349.00
Check your price
  • CD11b Antibody (Monoclonal, 5C6)
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516528
  • Size 500 ug
Applications
Flow Cytometry (1:50-1:100), Functional Assay (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
Neutrophil adherence receptor; Leukocyte adhesion receptor MO1; Cell surface glycoprotein MAC-1 subunit alpha; CR-3 alpha chain; CD11 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516528 425.00
Check your price
  • NFkB p50 Antibody (Monoclonal, 13A2F)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0236
  • Size 100 µL
Applications
ELISA (Assay Dependent), Western Blot (0.5 µg/ml)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1; NF-kappa-B1 p84/NF-kappa-B1 p98; EBP-1; DNA-binding factor KBF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0236 349.00
Check your price
  • NFkB p50 Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mink, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA517150
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1; NF-kappa-B1 p84/NF-kappa-B1 p98; EBP-1; DNA-binding factor KBF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517150 275.00
Check your price
  • Human CCL5 (RANTES) Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RRNTS20
  • Size 20 ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
T-cell-specific protein RANTES; T cell-specific protein P228; Small-inducible cytokine A5; SIS-delta; Eosinophil chemotactic cytokine; EoCP
Catalog # Size Price (AUD) Your Price (AUD) Qty
RRNTS20 304.00
Check your price
  • Mouse IL-3 Recombinant Protein
  • Host E. Coli
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # RMIL310
  • Size 10 ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
P-cell-stimulating factor; Multipotential colony-stimulating factor; Mast cell growth factor; Hematopoietic growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
RMIL310 265.00
Check your price
  • NFkB p50 Antibody
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA517654
  • Size 100 µL
Applications
ChIP assay (1:25), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1; EBP-1; DNA-binding factor KBF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517654 275.00
Check your price
  • Dectin 2 Antibody (Monoclonal, D2.11E4)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA182675
  • Size 500 µg
Applications
Flow Cytometry (1:10-1:100), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
Dendritic cell-associated C-type lectin 2; C-type lectin superfamily member 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182675 499.00
Check your price
  • CD4 Antibody (Monoclonal, YTS191.1)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA517008
  • Size 500 ug
Applications
Flow Cytometry (1:100 -1:200), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
T-cell surface antigen T4/Leu-3; T-cell differentiation antigen L3T4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517008 499.00
Check your price