1 - 30 of 30 products displayed
  • EpCAM Monoclonal Antibody for IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512153
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512153 309.00
Check your price
  • CD2 Monoclonal Antibody, Biotin conjugate for IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA517484
  • Size 100ug
Applications
Flow Cytometry (2.0 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
T-cell surface antigen T11/Leu-5; OX-34 antigen; LFA-3 receptor; LFA-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517484 159.00
Check your price
  • CD2 Monoclonal Antibody, Biotin conjugate for IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA517485
  • Size 500ug
Applications
Flow Cytometry (2.0 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
T-cell surface antigen T11/Leu-5; OX-34 antigen; LFA-3 receptor; LFA-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517485 309.00
Check your price
  • CD2 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # MA517486
  • Size 100ug
Applications
Flow Cytometry (0.1 ug/10^6 cells)
Alternate names:
T-cell surface antigen T11/Leu-5; OX-34 antigen; LFA-3 receptor; LFA-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517486 159.00
Check your price
  • CD2 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # MA517487
  • Size 500ug
Applications
Flow Cytometry (0.1 ug/10^6 cells)
Alternate names:
T-cell surface antigen T11/Leu-5; OX-34 antigen; LFA-3 receptor; LFA-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517487 425.00
Check your price
  • CD2 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate PE
  • Catalog # MA517488
  • Size 50ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
T-cell surface antigen T11/Leu-5; OX-34 antigen; LFA-3 receptor; LFA-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517488 159.00
Check your price
  • CD2 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate PE
  • Catalog # MA517489
  • Size 200ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
T-cell surface antigen T11/Leu-5; OX-34 antigen; LFA-3 receptor; LFA-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517489 399.00
Check your price
  • EpCAM Monoclonal Antibody for IF, ICC, IHC (P), Flow and RIA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA512436
  • Size 500µL
Applications
Flow Cytometry (0.5-1 ug/test), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1-2 µg/ml), Radioimmune Assays (Assay Dependent)
Alternate names:
Tumor-associated calcium signal transducer 1; Protein 289A; Epithelial glycoprotein 314; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512436 309.00
Check your price
  • EpCAM Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA513917
  • Size 500µL
Applications
Flow Cytometry (1ug/test), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1:50-1:500)
Alternate names:
Tumor-associated calcium signal transducer 1; Protein 289A; Epithelial glycoprotein 314; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513917 309.00
Check your price
  • EpCAM Monoclonal Antibody, Biotin conjugate for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA512150
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512150 375.00
Check your price
  • EpCAM Monoclonal Antibody for IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512604
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (1:10)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512604 309.00
Check your price
  • TROP2 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547030
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Pancreatic carcinoma marker protein GA733-1; Membrane component chromosome 1 surface marker 1; Cell surface glycoprotein Trop-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547030 375.00
Check your price
  • CD22 Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524023
  • Size 100µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
T-cell surface antigen Leu-14; Sialic acid-binding Ig-like lectin 2; B-lymphocyte cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524023 275.00
Check your price
  • TROP2 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547074
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cell surface glycoprotein Trop-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547074 375.00
Check your price
  • EpCAM Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA110197
  • Size 100Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110197 249.00
Check your price
  • EpCAM Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA110198
  • Size 100Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110198 275.00
Check your price
  • EpCAM Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA146104
  • Size 100µl
Applications
ELISA (1:100-1:2000), Flow Cytometry (1 µg/10^6 cells), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (2 µg/ml)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146104 309.00
Check your price
  • CD22 Polyclonal Antibody for Western Blot, IF, ICC and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547292
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (0.1-0.5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
T-cell surface antigen Leu-14; Sialic acid-binding Ig-like lectin 2; B-lymphocyte cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547292 349.00
Check your price
  • CD105 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523894
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Immunohistochemistry (Frozen) (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Cell surface MJ7/18 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523894 275.00
Check your price
  • CD105 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546971
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.5-5 µg/ml)
Alternate names:
Cell surface MJ7/18 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546971 399.00
Check your price
  • EpCAM Monoclonal Antibody for Western Blot, IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516973
  • Size 500ug
Applications
Flow Cytometry (1:10-1:200), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516973 499.00
Check your price
  • CD7/CD13/CD33 Monoclonal Antibody, FITC, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA112225
  • Size 100Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
gp150; Myeloid plasma membrane glycoprotein CD13; Microsomal aminopeptidase; Aminopeptidase M; Alanyl aminopeptidase; gp67; Sialic acid-binding Ig-like lectin 3; TP41; T-cell surface antigen Leu-9; T-cell leukemia antigen; GP40
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112225 375.00
Check your price
  • CD7/CD13/CD33 Monoclonal Antibody, FITC, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA112226
  • Size 50Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
gp150; Myeloid plasma membrane glycoprotein CD13; Microsomal aminopeptidase; Aminopeptidase M; Alanyl aminopeptidase; gp67; Sialic acid-binding Ig-like lectin 3; TP41; T-cell surface antigen Leu-9; T-cell leukemia antigen; GP40
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112226 309.00
Check your price
  • CD96 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524280
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells)
Alternate names:
T cell-activated increased late expression protein; Cell surface antigen CD96
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524280 275.00
Check your price
  • CD96 Monoclonal Antibody for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524281
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells)
Alternate names:
T cell-activated increased late expression protein; Cell surface antigen CD96
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524281 275.00
Check your price
  • CD2 Monoclonal Antibody for IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA517483
  • Size 500ug
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
T-cell surface antigen T11/Leu-5; OX-34 antigen; LFA-3 receptor; LFA-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517483 309.00
Check your price
  • EpCAM Monoclonal Antibody for Western Blot, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110196
  • Size 100ug
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1-10 ug/ml), Immunohistochemistry (Paraffin) (1-10 ug/ml), Immunoprecipitation (1-4 ug/ml), Western Blot (1-4 ug/ml)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110196 159.00
Check your price
  • EpCAM Monoclonal Antibody for Western Blot, ICC, IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110195
  • Size 100ug
Applications
Flow Cytometry (1-3 ug/ml), Immunocytochemistry (4 ug/ml), Immunohistochemistry (Frozen) (5-10 ug/ml), Immunohistochemistry (Paraffin) (5-10 ug/ml), Immunoprecipitation (1-2 ug/ml), Western Blot (1-4 ug/ml)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110195 159.00
Check your price
  • EpCAM Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512442
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512442 309.00
Check your price
  • CD7/C13 Monoclonal Antibody, FITC, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA112283
  • Size 100Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
gp150; Myeloid plasma membrane glycoprotein CD13; Microsomal aminopeptidase; Aminopeptidase M; Alanyl aminopeptidase; TP41; T-cell surface antigen Leu-9; T-cell leukemia antigen; GP40
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112283 375.00
Check your price