1 - 17 of 17 products displayed
  • CCRD6 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511867
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Chemokine-binding protein D6; Chemokine-binding protein 2; Chemokine receptor CCR-9; Chemokine receptor CCR-10; C-C chemokine receptor D6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511867 349.00
Check your price
  • CCRD6 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511866
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Chemokine-binding protein D6; Chemokine-binding protein 2; Chemokine receptor CCR-9; Chemokine receptor CCR-10; C-C chemokine receptor D6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511866 349.00
Check your price
  • CCRD6 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA121614
  • Size 100 µg
Applications
ELISA (1:100,000), Flow Cytometry (1-3 µl for 10^6 cells in 100 µl), Immunohistochemistry (Paraffin) (1:250), Western Blot (1:1000)
Alternate names:
Chemokine-binding protein D6; Chemokine-binding protein 2; Chemokine receptor CCR-9; Chemokine receptor CCR-10; C-C chemokine receptor D6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121614 399.00
Check your price
  • CCRD6 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533388
  • Size 50 ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (24 µg/ml)
Alternate names:
Chemokine-binding protein D6; Chemokine-binding protein 2; Chemokine receptor CCR-9; Chemokine receptor CCR-10; C-C chemokine receptor D6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533388 349.00
Check your price
  • CCRD6 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533387
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (20 µg/ml)
Alternate names:
Chemokine-binding protein D6; Chemokine-binding protein 2; Chemokine receptor CCR-9; Chemokine receptor CCR-10; C-C chemokine receptor D6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533387 349.00
Check your price
  • CCR9 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA121618
  • Size 100 µg
Applications
ELISA (1:100,000), Flow Cytometry (1:200), Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (1:250), Western Blot (1:1000)
Alternate names:
Chemokine C-C receptor 10; GPR-9-6; G-protein coupled receptor 28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121618 425.00
Check your price
  • CCR9 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533407
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (2-4 µg/ml)
Alternate names:
GPR-9-6; G-protein coupled receptor 28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533407 349.00
Check your price
  • CCR9 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533405
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (1.5 µg/ml)
Alternate names:
GPR-9-6; G-protein coupled receptor 28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533405 349.00
Check your price
  • CCR9 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533406
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (2-3 µg/ml)
Alternate names:
GPR-9-6; G-protein coupled receptor 28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533406 349.00
Check your price
  • CCR10 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA121617
  • Size 100 µg
Applications
ELISA (1:100,000), Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (1:250), Western Blot (1:1000)
Alternate names:
G-protein coupled receptor 2; Chemokine C-C receptor 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121617 475.00
Check your price
  • CCL21 Antibody (Monoclonal, 54111)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523751
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine A21; Secondary lymphoid-tissue chemokine; Beta-chemokine exodus-2; 6Ckine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523751 349.00
Check your price
  • Eotaxin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519868
  • Size 100 ug
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine A11; Eosinophil chemotactic protein; C-C motif chemokine 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519868 349.00
Check your price
  • CCL21 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523643
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Small-inducible cytokine A21; Secondary lymphoid-tissue chemokine; Beta-chemokine exodus-2; 6Ckine; Thymus-derived chemotactic agent 4; Small-inducible cytokine A21a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523643 349.00
Check your price
  • MIP-3 beta Antibody (Monoclonal, 54909)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523833
  • Size 100 µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Immunohistochemistry (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine A19; Macrophage inflammatory protein 3 beta; Epstein-Barr virus-induced molecule 1 ligand chemokine; CK beta-11; Beta-chemokine exodus-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523833 275.00
Check your price
  • MIP-3 beta Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546940
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Neutralization (2-10 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine A19; Macrophage inflammatory protein 3 beta; Epstein-Barr virus-induced molecule 1 ligand chemokine; CK beta-11; Beta-chemokine exodus-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546940 375.00
Check your price
  • MIP-3 beta Antibody (Monoclonal, 87102)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523845
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (3-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Small inducible cytokine A19, Macrophage inflammatory protein 3 beta, MIP-3-beta, EBI1-ligand chemokine, ELC, Beta chemokine exodus-3, CK beta-11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523845 275.00
Check your price
  • MIP-3 beta Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547958
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small inducible cytokine A19, Macrophage inflammatory protein 3 beta, MIP-3-beta, EBI1-ligand chemokine, ELC, Beta chemokine exodus-3, CK beta-11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547958 349.00
Check your price