1 - 30 of 30 products displayed
  • Donkey anti-Chicken IgY Secondary Antibody, HRP conjugate for Western Blot and ELISA
  • Host Donkey
  • Target Species Chicken
  • Conjugate HRP
  • Catalog # SA172004
  • Size 500µg
Applications
ELISA (1:15000), Western Blot (1:7500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172004 149.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Goat
  • Target Species Chicken
  • Conjugate HRP
  • Catalog # PA128798
  • Size 100µg
Applications
ELISA (1:10,000-1:50,000), Immunohistochemistry (Paraffin, Frozen) (1:500-1:2500), Immunomicroscopy (Assay Dependent), Western Blot (1:1000-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128798 168.00
Check your price
  • Bovine anti-Chicken IgY Secondary Antibody, Biotin conjugate for Western Blot, ICC, IHC and ELISA
  • Host Bovine
  • Target Species Chicken
  • Conjugate Biotin
  • Catalog # SA19505
  • Size 500µg
Applications
ELISA (1:1000-1:5000), Immunocytochemistry (1:100-1:1000), Immunohistochemistry (1:100-1:1000), Western Blot (1:1000-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA19505 177.00
Check your price
  • Bovine anti-Chicken IgY beads Secondary Antibody, Agarose conjugate for IP
  • Host Bovine
  • Target Species Chicken
  • Conjugate Agarose
  • Catalog # SA19588
  • Size 1mL
Applications
Immunoprecipitation (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA19588 372.00
Check your price
  • Rabbit anti-Chicken IgY Secondary Antibody, Rhodamine conjugate for ICC and IHC
  • Host Rabbit
  • Target Species Chicken
  • Conjugate Rhodamine
  • Catalog # SA19512
  • Size 500ug
Applications
Immunocytochemistry (1:50-1:200), Immunohistochemistry (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA19512 159.00
Check your price
  • Rabbit anti-Chicken IgY Secondary Antibody, FITC conjugate for ICC and IHC
  • Host Rabbit
  • Target Species Chicken
  • Conjugate FITC
  • Catalog # SA19511
  • Size 500ug
Applications
Immunocytochemistry (1:50-1:200), Immunohistochemistry (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA19511 166.00
Check your price
  • Rabbit anti-Chicken IgY Secondary Antibody, HRP conjugate for Western Blot, ICC, IHC and ELISA
  • Host Rabbit
  • Target Species Chicken
  • Conjugate HRP
  • Catalog # SA19509
  • Size 500ug
Applications
ELISA (1:2000-1:5000), Immunocytochemistry (1:250-1:3000), Immunohistochemistry (1:250-1:3000), Western Blot (1:5000-1:10000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA19509 196.00
Check your price
  • Rabbit anti-Chicken IgY Secondary Antibody, AP conjugate for Western Blot and ELISA
  • Host Rabbit
  • Target Species Chicken
  • Conjugate AP
  • Catalog # SA19514
  • Size 500ug
Applications
ELISA (1:2000-1:5000), Western Blot (1:2000-1:10000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA19514 197.00
Check your price
  • Bovine anti-Chicken IgY Secondary Antibody for Western Blot and ELISA
  • Host Bovine
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # SA19500
  • Size 500ug
Applications
ELISA (1:1000-1:5000), Western Blot (1:1000-1:10000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA19500 69.00
Check your price
  • Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 800 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Chicken
  • Conjugate DyLight® 800
  • Catalog # SA510076
  • Size 500ug
Applications
Flow Cytometry (1:50-200), Immunocytochemistry (1:50-500), Immunofluorescence (1:50-500), Immunohistochemistry (1:50-500), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510076 158.00
Check your price
  • Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 755 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Chicken
  • Conjugate DyLight® 755
  • Catalog # SA510075
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510075 158.00
Check your price
  • Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 680 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Chicken
  • Conjugate DyLight® 680
  • Catalog # SA510074
  • Size 500ug
Applications
Flow Cytometry (1:50-500), Immunocytochemistry (1:50 -500), Immunofluorescence (1:50-500), Immunohistochemistry (1:50 -500), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510074 156.00
Check your price
  • Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Chicken
  • Conjugate DyLight® 650
  • Catalog # SA510073
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510073 157.00
Check your price
  • Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 594 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Chicken
  • Conjugate DyLight® 594
  • Catalog # SA510072
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510072 156.00
Check your price
  • Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 550 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Chicken
  • Conjugate DyLight® 550
  • Catalog # SA510071
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510071 158.00
Check your price
  • Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Chicken
  • Conjugate DyLight® 488
  • Catalog # SA510070
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510070 160.00
Check your price
  • Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 350 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Chicken
  • Conjugate DyLight® 350
  • Catalog # SA510069
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510069 158.00
Check your price
  • Goat anti-Chicken IgY Secondary Antibody, WesternDot® 800 conjugate for Western Blot
  • Host Goat
  • Target Species Chicken
  • Conjugate WesternDot® 800
  • Catalog # W10822
  • Size 25blots
Applications
Western Blot (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10822 180.00
Check your price
  • Goat anti-Chicken IgY Secondary Antibody, WesternDot® 655 conjugate for Western Blot
  • Host Goat
  • Target Species Chicken
  • Conjugate WesternDot® 655
  • Catalog # W10820
  • Size 25blots
Applications
Western Blot (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10820 183.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, Alexa Fluor® 488 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Chicken
  • Conjugate Alexa Fluor® 488
  • Catalog # A11039
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11039 169.00
Check your price
  • Rabbit anti-Chicken IgY (H+L) Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Rabbit
  • Target Species Chicken
  • Conjugate Biotin
  • Catalog # 31720
  • Size 1.5mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31720 156.00
Check your price
  • Rabbit anti-Chicken IgY (H+L) Secondary Antibody, FITC conjugate for IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Chicken
  • Conjugate FITC
  • Catalog # 31501
  • Size 1.5mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31501 131.00
Check your price
  • Rabbit anti-Chicken IgY (H+L) Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Chicken
  • Conjugate HRP
  • Catalog # 31401
  • Size 1.5mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31401 179.00
Check your price
  • Rabbit anti-Chicken IgY (H+L) Secondary Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # 31104
  • Size 2mg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31104 142.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, Alexa Fluor® 647 conjugate for IF, ICC and IHC
  • Host Goat
  • Target Species Chicken
  • Conjugate Alexa Fluor® 647
  • Catalog # A21449
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21449 169.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Chicken
  • Conjugate Alexa Fluor® 594
  • Catalog # A11042
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11042 169.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Chicken
  • Conjugate Alexa Fluor® 568
  • Catalog # A11041
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11041 169.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, DSB-X Biotin conjugate for IF
  • Host Goat
  • Target Species Chicken
  • Conjugate
  • Catalog # D20701
  • Size 500µL
Applications
Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
D20701 Not Orderable Online
Check your price
  • Donkey anti-Chicken IgY Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot
  • Host Donkey
  • Target Species Chicken
  • Conjugate HRP
  • Catalog # SA1300
  • Size 250µL
Applications
Western Blot (1:250)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA1300 90.50
Check your price
  • Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 633 conjugate for IF, ICC and IHC
  • Host Goat
  • Target Species Chicken
  • Conjugate Alexa Fluor® 633
  • Catalog # A21103
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21103 169.00
Check your price