1 - 6 of 6 products displayed
  • CSTF2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530245
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Cleavage stimulation factor 64 kDa subunit; CF-1 64 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530245 349.00
Check your price
  • CSTF2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511777
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Cleavage stimulation factor 64 kDa subunit; CF-1 64 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511777 325.00
Check your price
  • CSTF2 Antibody (Monoclonal, 4E3-3C9)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518668
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Cleavage stimulation factor 64 kDa subunit; CF-1 64 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518668 309.00
Check your price
  • CSTF2T Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541856
  • Size 100 µl
Applications
Immunocytochemistry (4 µg/ml), Immunofluorescence (4 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
TauCstF-64; Cleavage stimulation factor 64 kDa subunit tau variant; CF-1 64 kDa subunit tau variant
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541856 309.00
Check your price
  • CSTF2T Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539109
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
TauCstF-64; Cleavage stimulation factor 64 kDa subunit tau variant; CF-1 64 kDa subunit tau variant
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539109 309.00
Check your price
  • CSTF2 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA537985
  • Size 100 µg
Applications
Immunoprecipitation (2µg/500µg lysate), Western Blot (2µg)
Alternate names:
Cleavage stimulation factor 64 kDa subunit; CF-1 64 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537985 309.00
Check your price