1 - 30 of 30 products displayed
  • Cyclin D1 Monoclonal Antibody for Western Blot, IF, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA512702
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
PRAD1 oncogene; BCL-1 oncogene; B-cell lymphoma 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512702 309.00
Check your price
  • Cyclin D1 Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514512
  • Size 500 µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:25-50), Western Blot (Assay Dependent)
Alternate names:
PRAD1 oncogene; BCL-1 oncogene; B-cell lymphoma 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514512 309.00
Check your price
  • Cyclin D3 Monoclonal Antibody for Western Blot, IF, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA512717
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:25), Immunoprecipitation (2 µg/ml), Western Blot (1:500-1:2000)
Alternate names:
D3-type cyclin, G1/S-specific cyclin D3, G1/S-specific cyclin-D3,OTTHUMP00000223415
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512717 309.00
Check your price
  • Phospho-Cyclin B1 (Ser126) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA536712
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
CCNB1, CCNB, cyclin B1, G2/mitotic-specific cyclin-B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536712 349.00
Check your price
  • Phospho-Cyclin B1 (Ser147) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538360
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
CCNB1, CCNB, cyclin B1, G2/mitotic-specific cyclin-B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538360 309.00
Check your price
  • Cyclin G1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536050
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Cyclin-G1, Cyclin-G, CCNG1, CCNG, CYCG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536050 309.00
Check your price
  • Cyclin D3 Monoclonal Antibody for Western Blot, IHC and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA112298
  • Size 100 µg
Applications
Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
RP5-973N23.3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112298 309.00
Check your price
  • CCNDBP1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528456
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Human homolog of Maid; Grap2 and cyclin-D-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528456 349.00
Check your price
  • CCNDBP1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537935
  • Size 100 µg
Applications
Western Blot (0.3-1 µg/ml)
Alternate names:
Human homolog of Maid; Grap2 and cyclin-D-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537935 309.00
Check your price
  • MNAT1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537378
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
p36; p35; RING finger protein MAT1; RING finger protein 66; Menage a trois; Cyclin-G1-interacting protein; CDK7/cyclin-H assembly factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537378 399.00
Check your price
  • Phospho-p21 (Thr145) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536677
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
p21; Melanoma differentiation-associated protein; CDK-interacting protein 1; Melanoma differentiation-associated protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536677 349.00
Check your price
  • Phospho-p21 (Thr145) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538144
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
p21; Melanoma differentiation-associated protein 6; CDK-interacting protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538144 309.00
Check your price
  • CDK5 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528795
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Tau protein kinase II catalytic subunit; Serine/threonine-protein kinase PSSALRE; Cell division protein kinase 5; CR6 protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528795 349.00
Check your price
  • CDK5 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA517053
  • Size 100 µL
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Tau protein kinase II catalytic subunit; Serine/threonine-protein kinase PSSALRE; Cell division protein kinase 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517053 309.00
Check your price
  • CDK2 Monoclonal Antibody, Biotin conjugate for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA513476
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:25-50), Western Blot (1-2 µg/ml)
Alternate names:
Cell division protein kinase 2; p33 protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513476 425.00
Check your price
  • Phospho-CDK2 (Thr160) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538128
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Cell division protein kinase 2; p33 protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538128 309.00
Check your price
  • CDK2 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA513725
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:100), Immunofluorescence (1:50-1:100), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Cell division protein kinase 2; p33 protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513725 349.00
Check your price
  • CDK2 Monoclonal Antibody for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHZ0142
  • Size 500 µl
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
Cell division protein kinase 2; p33 protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHZ0142 304.00
Check your price
  • Cdc47 Monoclonal Antibody for Western Blot, IHC (P), IP
  • Host Mouse
  • Target Species Human
  • Conjugate
  • Catalog # MA191554
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
P1.1-MCM3; CDC47 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191554 674.00
Check your price
  • SERTAD1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528460
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Transcriptional regulator interacting with the PHD-bromodomain 1; CDK4-binding protein p34SEI1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528460 349.00
Check your price
  • B-Myb Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA518449
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
MYBL2; BMYB; v-myb myeloblastosis viral oncogene homolog (avian)-like 2; HGNC:7548; MGC15600; MYB-related protein B; v-myb avian myeloblastosis viral oncogene homolog-like 2; B-MYB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518449 349.00
Check your price
  • Phospho-GSK3 alpha/beta (Tyr279, Tyr216) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # OPA103083
  • Size 100 µL
Applications
Western Blot (Assay-Dependent)
Alternate names:
Serine/threonine-protein kinase GSK3B; Serine/threonine-protein kinase GSK3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA103083 375.00
Check your price
  • PERK Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515305
  • Size 400 µL
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:1000)
Alternate names:
Pancreatic eIF2-alpha kinase; PRKR-like endoplasmic reticulum kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515305 349.00
Check your price
  • Cdc73 Polyclonal Antibody for Western Blot, IP and ChIP
  • Host Rabbit
  • Target Species Fruit fly, Human
  • Conjugate Unconjugated
  • Catalog # PA540082
  • Size 50 µg
Applications
ChIP assay (1-5 µg), Immunoprecipitation (12 µg), Western Blot (1:1000)
Alternate names:
Hyperparathyroidism 2 protein; Cell division cycle protein 73 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540082 309.00
Check your price
  • GSK3 beta Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Human, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA17621
  • Size 100 ug
Applications
ELISA (1:1000), Immunohistochemistry (Assay Dependent), Immunoprecipitation (5-10 µg), Western Blot (Assay-Dependent)
Alternate names:
Xgsk-3 protein; Serine/threonine-protein kinase GSK3B; Factor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17621 375.00
Check your price
  • PLAC1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521044
  • Size 100 ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1 ug/ml)
Alternate names:
CT92; cancer/testis antigen 92; placenta-specific protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521044 349.00
Check your price
  • PAGR1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520828
  • Size 100 ug
Applications
Western Blot (1 µg/ml)
Alternate names:
PTIP-associated protein 1; PAXIP1-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520828 349.00
Check your price
  • SMAD4 Oligoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710714
  • Size 100 µg
Applications
Flow Cytometry (2-4 µg x 10^6 cells), Immunocytochemistry (2 µg/ml), Immunofluorescence (2 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
SMAD family member 4; Deletion target in pancreatic carcinoma 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
710714 319.00
Check your price
  • SMAD4 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701682
  • Size 100 µg
Applications
Flow Cytometry (2-4 µg x 10^6 cells), Immunocytochemistry (2 µg/ml), Immunofluorescence (2 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
SMAD family member 4; Deletion target in pancreatic carcinoma 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
701682 325.00
Check your price
  • GSK3 alpha Polyclonal Antibody for Western Blot and ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA125969
  • Size 50 µg
Applications
ELISA (Assay Dependent), Western Blot (1:500)
Alternate names:
Serine/threonine-protein kinase GSK3A; Factor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125969 425.00
Check your price