1 - 3 of 3 products displayed
  • Dengue Virus Type 2 PrM Antibody (Monoclonal, DM-1)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171252
  • Size 1 mL
Applications
ELISA (5-10X), Immunofluorescence (5-10X), Western Blot (5-10X)
Alternate names:
Dengue prm, PrM protein, Dengue Virus 2 Prm, Dengue 2 prm
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171252 375.00
Check your price
  • Dengue Virus Type 2 PrM Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534966
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Dengue prm, PrM protein, Dengue Virus 2 Prm, Dengue 2 prm
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534966 349.00
Check your price
  • Dengue Virus Type 2 PrM Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534967
  • Size 100 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (1:1000-1:10,000)
Alternate names:
Dengue prm, PrM protein, Dengue Virus 2 Prm, Dengue 2 prm
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534967 349.00
Check your price