1 - 30 of 30 products displayed
  • DNA-PK Antibody (Monoclonal, 18-2 + 25-4 + 42-psc)
  • Host Mouse
  • Target Species Chicken, Hamster, Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA513404
  • Size 500 µL
Applications
Gel Shift (1mg/mL), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/mg protein lysate), Western Blot (1-2 µg/ml)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513404 349.00
Check your price
  • DNA-PK Antibody (Monoclonal, 25-4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513244
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (2µg/mg protein lysate), Western Blot (2-4 µg/ml)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513244 349.00
Check your price
  • DNA-PK Antibody, Biotin (Monoclonal, 18-2)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Biotin
  • Catalog # MA513235
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-100), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513235 399.00
Check your price
  • DNA-PK Antibody (Monoclonal, 42-psc (42-26 or 42-27))
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513249
  • Size 500 µL
Applications
Gel Shift (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:400)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513249 425.00
Check your price
  • DNA-PK Antibody (Monoclonal, 18-2)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA513238
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-100), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513238 349.00
Check your price
  • DNA-PK Antibody
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA123197
  • Size 50 µL
Applications
Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123197 349.00
Check your price
  • Phospho-DNA-PK (Thr2609) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129541
  • Size 50 µg
Applications
ELISA (1:300,000), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (1:500-1:2000)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129541 349.00
Check your price
  • Phospho-DNA-PK (Thr2609) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512913
  • Size 400 µL
Applications
Dot blot (1:500)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512913 349.00
Check your price
  • Phospho-DNA-PK (Thr2638) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538477
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538477 309.00
Check your price
  • Phospho-DNA-PK (Ser2056) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538478
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), peptide-ELISA (1:10,000)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538478 309.00
Check your price
  • DNA-PK Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529023
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529023 349.00
Check your price
  • Phospho-DNA-PK (Thr2647) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538475
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538475 309.00
Check your price
  • Phospho-DNA-PK (Ser2612) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538476
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538476 309.00
Check your price
  • DNA-PK Antibody
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA184583
  • Size 100 µL
Applications
Gel Shift (Assay Dependent), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (Assay Dependent), Western Blot (1/2000)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184583 399.00
Check your price
  • DNA-PK Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517644
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517644 275.00
Check your price
  • DNA-PK Antibody (Monoclonal, 3H6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515813
  • Size 100 µL
Applications
Immunocytochemistry (1/200-1/1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515813 349.00
Check your price
  • SSBP1 Antibody Polyclonal (PA1-27871)
  • Host Chicken
  • Target Species Mouse, Human, Rat
  • Conjugate
  • Catalog # PA127871
  • Size 50µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (1:2000)
Alternate names:
Single strand DNA-binding protein P16; PWP1-interacting protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127871 439.00
Check your price
  • CHD1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540067
  • Size 100 µl
Applications
Western Blot (1:500)
Alternate names:
ATP-dependent helicase CHD1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540067 309.00
Check your price
  • DDB1 Antibody (Monoclonal, 1322)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123335
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (1:1000)
Alternate names:
Xeroderma pigmentosum group E-complementing protein; XPE-binding factor; UV-damaged DNA-binding protein 1; UV-damaged DNA-binding factor; HBV X-associated protein 1; Damage-specific DNA-binding protein 1; DNA damage-binding protein a; DDB p127 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123335 309.00
Check your price
  • DDB1 Antibody (Monoclonal, 2B12D1)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 376200
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Xeroderma pigmentosum group E-complementing protein; XPE-binding factor; UV-damaged DNA-binding protein 1; UV-damaged DNA-binding factor; HBV X-associated protein 1; Damage-specific DNA-binding protein 1; DNA damage-binding protein a; DDB p127 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
376200 304.00
Check your price
  • DDB1 Antibody (Monoclonal, ZD001)
  • Host Mouse
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # 399901
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Xeroderma pigmentosum group E-complementing protein; XPE-binding factor; UV-damaged DNA-binding protein 1; UV-damaged DNA-binding factor; HBV X-associated protein 1; Damage-specific DNA-binding protein 1; DNA damage-binding protein a; DDB p127 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
399901 302.00
Check your price
  • DNA Ligase I Antibody (Monoclonal, 10H5)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA123189
  • Size 100 µg
Applications
Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Polydeoxyribonucleotide synthase [ATP] 1; DNA ligase I
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123189 309.00
Check your price
  • Ku80 Antibody (Monoclonal, 9403)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123314
  • Size 100 µg
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Western Blot (1:1000-1:5000)
Alternate names:
X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining); Thyroid-lupus autoantigen; Nuclear factor IV; Lupus Ku autoantigen protein p86; Ku86; Ku80; DNA repair protein XRCC5; CTC box-binding factor 85 kDa subunit; ATP-dependent DNA helicase II 80 kDa subunit; ATP-dependent DNA helicase 2 subunit 2; 86 kDa subunit of Ku antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123314 349.00
Check your price
  • Ku70 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123300
  • Size 100 µL
Applications
Western Blot (1:2500)
Alternate names:
X-ray repair complementing defective repair in Chinese hamster cells 6; Thyroid-lupus autoantigen; Lupus Ku autoantigen protein p70; DNA repair protein XRCC6; CTC box-binding factor 75 kDa subunit; ATP-dependent DNA helicase II 70 kDa subunit; ATP-dependent DNA helicase 2 subunit 1; 70 kDa subunit of Ku antigen; 5'-deoxyribose-5-phosphate lyase Ku70
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123300 309.00
Check your price
  • HLTF Antibody
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131579
  • Size 50 µg
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Sucrose nonfermenting protein 2-like 3; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 3; RING finger protein 80; HIP116; DNA-binding protein/plasminogen activator inhibitor 1 regulator; TNF-response element-binding protein; P113
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131579 399.00
Check your price
  • Amyloid Precursor Protein Antibody (Monoclonal, 3E9)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA125489
  • Size 50 µg
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (5-10 µg/ml)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP; Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125489 475.00
Check your price
  • GATA1 Antibody (Oligoclonal, 4HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710049
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Western Blot (1-5 ug/ml)
Alternate names:
NF-E1 DNA-binding protein; GATA-binding factor 1; Eryf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710049 309.00
Check your price
  • GATA1 Antibody (Monoclonal, 4H15L5)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700727
  • Size 100 µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
NF-E1 DNA-binding protein; GATA-binding factor 1; Eryf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
700727 304.00
Check your price
  • DNA Primase Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA129133
  • Size 500 µL
Applications
Western Blot (1:1000)
Alternate names:
DNA primase 49 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129133 349.00
Check your price
  • Ku80 Antibody (Monoclonal, SP130)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516375
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining); Thyroid-lupus autoantigen; Nuclear factor IV; Lupus Ku autoantigen protein p86; Ku86; Ku80; DNA repair protein XRCC5; CTC box-binding factor 85 kDa subunit; ATP-dependent DNA helicase II 80 kDa subunit; ATP-dependent DNA helicase 2 subunit 2; 86 kDa subunit of Ku antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516375 499.00
Check your price