1 - 30 of 30 products displayed
  • EGFR Antibody (Monoclonal, 111.6+199.12+ F4+11E8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513343
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513343 375.00
Check your price
  • EGFR Antibody (Monoclonal, 528 + 199.12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513697
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunoprecipitation (2 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513697 349.00
Check your price
  • EGFR Antibody (Monoclonal, 199.12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513319
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunoprecipitation (2 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513319 349.00
Check your price
  • EGFR Antibody (Monoclonal, EP38Y)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514485
  • Size 1 mL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (1:100), Western Blot (1-3 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514485 900.00
Check your price
  • EGFR Antibody (Monoclonal, 111.6)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA513269
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513269 349.00
Check your price
  • EGFR Antibody (Monoclonal, H9B4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513877
  • Size 500 µL
Applications
Immunoprecipitation (2 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513877 349.00
Check your price
  • EGFR Antibody (Monoclonal, 225)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA512880
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (2 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512880 309.00
Check your price
  • EGFR Antibody (Monoclonal, 528)
  • Host Mouse
  • Target Species Dog, Human
  • Conjugate Unconjugated
  • Catalog # MA512875
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (1:100-1:400), Immunoprecipitation (Assay Dependent)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512875 309.00
Check your price
  • EGFR Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA11110
  • Size 100 µg
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (1:200), Immunoprecipitation (2 µg), Western Blot (1:500)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11110 349.00
Check your price
  • Phospho-EGFR (Tyr1068) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OPA103170
  • Size 100 µL
Applications
Western Blot (Assay-Dependent)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA103170 349.00
Check your price
  • Phospho-EGFR (Tyr1173) Antibody (Monoclonal, EM-13)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110329
  • Size 100 µg
Applications
Flow Cytometry (2-8 ug/ml), Immunoprecipitation (Assay dependent), Western Blot (1 ug/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110329 349.00
Check your price
  • Phospho-EGFR (Tyr992) Antibody (Monoclonal, EM-12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110330
  • Size 100 ug
Applications
Flow Cytometry (2-8 ug/ml), Immunoprecipitation (Assay Dependent), Western Blot (1 ug/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110330 349.00
Check your price
  • Phospho-EGFR (Tyr1125) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512806
  • Size 400 µL
Applications
Dot blot (1:500)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512806 349.00
Check your price
  • Phospho-EGFR (Tyr1016) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512936
  • Size 400 µL
Applications
Dot blot (1:500), Western Blot (1:8000)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512936 349.00
Check your price
  • EGFR Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525976
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525976 349.00
Check your price
  • EGFR Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524584
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524584 349.00
Check your price
  • Phospho-EGFR (Tyr1148) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44792G
  • Size 10 blots
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (Assay Dependent)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44792G 309.00
Check your price
  • Phospho-EGFR (Tyr1173) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44794G
  • Size 10 blots
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44794G 309.00
Check your price
  • Phospho-EGFR (Tyr1086) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 369700
  • Size 50 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (0.1-0.5 µg/ml), Western Blot (0.1-0.5 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
369700 305.00
Check your price
  • Phospho-EGFR (Tyr992) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44786G
  • Size 10 blots
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44786G 304.00
Check your price
  • Phospho-EGFR (Tyr845) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44784G
  • Size 10 blots
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44784G 303.00
Check your price
  • Phospho-EGFR (Tyr1086) Antibody (Oligoclonal, 11HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710768
  • Size 100 µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710768 325.00
Check your price
  • Phospho-EGFR (Tyr1068) Antibody (Oligoclonal, 2HCLC)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710123
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (1-3 ug/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710123 309.00
Check your price
  • Phospho-EGFR (Tyr1173) Antibody (Monoclonal, 17H14L5)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701735
  • Size 100 µg
Applications
Flow Cytometry (2-4 µg x 10^6 cells), Immunocytochemistry (2 µg/ml), Immunofluorescence (2 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701735 309.00
Check your price
  • Phospho-EGFR (Tyr1068) Antibody (Monoclonal, 2H9L3)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701053
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:5000)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701053 309.00
Check your price
  • Nonphospho-EGFR (Tyr1173) Antibody (Monoclonal, 20G3)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44794M
  • Size 50µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44794M 309.00
Check your price
  • EGFR Antibody (Monoclonal, R19/48MIX)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHR5062
  • Size 100 µg
Applications
Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHR5062 305.00
Check your price
  • EGFR Antibody (Monoclonal, 199.12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHR5072
  • Size 500 µl
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHR5072 304.00
Check your price
  • EGFR Antibody, Biotin (Monoclonal, 528 + 199.12)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513694
  • Size 500 µL
Applications
Immunoprecipitation (2 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513694 399.00
Check your price
  • EGFR Antibody, Biotin (Monoclonal, 111.6)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513266
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513266 399.00
Check your price