1 - 5 of 5 products displayed
  • LMAN1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530081
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Lectin mannose-binding 1; Intracellular mannose-specific lectin MR60; Gp58; ER-Golgi intermediate compartment 53 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530081 349.00
Check your price
  • LMAN1 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127669
  • Size 50 µg
Applications
Western Blot (1:2000)
Alternate names:
Lectin mannose-binding 1; Intracellular mannose-specific lectin MR60; Gp58; ER-Golgi intermediate compartment 53 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127669 425.00
Check your price
  • LMAN1L Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525618
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Lectin mannose-binding 1-like; ERGIC53-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525618 349.00
Check your price
  • LMAN1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1074
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent), Western Blot (1:1000)
Alternate names:
p58; Lectin mannose-binding 1; ER-Golgi intermediate compartment 53 kDa protein; Intracellular mannose-specific lectin MR60; Gp58
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1074 349.00
Check your price
  • Lman1l Polyclonal Antibody for Western Blot
  • Host Sheep
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA547805
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Sublingual acinar membrane protein; Lectin mannose-binding 1-like; ERGIC53-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547805 349.00
Check your price