1 - 30 of 30 products displayed
  • Phospho-MEK1/MEK2 (Ser217, Ser221) Antibody (Monoclonal, C.158.9)
  • Host Rabbit
  • Target Species Chicken, Fruit fly, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515016
  • Size 100 µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1; Protein rolled; Extracellular-regulated kinase A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515016 309.00
Check your price
  • Phospho-ERK1/ERK2 (Thr202, Tyr204) Antibody (Monoclonal, S.812.9)
  • Host Rabbit
  • Target Species Fruit fly, Hamster, Human, Mink, Mouse, Non-human primate, Pig, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA515173
  • Size 200 µL
Applications
ChIP assay (1-3 µl), Immunohistochemistry (Paraffin) (1:400), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1; Protein rolled; Extracellular-regulated kinase A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515173 309.00
Check your price
  • Phospho-ERK1/ERK2 (Tyr204) Antibody (Monoclonal, B.742.5)
  • Host Rabbit
  • Target Species Fruit fly, Human, Mink, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA515174
  • Size 200 µL
Applications
Flow Cytometry (1:200), Immunocytochemistry (1:200), Immunofluorescence (1:200), Western Blot (1:1000)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1; Protein rolled; Extracellular-regulated kinase A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515174 309.00
Check your price
  • ERK1/ERK2 Antibody (Monoclonal, K.913.4)
  • Host Rabbit
  • Target Species Bovine, Dog, C. elegans, Chicken, Fruit fly, Hamster, Human, Mink, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA515134
  • Size 200 µL
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:250), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1; Protein rolled; Extracellular-regulated kinase A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515134 299.00
Check your price
  • ERK1/ERK2 Antibody (Monoclonal, ERK-7D8)
  • Host Mouse
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 136200
  • Size 200 µg
Applications
ELISA (Assay Dependent), Gel Shift (Assay Dependent), Immunocytochemistry (1µg/mL), Immunofluorescence (1µg/mL), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
136200 309.00
Check your price
  • ERK1 Antibody (Monoclonal, ERK-6B11)
  • Host Mouse
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 138600
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2
Catalog # Size Price (AUD) Your Price (AUD) Qty
138600 309.00
Check your price
  • MEK1/MEK2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA110034
  • Size 50 ug
Applications
Western Blot (0.5 ug/ml)
Alternate names:
MAPK/ERK kinase 1; ERK activator kinase 1; MAPK/ERK kinase 2; ERK activator kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110034 169.00
Check your price
  • MEK2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517678
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517678 299.00
Check your price
  • Phospho-MEK1 + MEK2 pSer217 Antibody Polyclonal (PA5-35603)
  • Host Rabbit
  • Target Species Mouse, Rat, Human
  • Conjugate
  • Catalog # PA535603
  • Size
Applications
Western Blot (1:500-1:2000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535603 339.00
Check your price
  • Phospho-MEK1/MEK2 (Ser222) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44452
  • Size 10 blots
Applications
Western Blot (1:1,000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44452 325.00
Check your price
  • Phospho-MEK1 / MAP2K1 pThr291 Antibody Polyclonal (PA5-35516)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # PA535516
  • Size
Applications
Western Blot (1:250-1:1000)
Alternate names:
MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535516 339.00
Check your price
  • MEK1 Antibody (Monoclonal, K.971.4)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515093
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:2000)
Alternate names:
MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515093 299.00
Check your price
  • MEK1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517677
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517677 299.00
Check your price
  • Phospho-MEK2 (Thr394) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538140
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538140 309.00
Check your price
  • MEK2 Antibody (Monoclonal, 7F5)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515600
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515600 349.00
Check your price
  • MEK2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA529291
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529291 349.00
Check your price
  • MEK2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527640
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527640 349.00
Check your price
  • MEK2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129852
  • Size 100 µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129852 399.00
Check your price
  • MEK2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA515167
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515167 325.00
Check your price
  • Phospho-MEK1 (Thr386) Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA14631
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14631 325.00
Check your price
  • Phospho-MEK1 (Thr292) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14630
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14630 309.00
Check your price
  • Phospho-MEK1/MEK2 (Ser221) Antibody (Monoclonal, K.742.1)
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515118
  • Size 100 µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:2000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515118 309.00
Check your price
  • Phospho-MEK1/MEK2 (Ser218, Ser222) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536823
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536823 349.00
Check your price
  • Phospho-MEK1/MEK2 (Ser222, Ser226) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536824
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536824 349.00
Check your price
  • MEK1/MEK2 Antibody (Monoclonal, J.653.9)
  • Host Mouse
  • Target Species Non-human primate, Rat, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515135
  • Size 100 µL
Applications
Flow Cytometry (1:25), Immunocytochemistry (1:25), Immunofluorescence (1:25), Immunohistochemistry (Paraffin) (1:25), Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515135 299.00
Check your price
  • MEK1/MEK2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531918
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
MAPK/ERK kinase 1; ERK activator kinase 1; MAPK/ERK kinase 2; ERK activator kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531918 325.00
Check your price
  • MEK1/MEK2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA531917
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531917 325.00
Check your price
  • Phospho-MEK1/MEK2 (Ser217, Ser221) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA14606
  • Size 100 µL
Applications
Western Blot (1:1,000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14606 325.00
Check your price
  • Phospho-MEK1/MEK2 (Ser218, Ser222, Ser226) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44454G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1µg/mL), Immunofluorescence (1µg/mL), Western Blot (1:500-1:2000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44454G 309.00
Check your price
  • Phospho-MEK1 (Ser218, Ser222) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512624
  • Size 400 µL
Applications
Dot blot (1:500), Immunohistochemistry (1:50-1:100)
Alternate names:
MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512624 325.00
Check your price