1 - 30 of 30 products displayed
  • Estrogen Receptor alpha Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA516476
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (10-20 µg/mL), Immunoprecipitation (10 µg/ml), Western Blot (2.5-5.0 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516476 349.00
Check your price
  • Estrogen Receptor alpha Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Hamster, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516440
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:250-500), Immunoprecipitation (10 µg/ml), Western Blot (2-5µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516440 349.00
Check your price
  • Phospho-Estrogen Receptor alpha (Ser106) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537569
  • Size 100 µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537569 309.00
Check your price
  • Phospho-Estrogen Receptor alpha (Ser167) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537570
  • Size 100 µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537570 309.00
Check your price
  • Phospho-Estrogen Receptor alpha (Ser104) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537568
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537568 309.00
Check your price
  • Phospho-Estrogen Receptor alpha (Ser102) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537567
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537567 309.00
Check your price
  • Phospho-Estrogen Receptor alpha (Tyr537) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537571
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537571 309.00
Check your price
  • Phospho-Estrogen Receptor alpha (Ser305) Polyclonal Antibody for IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538487
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538487 309.00
Check your price
  • Phospho-Estrogen Receptor alpha (Ser102) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539788
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539788 309.00
Check your price
  • Phospho-Estrogen Receptor alpha pSer118 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate
  • Catalog # PA535812
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535812 339.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA514104
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:100-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514104 550.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody, Biotin conjugate for IF, ICC, IP, GS and AP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA512325
  • Size 500 µL
Applications
Affinity Purification (Assay Dependent), Gel Shift (1mg/mL), Immunocytochemistry (1:500-1:2000), Immunofluorescence (1:500-1:2000), Immunoprecipitation (2 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512325 387.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IP and GS
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA512328
  • Size 500 µL
Applications
Gel Shift (1mg/mL), Immunoprecipitation (2 µg/ml), Western Blot (1:100-1:400)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512328 309.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA125623
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (15 µg/ml), Western Blot (1:250)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125623 399.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA513304
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin, Frozen) (1:40-1:80), Immunoprecipitation (10 µl/mg lysate), Western Blot (1:100-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513304 425.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA127107
  • Size 250 µL
Applications
Immunohistochemistry (Frozen) (1:40-1:80), Immunohistochemistry (Paraffin) (1:40-1:80), Immunoprecipitation (10 µl/mg lysate), Western Blot (1:50-1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127107 425.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA180216
  • Size 100 µL
Applications
Immunohistochemistry (Frozen) (1:40-1:60), Immunohistochemistry (Paraffin) (1:40-1:80), Immunoprecipitation (10 µl/mg lysate), Western Blot (1:50-1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180216 159.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA112600
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112600 275.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA139540
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA139540 1,100.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IHC and ChIP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 491002
  • Size 50 µg
Applications
ChIP assay (5 ug), Immunohistochemistry (Assay Dependent), Western Blot (7 ug/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
491002 309.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA139539
  • Size 7 mL
Applications
Immunohistochemistry (Paraffin) (Ready-to-use)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA139539 349.00
Check your price
  • Estrogen Receptor alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA1309
  • Size 50 µg
Applications
Immunocytochemistry (1:1000-1:4000), Immunofluorescence (1:1000-1:4000), Immunohistochemistry (Paraffin) (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1309 375.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IF and IHC
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1310
  • Size 100 µg
Applications
Immunofluorescence (10 µg/ml), Immunohistochemistry (1:20-1:100), Western Blot (4 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1310 375.00
Check your price
  • Estrogen Receptor alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1308
  • Size 50 µg
Applications
Immunocytochemistry (1:1000-1:4000), Immunofluorescence (1:1000-1:4000), Immunohistochemistry (Frozen) (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1308 349.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 417000
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
417000 433.00
Check your price
  • Phospho-Estrogen Receptor alpha (Ser104, Ser106) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA517795
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517795 309.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513191
  • Size 500 µL
Applications
Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunohistochemistry (Paraffin) (0.4-0.8 µg/mL), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513191 550.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514501
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:100-1:400), Western Blot (1:200-1:500)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514501 650.00
Check your price
  • Estrogen Receptor alpha Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532397
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532397 275.00
Check your price
  • Estrogen Receptor alpha Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137651
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137651 399.00
Check your price