1 - 30 of 30 products displayed
  • Phospho-Estrogen Receptor beta (Ser105) Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538488
  • Size 100µg
Applications
Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538488 309.00
Check your price
  • Estrogen Receptor beta Monoclonal Antibody for IF, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Camelid, Human, Non-human primate, Sheep, Pig, Reptile, Rat
  • Conjugate Unconjugated
  • Catalog # MA181281
  • Size 100µL
Applications
Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1/5-1/50)
Alternate names:
Nuclear receptor subfamily 3 group A member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181281 199.00
Check your price
  • Phospho-Estrogen Receptor alpha (Ser102) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539788
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539788 309.00
Check your price
  • Phospho-Estrogen Receptor alpha pSer118 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate
  • Catalog # PA535812
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535812 Not Orderable Online
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA139540
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA139540 1,100.00
Check your price
  • Estrogen Receptor alpha Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137651
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137651 399.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IHC and ChIP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 491002
  • Size 50µg
Applications
ChIP assay (5 ug), Immunohistochemistry (Assay Dependent), Western Blot (7 ug/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
491002 309.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA180216
  • Size 100µL
Applications
Immunohistochemistry (Frozen) (1:40-1:60), Immunohistochemistry (Paraffin) (1:40-1:80), Immunoprecipitation (10 µl/mg lysate), Western Blot (1:50-1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180216 159.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA112600
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112600 275.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA139539
  • Size 7mL
Applications
Immunohistochemistry (Paraffin) (Ready-to-use)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA139539 349.00
Check your price
  • Phospho-beta-2 Adrenergic Receptor (Ser346) Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538728
  • Size 100µg
Applications
Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538728 309.00
Check your price
  • RXRB Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # P21938
  • Size 100µL
Applications
Western Blot (Assay Dependent)
Alternate names:
DAUDI6, H-2RIIBP, NR2B2, RCoR-1, MHC class I promoter binding protein, nuclear receptor subfamily 2 group B member 2, retinoic acid receptor RXR-beta, retinoid X nuclear receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
P21938 Not Orderable Online
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody, Biotin conjugate for Western Blot, IP, ChIP and GS
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Sheep, Pig, Rabbit, Rat, Bovine, Dog, Chicken
  • Conjugate Biotin
  • Catalog # MA513062
  • Size 500µL
Applications
ChIP assay (Assay Dependent), Gel Shift (1mg/mL), Immunoprecipitation (2 µg/ml), Western Blot (1:1000-1:5000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513062 399.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IP, ChIP and GS
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Hamster, Human, Mouse, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA513065
  • Size 500µL
Applications
ChIP assay (Assay Dependent), Gel Shift (1mg/mL), Immunoprecipitation (2 µg/ml), Western Blot (1:200-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513065 349.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA3310
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100 -1:200), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:2000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3310 275.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IF and IHC
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1310
  • Size 100µg
Applications
Immunofluorescence (10 µg/ml), Immunohistochemistry (1:20-1:100), Western Blot (4 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1310 375.00
Check your price
  • Estrogen Receptor alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA1309
  • Size 50µg
Applications
Immunocytochemistry (1:1000-1:4000), Immunofluorescence (1:1000-1:4000), Immunohistochemistry (Paraffin) (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1309 375.00
Check your price
  • Phospho-Estrogen Receptor alpha (Ser118) Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515206
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515206 309.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123214
  • Size 100µg
Applications
Immunoprecipitation (1-5 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123214 349.00
Check your price
  • Estrogen Receptor alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1308
  • Size 50µg
Applications
Immunocytochemistry (1:1000-1:4000), Immunofluorescence (1:1000-1:4000), Immunohistochemistry (Frozen) (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1308 349.00
Check your price
  • Progestin Receptor beta Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA3881
  • Size 100µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Progestin and adipoQ receptor family member VIII; Progestin and adipoQ receptor family member 8; Lysosomal membrane protein in brain 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3881 349.00
Check your price
  • Progestin Receptor beta Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA3880
  • Size 100µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay dependent)
Alternate names:
Progestin and adipoQ receptor family member VIII; Progestin and adipoQ receptor family member 8; Lysosomal membrane protein in brain 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3880 349.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IP, ChIP, GS and Neu
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Hamster, Human, Mouse, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA112692
  • Size 100µg
Applications
Blocking Assay (Assay Dependent), ChIP assay (Assay Dependent), Gel Shift (Assay Dependent), Immunoprecipitation (2 µg/ml protein lysate), Western Blot (1-2 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112692 499.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515344
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515344 349.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515402
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515402 349.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515268
  • Size 100ug
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515268 349.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515464
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515464 349.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515454
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515454 349.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515689
  • Size 100µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515689 349.00
Check your price
  • Estrogen Receptor alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534577
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534577 349.00
Check your price