1 - 2 of 2 products displayed
  • ENPP2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA512478
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Extracellular lysophospholipase D; Autotaxin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512478 349.00
Check your price
  • ENPP2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112732
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (5-10µg/ml)
Alternate names:
Extracellular lysophospholipase D; Autotaxin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112732 550.00
Check your price