1 - 2 of 2 products displayed
  • AKAP3 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131348
  • Size 100 µg
Applications
ELISA (Assay-dependent), Western Blot (1-3 µg/ml)
Alternate names:
Sperm oocyte-binding protein; Protein kinase A-anchoring protein 3; Fibrousheathin-1; Fibrousheathin I; Fibrous sheath protein of 95 kDa; Cancer/testis antigen 82; A-kinase anchor protein 110 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131348 475.00
Check your price
  • AKAP3 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518261
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Sperm oocyte-binding protein; Protein kinase A-anchoring protein 3; Fibrousheathin-1; Fibrousheathin I; Fibrous sheath protein of 95 kDa; Cancer/testis antigen 82; A-kinase anchor protein 110 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518261 349.00
Check your price