1 - 30 of 30 products displayed
  • RFP Tag Oligoclonal Antibody for IF, ICC and Flow
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # 710530
  • Size 100µg
Applications
Flow Cytometry (1 ug/ml X 10^6 cells), Immunocytochemistry (1-2 ug/ml), Immunofluorescence (1-2 ug/ml)
Alternate names:
Red fluorescent protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
710530 309.00
Check your price
  • Maltose Binding Protein Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA3045
  • Size 50µl
Applications
Western Blot (1:1000-1:2000)
Alternate names:
MBP, Maltose Binding Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3045 309.00
Check your price
  • GFP Tag Polyclonal Antibody for Western Blot, ICC and ELISA
  • Host Chicken
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA19533
  • Size 500ug
Applications
ELISA (1:500-1:2000), Immunocytochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Green Fluorescence Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19533 219.00
Check your price
  • eGFP Tag Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA1952
  • Size 100µg
Applications
Immunocytochemistry (1:20-1:100), Immunofluorescence (1:20-1:100), Western Blot (1:1,000)
Alternate names:
Enhanced Green Fluorescence Protein, Venus, enhanced GFP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1952 159.00
Check your price
  • RFP Tag Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA1986
  • Size 100µg
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:1000)
Alternate names:
Red Fluorescent Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1986 249.00
Check your price
  • Green Fluorescent Protein (GFP) (residues 26-39) Synthetic Peptide for Ctrl
  • Host N/A
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # PEP033
  • Size 50µg
Applications
Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP033 98.00
Check your price
  • GFP Tag Polyclonal Antibody, Alexa Fluor® 594 conjugate for IF, ICC and IHC
  • Host Rabbit
  • Target Species Tag
  • Conjugate Alexa Fluor® 594
  • Catalog # A21312
  • Size 100µL
Applications
Immunocytochemistry (1-10 ug/ml), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Green Fluorescence Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21312 345.00
Check your price
  • GFP Tag Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # A11121
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Green Fluorescence Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11121 274.00
Check your price
  • GFP Tag Monoclonal Antibody, DyLight® 650 conjugate for IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight® 650
  • Catalog # MA515256D650
  • Size 50µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100)
Alternate names:
Green Fluorescence Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515256D650 159.00
Check your price
  • GFP Tag Monoclonal Antibody, DyLight® 680 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight® 680
  • Catalog # MA515256D680
  • Size 50µL
Applications
Immunocytochemistry (1:25-100), Immunofluorescence (1:25-100), Western Blot (1:500-1:1000)
Alternate names:
Green Fluorescent Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515256D680 159.00
Check your price
  • GFP Tag Polyclonal Antibody, Alexa Fluor® 555 conjugate for IF, ICC, IHC and ELISA
  • Host Rabbit
  • Target Species Tag
  • Conjugate Alexa Fluor® 555
  • Catalog # A31851
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Green Fluorescence Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
A31851 354.00
Check your price
  • GFP Tag Polyclonal Antibody, Biotin conjugate for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Tag
  • Conjugate Biotin
  • Catalog # A10259
  • Size 100µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Green Fluorescence Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
A10259 324.00
Check your price
  • GFP Tag Polyclonal Antibody, Biotin conjugate for Western Blot, IF and ICC
  • Host Chicken
  • Target Species Tag
  • Conjugate Biotin
  • Catalog # A10263
  • Size 100µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Green Fluorescence Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
A10263 328.00
Check your price
  • eGFP Tag Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # OSE00003G
  • Size 500µg
Applications
Immunohistochemistry (5-50 µg/ml), Western Blot (5-50 µg/ml)
Alternate names:
Enhanced Green Fluorescence Protein, Venus, enhanced GFP
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSE00003G 309.00
Check your price
  • eGFP Tag Polyclonal Antibody for Western Blot and IHC
  • Host Sheep
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # OSS00005W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Enhanced Green Fluorescence Protein, Venus, enhanced GFP
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSS00005W 375.00
Check your price
  • c-Abl Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514398
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2µg/mg protein lysate), Western Blot (1 µg/mL)
Alternate names:
p150; Proto-oncogene c-Abl; Abelson tyrosine-protein kinase 1; Abelson murine leukemia viral oncogene homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514398 349.00
Check your price
  • Phospho-CaMKII alpha (Thr286) Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1047
  • Size 100µL
Applications
ELISA (Assay dependent), Flow Cytometry (1µg per 10^6 cells), Immunocytochemistry (1:100), Immunofluorescence (1:50), Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type 2, Phospho-CaMKI
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1047 375.00
Check your price
  • MCP-1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA122488
  • Size 50µg
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:200-1:1000), Western Blot (1:2000)
Alternate names:
Small-inducible cytokine A2; Platelet-derived growth factor-inducible protein JE; Monocyte chemotactic protein 1; Monocyte chemoattractant protein 1; Immediate-early serum-responsive protein JE
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122488 349.00
Check your price
  • SETDB1 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA514960
  • Size 100µL
Applications
Immunocytochemistry (1:25), Immunofluorescence (1:25), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
SET domain bifurcated 1; Lysine N-methyltransferase 1E; Histone H3-K9 methyltransferase 4; ERG-associated protein with SET domain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514960 275.00
Check your price
  • CDK5 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHZ0492
  • Size 500µl
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
Tau protein kinase II catalytic subunit; Serine/threonine-protein kinase PSSALRE; Cell division protein kinase 5; CR6 protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHZ0492 303.00
Check your price
  • Phospho-CENPA (Ser7) Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA517195
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Centromere protein A; Centromere autoantigen A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517195 309.00
Check your price
  • Lactoferrin Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518107
  • Size 400µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000-1:16,000)
Alternate names:
Talalactoferrin; Growth-inhibiting protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518107 349.00
Check your price
  • Maltose Binding Protein Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA1989
  • Size 100µg
Applications
Western Blot (2 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1989 159.00
Check your price
  • PPP2R1A Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA514952
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:1000)
Alternate names:
PP2A subunit A isoform R1-alpha; PP2A subunit A isoform PR65-alpha; Medium tumor antigen-associated 61 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514952 275.00
Check your price
  • SKP2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517009
  • Size 100µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Western Blot (1:1000)
Alternate names:
p45skp2; F-box/LRR-repeat protein 1; F-box protein Skp2; Cyclin-A/CDK2-associated protein p45; F-box/WD-40 protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517009 275.00
Check your price
  • BMI-1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535360
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
BMI1; PCGF4; RNF51; Polycomb complex protein BMI-1; Polycomb group RING finger protein 4; RING finger protein 51
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535360 349.00
Check your price
  • LSD1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA514934
  • Size 100µL
Applications
Immunocytochemistry (1:400), Immunofluorescence (1:400), Immunohistochemistry (Frozen) (1:800), Immunohistochemistry (Paraffin) (1:800), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Flavin-containing amine oxidase domain-containing protein 2; BRAF35-HDAC complex protein BHC110
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514934 275.00
Check your price
  • SKP2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA517497
  • Size 100µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1:1000)
Alternate names:
p45skp2; F-box/LRR-repeat protein 1; F-box protein Skp2; Cyclin-A/CDK2-associated protein p45
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517497 275.00
Check your price
  • ASF1B Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA514836
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
CCG1-interacting factor A-II; Anti-silencing function protein 1 homolog B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514836 275.00
Check your price
  • LIN28A Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1096X
  • Size 20µL
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:20-1:50), Immunoprecipitation (5 µg, 1:1000), Western Blot (1:1000)
Alternate names:
Testis-expressed protein 17; Zinc finger CCHC domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1096X 119.00
Check your price