1 - 30 of 30 products displayed
  • Fyn Monoclonal Antibody for Western Blot, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA119331
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (5 µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (2 µg/ml)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119331 299.00
Check your price
  • Fyn Monoclonal Antibody for Western Blot, IF, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 137800
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
137800 309.00
Check your price
  • Fyn Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515865
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515865 349.00
Check your price
  • Fyn Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515866
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515866 349.00
Check your price
  • Fyn Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA528975
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528975 349.00
Check your price
  • Fyn Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527351
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527351 349.00
Check your price
  • Fyn Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514780
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514780 349.00
Check your price
  • Phospho-Fyn (Tyr530) Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536644
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536644 349.00
Check your price
  • Phospho-Fyn (Tyr530) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538115
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538115 309.00
Check your price
  • Fyn Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA517499
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517499 275.00
Check your price
  • Fyn Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA112607
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (Assay Dependent)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112607 499.00
Check your price
  • FRK Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515425
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Protein-tyrosine kinase 5; Nuclear tyrosine protein kinase RAK; FYN-related kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515425 349.00
Check your price
  • FRK Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515547
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Protein-tyrosine kinase 5; Nuclear tyrosine protein kinase RAK; FYN-related kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515547 349.00
Check your price
  • Fyn Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA19045
  • Size 100 ug
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19045 309.00
Check your price
  • Fyn Monoclonal Antibody for Western Blot, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHO0482
  • Size 500 µl
Applications
Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay Dependent)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO0482 304.00
Check your price
  • FRK Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534782
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Protein-tyrosine kinase 5; Nuclear tyrosine protein kinase RAK; FYN-related kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534782 349.00
Check your price
  • FRK Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528573
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Protein-tyrosine kinase 5; Nuclear tyrosine protein kinase RAK; FYN-related kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528573 349.00
Check your price
  • FRK Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528373
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Protein-tyrosine kinase 5; Nuclear tyrosine protein kinase RAK; FYN-related kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528373 349.00
Check your price
  • PKN2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534784
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Protein-kinase C-related kinase 2; Protein kinase C-like 2; PKN gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534784 349.00
Check your price
  • ZAP70 Monoclonal Antibody for Western Blot, Flow and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 376500
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Syk-related tyrosine kinase; 70 kDa zeta-chain associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
376500 304.00
Check your price
  • ZAP70 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515137
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Syk-related tyrosine kinase; 70 kDa zeta-chain associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515137 275.00
Check your price
  • Lyn Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFPA0074
  • Size 100 µL
Applications
Western Blot (1:2000)
Alternate names:
p56Lyn; p53Lyn; V-yes-1 Yamaguchi sarcoma viral related oncogene homolog; Lck/Yes-related novel protein tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0074 309.00
Check your price
  • Phospho-ZAP70/Syk (Tyr319, Tyr352) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Horse, Hamster, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA515095
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Spleen tyrosine kinase; p72-Syk; Syk-related tyrosine kinase; 70 kDa zeta-chain associated protein; p72Syk
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515095 309.00
Check your price
  • Phospho-ZAP70/Syk (Tyr319, Tyr352) Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Horse, Hamster, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA517815
  • Size 100 µL
Applications
Flow Cytometry (1:25), Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Spleen tyrosine kinase; p72-Syk; Syk-related tyrosine kinase; 70 kDa zeta-chain associated protein; p72Syk
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517815 309.00
Check your price
  • Phospho-ZAP70 (Tyr315, Tyr319) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA125547
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
Syk-related tyrosine kinase; 70 kDa zeta-chain associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125547 349.00
Check your price
  • Phospho-ZAP70 (Tyr493) Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517778
  • Size 100 µL
Applications
Flow Cytometry (1:100), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Syk-related tyrosine kinase; 70 kDa zeta-chain associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517778 309.00
Check your price
  • Phospho-ZAP70 (Tyr292) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA125546
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
Syk-related tyrosine kinase; 70 kDa zeta-chain associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125546 349.00
Check your price
  • ZAP70 Monoclonal Antibody for Western Blot, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515180
  • Size 100 µL
Applications
Flow Cytometry (1:200), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Syk-related tyrosine kinase; 70 kDa zeta-chain associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515180 275.00
Check your price
  • ZAP70 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515281
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Syk-related tyrosine kinase; 70 kDa zeta-chain associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515281 349.00
Check your price
  • ZAP70 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0197
  • Size 100 µL
Applications
ELISA (Assay Dependent), Immunoprecipitation (2 µg), Western Blot (0.1 µg/ml)
Alternate names:
Syk-related tyrosine kinase; 70 kDa zeta-chain associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0197 309.00
Check your price