1 - 2 of 2 products displayed
  • NPRL2 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518862
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Tumor suppressor candidate 4; Gene 21 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518862 325.00
Check your price
  • Androgen Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1111B
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Western Blot (2 ug/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 4; Dihydrotestosterone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1111B 375.00
Check your price