1 - 30 of 30 products displayed
  • PPAR alpha/gamma Polyclonal Antibody for Western Blot, IP and GS
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA3820
  • Size 100µL
Applications
Gel Shift (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
Nuclear receptor subfamily 1 group C member 1; Nuclear receptor subfamily 1 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3820 375.00
Check your price
  • RARG Polyclonal Antibody for Western Blot, IP and GS
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA3813
  • Size 100µL
Applications
Gel Shift (1-2 µl), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
Nuclear receptor subfamily 1 group B member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3813 349.00
Check your price
  • RARG Polyclonal Antibody for Western Blot, IP and GS
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA3814
  • Size 100µL
Applications
Gel Shift (1-2 µl), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:5000)
Alternate names:
Nuclear receptor subfamily 1 group B member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3814 349.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate HRP
  • Catalog # MA516308HRP
  • Size 50µL
Applications
Western Blot (1:500-1:2000)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308HRP 159.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody, DyLight 800 4X PEG conjugate for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate DyLight 800 4X PEG
  • Catalog # MA516308D800
  • Size 50µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308D800 225.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody, DyLight 800 4X PEG conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate DyLight 800 4X PEG
  • Catalog # MA515739D800
  • Size 50µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739D800 159.00
Check your price
  • beta Actin Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate HRP
  • Catalog # MA515739HRP
  • Size 50µL
Applications
Western Blot (1:500-1:2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739HRP 159.00
Check your price
  • Rabbit anti-Mouse IgG Secondary Antibody, HRP conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # PA128568
  • Size 1mg
Applications
Dot blot (Assay dependent), ELISA (1:6000-1:30,000), Immunohistochemistry (Frozen) (1:1500-1:7500), Immunohistochemistry (Paraffin) (1:1500-1:7500), Immunomicroscopy (Assay dependent), Western Blot (1:3000-1:15,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128568 208.00
Check your price
  • Rabbit anti-Mouse IgG Secondary Antibody, HRP conjugate for Western Blot, IHC (F), IHC (P), ELISA, DB and IM
  • Host Rabbit
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # PA128761
  • Size 100µg
Applications
Dot blot (Assay dependent), ELISA (1:5000-1:20,000), Immunohistochemistry (Frozen) (1:200-1:1000), Immunohistochemistry (Paraffin) (1:200-1:1000), Immunomicroscopy (Assay dependent), Western Blot (1:500-1:2500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128761 139.00
Check your price
  • Goat anti-Mouse IgG Secondary Antibody, AP conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate AP
  • Catalog # PA128755
  • Size 100µg
Applications
Dot blot (Assay dependent), ELISA (1:4000-1:16,000), Immunohistochemistry (Paraffin, Frozen) (1:200-1:1000), Immunomicroscopy (Assay dependent), Western Blot (1:1000-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128755 167.00
Check your price
  • ERK1/ERK2 Oligoclonal Antibody for Western Blot and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710318
  • Size 100µg
Applications
ChIP assay (1 ul), Western Blot (0.5-2 ug/ml)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710318 309.00
Check your price
  • ERK1/ERK2 Oligoclonal Antibody for Western Blot and ChIP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 710214
  • Size 100µg
Applications
ChIP assay (1 ul), Western Blot (1-2 ug/ml)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1; p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2
Catalog # Size Price (AUD) Your Price (AUD) Qty
710214 309.00
Check your price
  • RUNX1 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710233
  • Size 100µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
SL3/AKV core-binding factor alpha B subunit; SL3-3 enhancer factor 1 alpha B subunit; Polyomavirus enhancer-binding protein 2 alpha B subunit; Oncogene AML-1; Core-binding factor subunit alpha-2; Acute myeloid leukemia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
710233 309.00
Check your price
  • RUNX1 Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 701202
  • Size 100µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
SL3/AKV core-binding factor alpha B subunit; SL3-3 enhancer factor 1 alpha B subunit; Polyomavirus enhancer-binding protein 2 alpha B subunit; Oncogene AML-1; Core-binding factor subunit alpha-2; Acute myeloid leukemia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
701202 309.00
Check your price
  • NCoR2 Monoclonal Antibody for Western Blot, IHC and ChIP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1843
  • Size 100µg
Applications
ChIP assay (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (4 µg/ml)
Alternate names:
Thyroid-, retinoic-acid-receptor-associated corepressor; T3 receptor-associating factor; Silencing mediator of retinoic acid and thyroid hormone receptor; SMAP270; CTG repeat protein 26
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1843 349.00
Check your price
  • Actin Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Human, Mouse, Non-human primate, Plant, Pig, Protozoa, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA511866
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (2 ug), Western Blot (1:5000-1:20,000)
Alternate names:
Smooth muscle gamma-actin; Gamma-2-actin; Alpha-actin-3; Beta-actin; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1; Alpha-cardiac actin; Alpha-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511866 399.00
Check your price
  • SMAD1 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710199
  • Size 100µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
SMAD family member 1; Mothers-against-DPP-related 1; Dwarfin-A; Transforming growth factor-beta-signaling protein 1; Mad-related protein 1; JV4-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710199 309.00
Check your price
  • SMAD1 Monoclonal Antibody for Western Blot, Flow and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 701168
  • Size 100µg
Applications
ChIP assay (3µg), Flow Cytometry (1-3µg x 10^6 cells), Western Blot (1-3µg/ml)
Alternate names:
SMAD family member 1; Mothers-against-DPP-related 1; Dwarfin-A; Transforming growth factor-beta-signaling protein 1; Mad-related protein 1; JV4-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701168 304.00
Check your price
  • Histone H3 Oligoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710546
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (2 ug/ml), Immunofluorescence (2 ug/ml), Western Blot (0.5 ug/ml)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
710546 309.00
Check your price
  • alpha Tubulin Monoclonal Antibody for Western Blot, IF, IHC (F), IP, ELISA and RIA
  • Host Rat
  • Target Species Bovine, Dog, Fruit fly, Human, Mouse, Non-human primate, Plant, Pig, Rabbit, Rat, Xenopus, Yeast
  • Conjugate Unconjugated
  • Catalog # MA180017
  • Size 500µg
Applications
ELISA (1/100-1/1000), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tubulin alpha-3 chain; Tubulin B-alpha-1; Alpha-tubulin 3; Tubulin alpha-1 chain; Alpha-tubulin 1; Alpha-tubulin isotype M-alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180017 399.00
Check your price
  • Phospho-GIT1 (Tyr554) Polyclonal Antibody for Western Blot and DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512982
  • Size 400µL
Applications
Dot blot (1:500)
Alternate names:
GRK-interacting protein 1; G protein-coupled receptor kinase-interactor 1; Cool-associated and tyrosine-phosphorylated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512982 349.00
Check your price
  • Phospho-p70 S6 Kinase (Thr421, Ser424) Monoclonal Antibody for Western Blot, Flow and ChIP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 701083
  • Size 100µg
Applications
ChIP assay (1 ul), Flow Cytometry (1:100-1:500), Western Blot (1:500-1:5000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701083 Contact Us ›
Check your price
  • SQSTM1 Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701510
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (1 ug/ml), Immunofluorescence (1 ug/ml), Western Blot (0.5 ug/ml)
Alternate names:
Ubiquitin-binding protein p62; Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; EBI3-associated protein of 60 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
701510 309.00
Check your price
  • Phospho-CREB (Ser133) Monoclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 700129
  • Size 100µg
Applications
Flow Cytometry (1-3µg x 10^6 cells), Western Blot (1-3µg/ml)
Alternate names:
Active transcription factor CREB, cAMP responsive element binding protein 1, CREB1, MGC9284, Transactivator protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
700129 303.00
Check your price
  • GAPDH Loading Control Monoclonal Antibody, DyLight 800 4X PEG conjugate for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate DyLight 800 4X PEG
  • Catalog # MA515738D800
  • Size 50µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738D800 225.00
Check your price
  • GAPDH Loading Control Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast
  • Conjugate HRP
  • Catalog # MA515738HRP
  • Size 50µL
Applications
Western Blot (1:500-1:2000)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738HRP 159.00
Check your price
  • GAPDH Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1987
  • Size 100µg
Applications
Western Blot (1:1000-1:5000)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1987 249.00
Check your price
  • GAPDH Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1988
  • Size 100µg
Applications
Western Blot (1:500-1:5000)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1988 249.00
Check your price
  • RALBP1 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA1035X
  • Size 20µL
Applications
Immunoprecipitation (2 µg), Western Blot (1:1000)
Alternate names:
Ral-interacting protein 1; Dinitrophenyl S-glutathione ATPase; 76 kDa Ral-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1035X 119.00
Check your price
  • c-Myc Tag Monoclonal Antibody, DyLight 800 4X PEG conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight 800 4X PEG
  • Catalog # MA121316D800
  • Size 50µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Transcription factor p64; Proto-oncogene c-Myc; Class E basic helix-loop-helix protein 39
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121316D800 225.00
Check your price