1 - 3 of 3 products displayed
  • Giardia lamblia 65 kDa Antibody (Monoclonal, F86B)
  • Host Mouse
  • Target Species Protozoa
  • Conjugate Unconjugated
  • Catalog # MA17437
  • Size 100 µg
Applications
ELISA (1:20), Immunofluorescence (1:10), Western Blot (1:10)
Alternate names:
G. lamblia
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17437 309.00
Check your price
  • Giardia lamblia 65 kDa Antibody (Monoclonal, D33I)
  • Host Mouse
  • Target Species Protozoa
  • Conjugate Unconjugated
  • Catalog # MA17440
  • Size 100 µg
Applications
ELISA (1:20), Immunofluorescence (1:10), Western Blot (1:10)
Alternate names:
G. lamblia
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17440 309.00
Check your price
  • Giardia lamblia 65 kDa Antibody (Monoclonal, S1H)
  • Host Mouse
  • Target Species Protozoa
  • Conjugate Unconjugated
  • Catalog # MA518207
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
G. lamblia
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518207 299.00
Check your price