1 - 3 of 3 products displayed
  • Hepatitis C Virus NS4b Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17616
  • Size 100ug
Applications
ELISA (10^-6), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
HepC, HCV, HepC NS4, HCV NS4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17616 309.00
Check your price
  • Hepatitis C Virus NS4b Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17365
  • Size 100µg
Applications
ELISA (1:20-1:200), Western Blot (1:10-1:50)
Alternate names:
HepC, HCV, HepC NS4, HCV NS4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17365 375.00
Check your price
  • Hepatitis C Virus NS4b Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17358
  • Size 100µg
Applications
ELISA (1:20-1:200), Western Blot (1:10-1:50)
Alternate names:
HepC, HCV, HepC NS4, HCV NS4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17358 375.00
Check your price