1 - 30 of 30 products displayed
  • Hepatitis B Virus Core Antigen Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA516368
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
HepB, Hepatitis B virus nucleocapsid particles, HBcAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516368 225.00
Check your price
  • Hepatitis B Virus Core Antigen Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA121697
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
HepB, Hepatitis B virus nucleocapsid particles, HBcAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121697 349.00
Check your price
  • Hepatitis B Virus Core Antigen Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17608
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
HepB, Hepatitis B virus nucleocapsid particles, HBcAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17608 275.00
Check your price
  • Hepatitis B Virus Core Antigen Monoclonal Antibody for IHC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17606
  • Size 100ug
Applications
ELISA (1:1000), Flow Cytometry (1:1000), Immunohistochemistry (1:1000), Immunoprecipitation (1:1000)
Alternate names:
HepB, Hepatitis B virus nucleocapsid particles, HBcAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17606 275.00
Check your price
  • Hepatitis B Virus Core Antigen Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17609
  • Size 100µg
Applications
ELISA (1:1000), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
HepB, Hepatitis B virus nucleocapsid particles, HBcAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17609 275.00
Check your price
  • Hepatitis B Virus Core Antigen Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17607
  • Size 100ug
Applications
ELISA (1:1000), Immunohistochemistry (1:1000), Immunoprecipitation (1:1000), Western Blot (1:1000)
Alternate names:
HepB, Hepatitis B virus nucleocapsid particles, HBcAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17607 275.00
Check your price
  • Hepatitis B Virus Core Antigen Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17610
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
HepB, Hepatitis B virus nucleocapsid particles, HBcAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17610 309.00
Check your price
  • Hepatitis B Virus E Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA120275
  • Size 100µg
Applications
ELISA (Assay dependent)
Alternate names:
HepB, HBVgp4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120275 309.00
Check your price
  • Hepatitis B Virus E Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA120273
  • Size 200µg
Applications
ELISA (Assay dependent)
Alternate names:
HepB, HBVgp4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120273 349.00
Check your price
  • Hepatitis B Virus preS2 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA123356
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
HepB, HBVgp4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123356 399.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA514720
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514720 349.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA514678
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514678 349.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA514679
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514679 349.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA514731
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514731 349.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MIH9701
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9701 600.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MIH9705
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9705 600.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MIH9706
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9706 600.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17348
  • Size 100µg
Applications
ELISA (1:20)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17348 309.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17346
  • Size 100µg
Applications
ELISA (1:20)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17346 309.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17350
  • Size 100µg
Applications
ELISA (1:20)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17350 309.00
Check your price
  • Hepatitis B Virus Surface Polyclonal Antibody for Western Blot and ELISA
  • Host Horse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA173083
  • Size 1mg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173083 349.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA513059
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513059 275.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA119263
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119263 249.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA119264
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119264 199.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IHC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17604
  • Size 100ug
Applications
ELISA (1:1000), Flow Cytometry (1:1000), Immunohistochemistry (1:1000), Immunoprecipitation (1:1000)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17604 309.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for Flow, IP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17603
  • Size 100ug
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17603 309.00
Check your price
  • KIAA0020 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531247
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Minor histocompatibility antigen HA-8; HBV XAg-transactivated protein 5; HBV X-transactivated gene 5 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531247 349.00
Check your price
  • KIAA0020 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527851
  • Size 100µL
Applications
Western Blot (1:5000-1:20,000)
Alternate names:
Minor histocompatibility antigen HA-8; HBV XAg-transactivated protein 5; HBV X-transactivated gene 5 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527851 349.00
Check your price
  • EXOC3L2 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524947
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
HBV XAg-transactivated protein 7; HBV X-transactivated gene 7 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524947 349.00
Check your price
  • EXOC3L2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA545824
  • Size 100µl
Applications
Western Blot (1 µg/ml)
Alternate names:
HBV XAg-transactivated protein 7; HBV X-transactivated gene 7 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545824 309.00
Check your price