1 - 7 of 7 products displayed
  • Hepatitis C Virus NS5 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA173109
  • Size 1 mL
Applications
ELISA (1:1000), Western Blot (1:100)
Alternate names:
HepC, HCV, HepC NS5, HCV NS5, NICE-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173109 349.00
Check your price
  • Hepatitis C Virus NS5 Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA182923
  • Size 200 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
HepC, HCV, HepC NS5, HCV NS5, NICE-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182923 449.00
Check your price
  • Hepatitis C Virus NS5a Monoclonal Antibody for Western Blot, IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17368
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Western Blot (1:10-1:500)
Alternate names:
HepC, HCV, HepC NS5, HCV NS5, NICE-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17368 375.00
Check your price
  • Hepatitis C Virus NS5b Monoclonal Antibody for Western Blot, IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17343
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
HepC, HCV, HepC NS5, HCV NS5, NICE-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17343 349.00
Check your price
  • Hepatitis C Virus NS5a Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17344
  • Size 100 µg
Applications
ELISA (1:20), Immunofluorescence (1:10)
Alternate names:
HepC, HCV, HepC NS5, HCV NS5, NICE-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17344 349.00
Check your price
  • Hepatitis C Virus NS5b Polyclonal Antibody for Western Blot, IP and ELISA
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534981
  • Size 100 ug
Applications
ELISA (1:1000), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
HepC, HCV, HepC NS5, HCV NS5, NICE-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534981 249.00
Check your price
  • Hepatitis C Virus NS5b Monoclonal Antibody for Western Blot, IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17342
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
HepC, HCV, HepC NS5, HCV NS5, NICE-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17342 375.00
Check your price