1 - 30 of 30 products displayed
  • HGF Monoclonal Antibody for Western Blot, ELISA, Neu and IA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124767
  • Size 100µg
Applications
ELISA (0.5-1 µg/ml), Inhibition Assays (0.01-0.50 ug/ml), Neutralization (0.01-0.5 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Scatter factor; Hepatopoietin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124767 349.00
Check your price
  • HGF Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA128023
  • Size 100µg
Applications
ELISA (0.5 µg/ml), Neutralization (Assay dependent), Western Blot (1µg/ml)
Alternate names:
Scatter factor; Hepatopoietin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128023 309.00
Check your price
  • c-Met Polyclonal Antibody for Western Blot, IHC, ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA120184
  • Size 25µg
Applications
ELISA (0.5-1 µg/ml), Immunohistochemistry (Assay dependent), Neutralization (0.3-1 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
AUTS9, HGFR, RCCP2, c-Met
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120184 349.00
Check your price
  • NR5A2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534018
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (10-19 µg/ml)
Alternate names:
Liver receptor homolog 1; Hepatocytic transcription factor; CYP7A promoter-binding factor; B1-binding factor; Alpha-1-fetoprotein transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534018 349.00
Check your price
  • NR5A2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534017
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (7 µg/ml)
Alternate names:
Liver receptor homolog 1; Hepatocytic transcription factor; CYP7A promoter-binding factor; B1-binding factor; Alpha-1-fetoprotein transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534017 349.00
Check your price
  • NR5A2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528347
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Liver receptor homolog 1; Hepatocytic transcription factor; CYP7A promoter-binding factor; B1-binding factor; Alpha-1-fetoprotein transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528347 349.00
Check your price
  • TCF2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA124459
  • Size 50µg
Applications
Western Blot (0.1-2 µg/ml)
Alternate names:
Transcription factor 2; Homeoprotein LFB3; Variant hepatic nuclear factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124459 425.00
Check your price
  • Phospho-FOXO pan pThr24 / pThr32 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat, Human, Mouse
  • Conjugate
  • Catalog # PA535576
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Fork head domain transcription factor AFX1; Forkhead in rhabdomyosarcoma; Forkhead box protein O1A; Afxh; Forkhead in rhabdomyosarcoma-like 1; AF6q21 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535576 Not Orderable Online
Check your price
  • MPM2 Monoclonal Antibody for Western Blot, IF, ICC, ELISA
  • Host Mouse
  • Target Species Human, Yeast
  • Conjugate
  • Catalog # MA191855
  • Size 50µg
Applications
ELISA (Assay dependent), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 ug/ml), Western Blot (Assay Dependent)
Alternate names:
Winged-helix factor from INS-1 cells; Transcription factor Trident; MPM-2 reactive phosphoprotein 2; M-phase phosphoprotein 2; Hepatocyte nuclear factor 3 forkhead homolog 11; Forkhead-related protein FKHL16
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191855 Not Orderable Online
Check your price
  • Phospho-HNF4A (Ser313) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536857
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Transcription factor HNF-4; Transcription factor 14; Nuclear receptor subfamily 2 group A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536857 349.00
Check your price
  • Phospho-HNF4A (Ser304) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537595
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Transcription factor HNF-4; Transcription factor 14; Nuclear receptor subfamily 2 group A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537595 309.00
Check your price
  • Phospho-HNF4A (Ser313) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539695
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Transcription factor HNF-4; Transcription factor 14; Nuclear receptor subfamily 2 group A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539695 309.00
Check your price
  • HNF4A Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514891
  • Size 100µL
Applications
Immunocytochemistry (1:6000), Immunofluorescence (1:6000), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:1000)
Alternate names:
Transcription factor HNF-4; Transcription factor 14; Nuclear receptor subfamily 2 group A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514891 275.00
Check your price
  • HNF4A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527159
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Transcription factor HNF-4; Transcription factor 14; Nuclear receptor subfamily 2 group A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527159 349.00
Check your price
  • HNF4A Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518363
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Transcription factor HNF-4; Transcription factor 14; Nuclear receptor subfamily 2 group A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518363 349.00
Check your price
  • HNF4A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512370
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Transcription factor HNF-4; Transcription factor 14; Nuclear receptor subfamily 2 group A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512370 349.00
Check your price
  • HNF4A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526443
  • Size 400µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Transcription factor HNF-4; Transcription factor 14; Nuclear receptor subfamily 2 group A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526443 349.00
Check your price
  • HNF4A Monoclonal Antibody for Western Blot, IF, IHC, IP, ChIP, ELISA and GS
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 417700
  • Size 100µg
Applications
ChIP assay (Assay Dependent), ELISA (Assay Dependent), Gel Shift (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Transcription factor HNF-4; Transcription factor 14; Nuclear receptor subfamily 2 group A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
417700 428.00
Check your price
  • HNF4A Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 417800
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Transcription factor HNF-4; Transcription factor 14; Nuclear receptor subfamily 2 group A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
417800 433.00
Check your price
  • FOXJ1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536210
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Hepatocyte nuclear factor 3 forkhead homolog 4; Forkhead-related protein FKHL13
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536210 309.00
Check your price
  • FOXA1 Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518168
  • Size 100ug
Applications
Immunohistochemistry (5-10 µg/ml), Western Blot (0.2-0.6 ug/ml)
Alternate names:
Forkhead box protein A1; Transcription factor 3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518168 349.00
Check your price
  • FOXA1 Monoclonal Antibody for IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516364
  • Size 500µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Transcription factor 3A; Forkhead box protein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516364 499.00
Check your price
  • FOXA1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532411
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Transcription factor 3A; Forkhead box protein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532411 349.00
Check your price
  • FOXA1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527157
  • Size 100µL
Applications
ChIP assay (Assay-dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
Transcription factor 3A; Forkhead box protein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527157 349.00
Check your price
  • FOXA2 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515542
  • Size 100µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Transcription factor 3B; Forkhead box protein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515542 349.00
Check your price
  • FOXA2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518169
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Transcription factor 3B; Forkhead box protein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518169 349.00
Check your price
  • FOXA2 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131936
  • Size 100µg
Applications
ELISA (Assay-dependent), Western Blot (1-3 µg/ml)
Alternate names:
Transcription factor 3B; Forkhead box protein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131936 475.00
Check your price
  • FOXA2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527158
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Transcription factor 3B; Forkhead box protein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527158 349.00
Check your price
  • FOXA2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519518
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Forkhead box protein A2; Transcription factor 3B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519518 349.00
Check your price
  • FOXA2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535033
  • Size 400µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
Transcription factor 3B; Forkhead box protein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535033 349.00
Check your price