1 - 3 of 3 products displayed
  • ErbB2 (HER-2) Antibody (Monoclonal, SP3)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516348
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516348 700.00
Check your price
  • ErbB2 (HER-2) Antibody (Monoclonal, SP3)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA139544
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA139544 924.00
Check your price
  • ErbB2 (HER-2) Antibody (Monoclonal, SP3)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514509
  • Size 500 µL
Applications
Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (1:100)
Alternate names:
p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514509 650.00
Check your price