1 - 7 of 7 products displayed
  • KCNH2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533867
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
Voltage-gated potassium channel subunit Kv11.1; Ether-a-go-go-related gene potassium channel 1; Eag homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533867 349.00
Check your price
  • KCNH2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533866
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
Voltage-gated potassium channel subunit Kv11.1; Ether-a-go-go-related gene potassium channel 1; Eag homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533866 349.00
Check your price
  • KCNH2 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA3860
  • Size 100 µL
Applications
Immunocytochemistry (1:3000), Immunofluorescence (1:3,000), Immunoprecipitation (1:100), Western Blot (1:20,000)
Alternate names:
Voltage-gated potassium channel subunit Kv11.1; Ether-a-go-go-related gene potassium channel 1; Eag homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3860 349.00
Check your price
  • KCNH7 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526177
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Voltage-gated potassium channel subunit Kv11.3; Ether-a-go-go-related gene potassium channel 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526177 349.00
Check your price
  • ALG10B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525540
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Potassium channel regulator 1; Asparagine-linked glycosylation protein 10 homolog B; Alpha-2-glucosyltransferase ALG10-B; Alpha-1,2-glucosyltransferase ALG10-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525540 349.00
Check your price
  • KCNH6 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541061
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1.25 µg/ml)
Alternate names:
Voltage-gated potassium channel subunit Kv11.2; Ether-a-go-go-related gene potassium channel 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541061 309.00
Check your price
  • KCNH7 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532079
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Voltage-gated potassium channel subunit Kv11.3; Ether-a-go-go-related gene potassium channel 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532079 349.00
Check your price