1 - 2 of 2 products displayed
  • HSV Type 2 gG Monoclonal Antibody for Western Blot, IF, IHC (F) and IP
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516575
  • Size 1mg
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
HSV2, HSV-2, Human herpes virus 2, HHV-2, Herpes Simplex II, HSV Type 2, Herpes Simplex 2, HSV Type II, Glycoprotein G
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516575 375.00
Check your price
  • HSV Type 2 gG Monoclonal Antibody for IF
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17262
  • Size 100µg
Applications
Immunofluorescence (1:10)
Alternate names:
HSV2, HSV-2, Human herpes virus 2, HHV-2, Herpes Simplex II, HSV Type 2, Herpes Simplex 2, HSV Type II, Glycoprotein G
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17262 309.00
Check your price