1 - 30 of 30 products displayed
  • HK1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514789
  • Size 100 µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (1:800), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Hexokinase, tumor isozyme; Hexokinase type I; Brain form hexokinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514789 275.00
Check your price
  • HK1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA515447
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Hexokinase, tumor isozyme; Hexokinase type I; Brain form hexokinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515447 349.00
Check your price
  • HK1 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515680
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1/200-1/1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Hexokinase type I; Brain form hexokinase; Hexokinase, tumor isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515680 349.00
Check your price
  • HK1 Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515675
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Hexokinase type I; Brain form hexokinase; Hexokinase, tumor isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515675 349.00
Check your price
  • HK1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515448
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Hexokinase type I; Brain form hexokinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515448 349.00
Check your price
  • Hexokinase Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Rabbit
  • Target Species Yeast
  • Conjugate HRP
  • Catalog # PA127232
  • Size 1 mg
Applications
ELISA (1:4000-1:20,000), Western Blot (1:500-1:2000)
Alternate names:
Hexokinase-A; Hexokinase PI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127232 349.00
Check your price
  • HK1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528217
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Hexokinase type I; Brain form hexokinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528217 349.00
Check your price
  • HK2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515443
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Muscle form hexokinase; Hexokinase type II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515443 349.00
Check your price
  • HK2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Horse, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529326
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
Hexokinase type II; Muscle form hexokinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529326 349.00
Check your price
  • HK2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA517257
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Muscle form hexokinase; Hexokinase type II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517257 275.00
Check your price
  • HK2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515444
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Muscle form hexokinase; Hexokinase type II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515444 349.00
Check your price
  • HK3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528666
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Hexokinase type III
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528666 349.00
Check your price
  • HK3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529304
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Hexokinase type III
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529304 349.00
Check your price
  • HK3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515442
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Hexokinase type III
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515442 349.00
Check your price
  • HK3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA515441
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Hexokinase type III
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515441 349.00
Check your price
  • GCK Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA515072
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Hexokinase-D; Hexokinase-4; Hexokinase type IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515072 349.00
Check your price
  • HK2 Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA514849
  • Size 100 µL
Applications
Immunocytochemistry (1:1600), Immunofluorescence (1:1600), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:1000)
Alternate names:
Hexokinase type II; Muscle form hexokinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514849 275.00
Check your price
  • HK2 Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515679
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Muscle form hexokinase; Hexokinase type II
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515679 349.00
Check your price
  • HK2 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700422
  • Size 100 µg
Applications
Western Blot (4-6 ug/ml)
Alternate names:
Muscle form hexokinase; Hexokinase type II
Catalog # Size Price (AUD) Your Price (AUD) Qty
700422 309.00
Check your price
  • GCK Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515878
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Hexokinase-D; Hexokinase-4; Hexokinase type IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515878 349.00
Check your price
  • GCK Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515073
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Hexokinase-D; Hexokinase-4; Hexokinase type IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515073 349.00
Check your price
  • GCKR Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519103
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
GCKR; glucokinase (hexokinase 4) regulator; GKRP; glucokinase (hexokinase 4) regulatory protein; glucokinase regulator; glucokinase regulatory protein; hexokinase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519103 349.00
Check your price
  • GCK Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529325
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Hexokinase-D; Hexokinase-4; Hexokinase type IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529325 349.00
Check your price
  • GCK Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517711
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Hexokinase-D; Hexokinase-4; Hexokinase type IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517711 275.00
Check your price
  • HKDC1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535894
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000)
Alternate names:
Hexokinase domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535894 309.00
Check your price
  • TIGAR Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520346
  • Size 100 ug
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
TP53-induced glycolysis regulatory phosphatase; TP53-induced glycolysis and apoptosis regulator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520346 349.00
Check your price
  • TIGAR Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA141382
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (1-4 µg/ml)
Alternate names:
TP53-induced glycolysis regulatory phosphatase; TP53-induced glycolysis and apoptosis regulator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141382 309.00
Check your price
  • TIGAR Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520347
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
TP53-induced glycolysis regulatory phosphatase; TP53-induced glycolysis and apoptosis regulator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520347 349.00
Check your price
  • TIGAR Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Dog, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA141487
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
TP53-induced glycolysis regulatory phosphatase; TP53-induced glycolysis and apoptosis regulator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141487 309.00
Check your price
  • TIGAR Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523053
  • Size 100 µL
Applications
Immunohistochemistry (1:10-1:500), Western Blot (1:5000)
Alternate names:
TP53-induced glycolysis regulatory phosphatase; TP53-induced glycolysis and apoptosis regulator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523053 349.00
Check your price