1 - 7 of 7 products displayed
  • HK3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA515441
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Hexokinase type III
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515441 349.00
Check your price
  • HK3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528666
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Hexokinase type III
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528666 349.00
Check your price
  • HK3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529304
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Hexokinase type III
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529304 349.00
Check your price
  • HK3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515442
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Hexokinase type III
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515442 349.00
Check your price
  • HK2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515443
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Muscle form hexokinase; Hexokinase type II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515443 349.00
Check your price
  • HK2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515444
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Muscle form hexokinase; Hexokinase type II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515444 349.00
Check your price
  • HK2 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700422
  • Size 100µg
Applications
Western Blot (4-6 ug/ml)
Alternate names:
Muscle form hexokinase; Hexokinase type II
Catalog # Size Price (AUD) Your Price (AUD) Qty
700422 309.00
Check your price