1 - 2 of 2 products displayed
  • HEYL Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529920
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Hairy-related transcription factor 3; Class B basic helix-loop-helix protein 33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529920 349.00
Check your price
  • HEYL Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535185
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Hairy-related transcription factor 3; Class B basic helix-loop-helix protein 33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535185 349.00
Check your price