1 - 30 of 30 products displayed
  • c-Met Polyclonal Antibody for Western Blot, IHC, ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA120184
  • Size 25µg
Applications
ELISA (0.5-1 µg/ml), Immunohistochemistry (Assay dependent), Neutralization (0.3-1 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
AUTS9, HGFR, RCCP2, c-Met
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120184 349.00
Check your price
  • c-Met Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516352
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516352 499.00
Check your price
  • Non-Phospho-c-Met (Tyr1003) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA532358
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532358 Not Orderable Online
Check your price
  • c-Met Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532357
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532357 349.00
Check your price
  • Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700139
  • Size 100µl
Applications
ELISA (1:100), Western Blot (1:2500-1:5000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
700139 309.00
Check your price
  • c-Met Monoclonal Antibody for Western Blot, IF, ICC, Flow and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700261
  • Size 100µg
Applications
ELISA (1-5 ug/ml), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:5000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
700261 309.00
Check your price
  • Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA128463
  • Size 50µL
Applications
ELISA (Assay-dependent), Western Blot (1:1000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128463 349.00
Check your price
  • Phospho-c-Met (Tyr1349) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA128464
  • Size 50µL
Applications
Western Blot (0.1-1 µg/ml)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128464 349.00
Check your price
  • Phospho-c-Met (Tyr1003) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA128461
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128461 349.00
Check your price
  • c-Met Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129042
  • Size 7mL
Applications
Immunohistochemistry (Paraffin) (Neat)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129042 425.00
Check your price
  • c-Met Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527235
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527235 349.00
Check your price
  • Phospho-c-Met (Tyr1365) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA128465
  • Size 50µL
Applications
Western Blot (0.1-1 µg/ml)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128465 349.00
Check your price
  • c-Met Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA128026
  • Size 25µg
Applications
ELISA (0.5-1 µg/ml), Neutralization (0.5-2 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128026 349.00
Check your price
  • Phospho-c-Met (Tyr1356) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA540218
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
MET; Hepatocyte growth factor receptor; HGF receptor; HGF/SF receptor; Proto-oncogene c-Met; Scatter factor receptor; SF receptor; Tyrosine-protein kinase Met
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540218 309.00
Check your price
  • Phospho-c-Met (Tyr1234, Tyr1235) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515083
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515083 309.00
Check your price
  • Phospho-c-Met (Tyr1349) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515082
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515082 309.00
Check your price
  • Phospho-c-Met (Tyr1349) Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA512629
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512629 349.00
Check your price
  • c-Met Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515198
  • Size 100µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515198 275.00
Check your price
  • c-Met Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA517148
  • Size 100µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517148 275.00
Check your price
  • Phospho-c-Met (Tyr1349) Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710792
  • Size 100µg
Applications
Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
710792 309.00
Check your price
  • Phospho-c-Met (Tyr1349) Monoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701760
  • Size 100µg
Applications
Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
701760 309.00
Check your price
  • Phospho-Met pTyr1234 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # PA535767
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535767 Not Orderable Online
Check your price
  • Phospho-c-Met (Tyr1003) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44882G
  • Size 10blots
Applications
Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (Assay Dependent)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
44882G 309.00
Check your price
  • c-Met Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 370100
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
370100 309.00
Check your price
  • c-Met Monoclonal Antibody for IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 187366
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
187366 357.00
Check your price
  • c-Met Polyclonal Antibody for Western Blot, IHC (P), Flow and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 718000
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1:100-1:500), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
718000 304.00
Check your price
  • Phospho-c-Met (Tyr1003) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA514944
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514944 309.00
Check your price
  • Phospho-c-Met (Tyr1234, Tyr1235) Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517770
  • Size 100µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517770 309.00
Check your price
  • c-Met Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA514847
  • Size 100µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514847 275.00
Check your price
  • Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44888G
  • Size 10blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
44888G 304.00
Check your price