1 - 30 of 30 products displayed
  • HIF-1 beta Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), IP, ChIP and GS
  • Host Rabbit
  • Target Species Bovine, Ferret, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA316508
  • Size 100µL
Applications
ChIP assay (1:10-1:500), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:150), Immunoprecipitation (1:10-1:500), Western Blot (1:2000)
Alternate names:
Hypoxia-inducible factor 1-beta; Dioxin receptor, nuclear translocator; Class E basic helix-loop-helix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316508 375.00
Check your price
  • HIF-1 beta Monoclonal Antibody, DyLight® 650 conjugate for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate DyLight® 650
  • Catalog # MA516012
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 1-beta; Dioxin receptor, nuclear translocator; Class E basic helix-loop-helix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516012 375.00
Check your price
  • HIF-1 beta Monoclonal Antibody, DyLight® 550 conjugate for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate DyLight® 550
  • Catalog # MA516015
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 1-beta; Dioxin receptor, nuclear translocator; Class E basic helix-loop-helix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516015 375.00
Check your price
  • HIF-1 beta Monoclonal Antibody, DyLight® 488 conjugate for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate DyLight® 488
  • Catalog # MA516013
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 1-beta; Dioxin receptor, nuclear translocator; Class E basic helix-loop-helix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516013 375.00
Check your price
  • HIF-1 beta Monoclonal Antibody, Biotin conjugate for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate Biotin
  • Catalog # MA516011
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 1-beta; Dioxin receptor, nuclear translocator; Class E basic helix-loop-helix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516011 375.00
Check your price
  • HIF-1 beta Monoclonal Antibody, HRP conjugate for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate HRP
  • Catalog # MA516014
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 1-beta; Dioxin receptor, nuclear translocator; Class E basic helix-loop-helix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516014 375.00
Check your price
  • HIF-1 beta Monoclonal Antibody for IF, ICC, IHC, IHC (P), ChIP and GS
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # MA116512
  • Size 100µL
Applications
ChIP assay (1:10-1:500), Gel Shift (Assay-Dependent), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (Assay-Dependent), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 1-beta; Dioxin receptor, nuclear translocator; Class E basic helix-loop-helix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116512 349.00
Check your price
  • HIF-1 beta Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and GS
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1515
  • Size 100µL
Applications
Gel Shift (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (1:1,000), Immunohistochemistry (Paraffin) (5 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:3000)
Alternate names:
Hypoxia-inducible factor 1-beta; Dioxin receptor, nuclear translocator; Class E basic helix-loop-helix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1515 375.00
Check your price
  • HIF1A Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), Flow and ChIP
  • Host Rabbit
  • Target Species Goat, Hamster, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA116601
  • Size 100µL
Applications
ChIP assay (Assay-Dependent), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Frozen) (5-10 µg/ml), Immunohistochemistry (Paraffin) (5-10 µg/ml), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116601 375.00
Check your price
  • HIF1A Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), IP, GS and IE
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Non-human primate, Pig
  • Conjugate Unconjugated
  • Catalog # MA1516
  • Size 100µL
Applications
Gel Shift (Assay dependent), Immunocytochemistry (1:20-1:200), Immunoelectrophoresis (Assay Dependent), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay dependent), Western Blot (1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1516 375.00
Check your price
  • HIF1A Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA116518
  • Size 100µL
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Immunohistochemistry (Frozen) (1:100-1:5000), Immunohistochemistry (Paraffin) (1:100-1:5000), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116518 375.00
Check your price
  • HIF1A Monoclonal Antibody, Biotin conjugate for Western Blot, IHC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA516016
  • Size 100µL
Applications
Immunohistochemistry (Frozen) (1:100-1:5000), Immunohistochemistry (Paraffin) (1:100-1:5000), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516016 375.00
Check your price
  • HIF1A Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Goat, Hamster, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate DyLight® 650
  • Catalog # PA522777
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522777 375.00
Check your price
  • HIF1A Polyclonal Antibody, DyLight® 488 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Goat, Hamster, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate DyLight® 488
  • Catalog # PA522778
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522778 375.00
Check your price
  • HIF1A Monoclonal Antibody, DyLight® 550 conjugate for IF, ICC, IHC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate DyLight® 550
  • Catalog # MA516020
  • Size 100µL
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Immunohistochemistry (Frozen) (1:100-1:5000), Immunohistochemistry (Paraffin) (1:100-1:5000), Immunoprecipitation (1:10-1:500)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516020 375.00
Check your price
  • HIF1A Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), IP, ChIP and GS
  • Host Rabbit
  • Target Species Dog, Guinea pig, Human, Mouse, Non-human primate, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA316521
  • Size 100µL
Applications
ChIP assay (25 µl/1.5x10^7 cells), Gel Shift (1:1-1:100), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (1:10-1:500), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:1000), Western Blot (1:500-1:1000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316521 375.00
Check your price
  • HIF1A Polyclonal Antibody for IF, ICC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA116627
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:1000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116627 375.00
Check your price
  • HIF1A Polyclonal Antibody, DyLight® 550 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Goat, Hamster, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate DyLight® 550
  • Catalog # PA522780
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522780 375.00
Check your price
  • HIF1A Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA116517
  • Size 100µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1; ARNT-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116517 349.00
Check your price
  • HIF1A Polyclonal Antibody, HRP conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Goat, Hamster, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate HRP
  • Catalog # PA522779
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522779 375.00
Check your price
  • HIF1A Monoclonal Antibody, DyLight® 650 conjugate for IF, ICC, IHC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate DyLight® 650
  • Catalog # MA516017
  • Size 100µL
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Immunohistochemistry (Frozen) (1:100-1:5000), Immunohistochemistry (Paraffin) (1:100-1:5000), Immunoprecipitation (1:10-1:500)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516017 375.00
Check your price
  • HIF1A Monoclonal Antibody, DyLight® 488 conjugate for IF, ICC, IHC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate DyLight® 488
  • Catalog # MA516018
  • Size 100µL
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Immunohistochemistry (Frozen) (1:100-1:5000), Immunohistochemistry (Paraffin) (1:100-1:5000), Immunoprecipitation (1:10-1:500)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516018 375.00
Check your price
  • HIF1A Monoclonal Antibody, HRP conjugate for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate HRP
  • Catalog # MA516019
  • Size 100µL
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Immunohistochemistry (Paraffin) (1:100-1:5000), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516019 375.00
Check your price
  • HIF1A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523031
  • Size 100µL
Applications
Western Blot (1 µg/ml)
Alternate names:
PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1; ARNT-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523031 349.00
Check your price
  • HIF1A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523032
  • Size 100µL
Applications
Western Blot (1 µg/ml)
Alternate names:
PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1; ARNT-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523032 349.00
Check your price
  • HIF1A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523030
  • Size 100µL
Applications
Western Blot (1 µg/ml)
Alternate names:
PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1; ARNT-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523030 349.00
Check your price
  • HIF1A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523029
  • Size 100µL
Applications
Western Blot (1 µg/ml)
Alternate names:
PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1; ARNT-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523029 349.00
Check your price
  • HIF1A Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), Flow, IP, ChIP, ELISA and GS
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat, Xenopus, Yeast
  • Conjugate Unconjugated
  • Catalog # MA116504
  • Size 100µL
Applications
ChIP assay (1:10-1:500), ELISA (Assay-Dependent), Flow Cytometry (Assay-Dependent), Gel Shift (1:1-1:100), Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Frozen) (1:20-1:50), Immunohistochemistry (Paraffin) (1:20-1:50), Immunoprecipitation (1:10-1:500), Western Blot (1:500)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116504 375.00
Check your price
  • HIF1A Monoclonal Antibody for IF, ICC, IHC, IHC (P), Flow, IP, ChIP, ELISA and GS
  • Host Mouse
  • Target Species Avian, Bovine, Dog, Ferret, Human, Mouse, Non-human primate, Sheep, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA116511
  • Size 100µL
Applications
ChIP assay (Assay-Dependent), ELISA (Assay-Dependent), Flow Cytometry (Assay-Dependent), Gel Shift (Assay-Dependent), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:100-1:300), Immunoprecipitation (1:10), Western Blot (1:500-1:1000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116511 375.00
Check your price
  • HIF1A Monoclonal Antibody, Biotin conjugate for Western Blot, IHC, IHC (P) and IP
  • Host Mouse
  • Target Species Avian, Bovine, Dog, Ferret, Human, Mouse, Non-human primate, Sheep, Pig, Rat
  • Conjugate Biotin
  • Catalog # MA146457
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:300), Immunoprecipitation (1:10), Western Blot (1:500)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146457 325.00
Check your price