1 - 15 of 15 products displayed
  • PHD3 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), IP and IM
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116526
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunomicroscopy (Assay-Dependent), Western Blot (1:1000-1:2000)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1; SM-20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116526 349.00
Check your price
  • PHD1 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IM
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116569
  • Size 100 µg
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunomicroscopy (Assay-Dependent), Immunoprecipitation (1:50), Western Blot (1:500-1:2500)
Alternate names:
Prolyl hydroxylase domain-containing protein 1; Hypoxia-inducible factor prolyl hydroxylase 1; Falkor; HPH-3; Estrogen-induced tag 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116569 Not Orderable Online
Check your price
  • PHD3 Monoclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA516144
  • Size 200 µL
Applications
Immunohistochemistry (Paraffin) (1:400), Western Blot (10 µg/ml)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1; SM-20
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516144 349.00
Check your price
  • PHD3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA120196
  • Size 100 µL
Applications
Western Blot (1:1000-1:3000)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1; SM-20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120196 499.00
Check your price
  • PHD2 Monoclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516143
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:400), Western Blot (0.5-2 µg/ml)
Alternate names:
SM-20; Prolyl hydroxylase domain-containing protein 2; Hypoxia-inducible factor prolyl hydroxylase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516143 349.00
Check your price
  • PHD2 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA116524
  • Size 100 µg
Applications
Flow Cytometry (3 µg/ml), Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:2500)
Alternate names:
SM-20; Prolyl hydroxylase domain-containing protein 2; Hypoxia-inducible factor prolyl hydroxylase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116524 349.00
Check your price
  • PHD3 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516140
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Western Blot (1:250)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516140 349.00
Check your price
  • PHD3 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA522900
  • Size 100 µg
Applications
ELISA (Assay-Dependent)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA522900 309.00
Check your price
  • PHD3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518957
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518957 349.00
Check your price
  • PHD3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529260
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529260 349.00
Check your price
  • PHD2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517050
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
SM-20; Prolyl hydroxylase domain-containing protein 2; Hypoxia-inducible factor prolyl hydroxylase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517050 275.00
Check your price
  • PHD1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532081
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Prolyl hydroxylase domain-containing protein 1; Hypoxia-inducible factor prolyl hydroxylase 1; HPH-3; Estrogen-induced tag 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532081 349.00
Check your price
  • HIF1A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523031
  • Size 100 µL
Applications
Western Blot (1 µg/ml)
Alternate names:
PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1; ARNT-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523031 349.00
Check your price
  • HIF1A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523030
  • Size 100 µL
Applications
Western Blot (1 µg/ml)
Alternate names:
PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1; ARNT-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523030 349.00
Check your price
  • SIAH2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536302
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Seven in absentia homolog 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536302 309.00
Check your price