1 - 30 of 30 products displayed
  • P4HTM Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530753
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Hypoxia-inducible factor prolyl hydroxylase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530753 349.00
Check your price
  • P4HTM Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530752
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Hypoxia-inducible factor prolyl hydroxylase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530752 349.00
Check your price
  • P4HTM Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IM
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA116560
  • Size 100µg
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunomicroscopy (Assay-Dependent), Western Blot (1:500-1:2500)
Alternate names:
Hypoxia-inducible factor prolyl hydroxylase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116560 349.00
Check your price
  • PHD3 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), IP and IM
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116526
  • Size 100µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunomicroscopy (Assay-Dependent), Western Blot (1:1000-1:2000)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1; SM-20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116526 349.00
Check your price
  • PHD2 Polyclonal Antibody for IF, ICC, IHC, IHC (P), Flow, IP and IM
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA116525
  • Size 100µL
Applications
Flow Cytometry (1:10-1:1000), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunomicroscopy (Assay-Dependent), Western Blot (1:500-1:2500)
Alternate names:
SM-20; Prolyl hydroxylase domain-containing protein 2; Hypoxia-inducible factor prolyl hydroxylase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116525 349.00
Check your price
  • PHD1 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IM
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116569
  • Size 100µg
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunomicroscopy (Assay-Dependent), Immunoprecipitation (1:50), Western Blot (1:500-1:2500)
Alternate names:
Prolyl hydroxylase domain-containing protein 1; Hypoxia-inducible factor prolyl hydroxylase 1; Falkor; HPH-3; Estrogen-induced tag 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116569 Not Orderable Online
Check your price
  • PHD2 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA116524
  • Size 100µg
Applications
Flow Cytometry (3 µg/ml), Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:2500)
Alternate names:
SM-20; Prolyl hydroxylase domain-containing protein 2; Hypoxia-inducible factor prolyl hydroxylase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116524 349.00
Check your price
  • PHD2 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116946
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (2.5 µg/ml), Immunoprecipitation (1:10-1:500), Western Blot (2 µg/ml)
Alternate names:
SM-20; Prolyl hydroxylase domain-containing protein 2; Hypoxia-inducible factor prolyl hydroxylase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116946 349.00
Check your price
  • PHD2 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA133195
  • Size 25µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
SM-20; Prolyl hydroxylase domain-containing protein 2; Hypoxia-inducible factor prolyl hydroxylase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA133195 425.00
Check your price
  • PHD3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA120196
  • Size 100µL
Applications
Western Blot (1:1000-1:3000)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1; SM-20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120196 499.00
Check your price
  • PHD2 Monoclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516143
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:400), Western Blot (0.5-2 µg/ml)
Alternate names:
SM-20; Prolyl hydroxylase domain-containing protein 2; Hypoxia-inducible factor prolyl hydroxylase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516143 349.00
Check your price
  • PHD2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517050
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
SM-20; Prolyl hydroxylase domain-containing protein 2; Hypoxia-inducible factor prolyl hydroxylase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517050 275.00
Check your price
  • PHD1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532081
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Prolyl hydroxylase domain-containing protein 1; Hypoxia-inducible factor prolyl hydroxylase 1; HPH-3; Estrogen-induced tag 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532081 349.00
Check your price
  • HIF-1 beta Monoclonal Antibody, DyLight® 650 conjugate for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate DyLight® 650
  • Catalog # MA516012
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 1-beta; Dioxin receptor, nuclear translocator; Class E basic helix-loop-helix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516012 375.00
Check your price
  • HIF-1 beta Monoclonal Antibody, DyLight® 550 conjugate for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate DyLight® 550
  • Catalog # MA516015
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 1-beta; Dioxin receptor, nuclear translocator; Class E basic helix-loop-helix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516015 375.00
Check your price
  • HIF-1 beta Monoclonal Antibody, DyLight® 488 conjugate for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate DyLight® 488
  • Catalog # MA516013
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 1-beta; Dioxin receptor, nuclear translocator; Class E basic helix-loop-helix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516013 375.00
Check your price
  • HIF-1 beta Monoclonal Antibody, Biotin conjugate for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate Biotin
  • Catalog # MA516011
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 1-beta; Dioxin receptor, nuclear translocator; Class E basic helix-loop-helix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516011 375.00
Check your price
  • HIF-1 beta Monoclonal Antibody, HRP conjugate for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate HRP
  • Catalog # MA516014
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 1-beta; Dioxin receptor, nuclear translocator; Class E basic helix-loop-helix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516014 375.00
Check your price
  • PHD3 Monoclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA516144
  • Size 200µL
Applications
Immunohistochemistry (Paraffin) (1:400), Western Blot (10 µg/ml)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1; SM-20
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516144 349.00
Check your price
  • PHD3 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516140
  • Size 100µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Western Blot (1:250)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516140 349.00
Check your price
  • PHD3 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA522900
  • Size 100µg
Applications
ELISA (Assay-Dependent)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA522900 309.00
Check your price
  • PHD3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518957
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518957 349.00
Check your price
  • PHD3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529260
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529260 349.00
Check your price
  • HIF1A Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), Flow and ChIP
  • Host Rabbit
  • Target Species Goat, Hamster, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA116601
  • Size 100µL
Applications
ChIP assay (Assay-Dependent), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Frozen) (5-10 µg/ml), Immunohistochemistry (Paraffin) (5-10 µg/ml), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116601 375.00
Check your price
  • HIF1A Monoclonal Antibody, Biotin conjugate for Western Blot, IHC, IHC (P) and IP
  • Host Mouse
  • Target Species Avian, Bovine, Dog, Ferret, Human, Mouse, Non-human primate, Sheep, Pig, Rat
  • Conjugate Biotin
  • Catalog # MA146457
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:300), Immunoprecipitation (1:10), Western Blot (1:500)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146457 325.00
Check your price
  • HIF1A Monoclonal Antibody for IF, ICC, IHC, IHC (P), Flow, IP, ChIP, ELISA and GS
  • Host Mouse
  • Target Species Avian, Bovine, Dog, Ferret, Human, Mouse, Non-human primate, Sheep, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA116511
  • Size 100µL
Applications
ChIP assay (Assay-Dependent), ELISA (Assay-Dependent), Flow Cytometry (Assay-Dependent), Gel Shift (Assay-Dependent), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:100-1:300), Immunoprecipitation (1:10), Western Blot (1:500-1:1000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116511 375.00
Check your price
  • HIF1A Monoclonal Antibody, Biotin conjugate for Western Blot, IHC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA516016
  • Size 100µL
Applications
Immunohistochemistry (Frozen) (1:100-1:5000), Immunohistochemistry (Paraffin) (1:100-1:5000), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516016 375.00
Check your price
  • HIF1A Monoclonal Antibody, HRP conjugate for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate HRP
  • Catalog # MA516019
  • Size 100µL
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Immunohistochemistry (Paraffin) (1:100-1:5000), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516019 375.00
Check your price
  • HIF1A Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), Flow, IP, ChIP, ELISA and GS
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat, Xenopus, Yeast
  • Conjugate Unconjugated
  • Catalog # MA116504
  • Size 100µL
Applications
ChIP assay (1:10-1:500), ELISA (Assay-Dependent), Flow Cytometry (Assay-Dependent), Gel Shift (1:1-1:100), Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Frozen) (1:20-1:50), Immunohistochemistry (Paraffin) (1:20-1:50), Immunoprecipitation (1:10-1:500), Western Blot (1:500)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116504 375.00
Check your price
  • HIF1A Monoclonal Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC, IHC, IHC (F) and ChIP
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat, Xenopus, Yeast
  • Conjugate DyLight® 650
  • Catalog # MA516008
  • Size 100µL
Applications
ChIP assay (Assay-Dependent), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:100), Immunoprecipitation (Assay-Dependent), Western Blot (1:500)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516008 375.00
Check your price