1 - 24 of 24 products displayed
  • HIF-2 alpha Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519494
  • Size 100 µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1:2500), Western Blot (1 µg/ml)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519494 349.00
Check your price
  • HIF-2 alpha Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524200
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
PAS domain-containing protein 2; Member of PAS protein 2; Hypoxia-inducible factor 2-alpha; HIF-1-alpha-like factor; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524200 349.00
Check your price
  • HIF-2 alpha Antibody, HRP (Monoclonal, ep190b)
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate HRP
  • Catalog # MA516023
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:150-1:300), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516023 375.00
Check your price
  • HIF-2 alpha Antibody, Biotin
  • Host Rabbit
  • Target Species Fish, Hamster, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate Biotin
  • Catalog # PA522693
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:200-1:1000)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522693 375.00
Check your price
  • HIF-2 alpha Antibody, HRP
  • Host Rabbit
  • Target Species Fish, Hamster, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate HRP
  • Catalog # PA522696
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:200-1:1000)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522696 375.00
Check your price
  • HIF-2 alpha Antibody, Biotin (Monoclonal, ep190b)
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA146459
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:150-1:300), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146459 325.00
Check your price
  • HIF-2 alpha Antibody (Monoclonal, ep190b)
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA116519
  • Size 100 µL
Applications
ChIP assay (Assay-Dependent), ELISA (1:100-1:2000), Flow Cytometry (1:400), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:150-1:300), Immunoprecipitation (Assay-Dependent), Western Blot (1-2 µg/ml)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116519 375.00
Check your price
  • HIF-2 alpha Antibody
  • Host Rabbit
  • Target Species Fish, Hamster, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA116510
  • Size 100 µL
Applications
ChIP assay (1:10-1:500), ELISA (1:100-1:2000), Flow Cytometry (Assay-Dependent), Gel Shift (Assay-Dependent), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (5 µg/1mg lysate), Western Blot
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116510 375.00
Check your price
  • HIF-2 alpha Antibody, DyLight® 550 (Monoclonal, ep190b)
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate DyLight® 550
  • Catalog # MA516024
  • Size 100 µL
Applications
Flow Cytometry (1 µg), Immunohistochemistry (Paraffin) (1:150-1:300), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516024 375.00
Check your price
  • HIF-2 alpha Antibody, DyLight® 650 (Monoclonal, ep190b)
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate DyLight® 650
  • Catalog # MA516021
  • Size 100 µL
Applications
Flow Cytometry (1 µg), Immunohistochemistry (Paraffin) (1:150-1:300), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516021 375.00
Check your price
  • HIF-2 alpha Antibody, DyLight® 488 (Monoclonal, ep190b)
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate DyLight® 488
  • Catalog # MA516022
  • Size 100 µL
Applications
Flow Cytometry (1 µg), Immunohistochemistry (Paraffin) (1:150-1:300), Western Blot (1:500)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516022 375.00
Check your price
  • HIF-2 alpha Antibody, DyLight® 650
  • Host Rabbit
  • Target Species Fish, Hamster, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate DyLight® 650
  • Catalog # PA522694
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:200-1:1000)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522694 375.00
Check your price
  • HIF-2 alpha Antibody, DyLight® 550
  • Host Rabbit
  • Target Species Fish, Hamster, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate DyLight® 550
  • Catalog # PA522697
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:200-1:1000)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522697 375.00
Check your price
  • HIF-2 alpha Antibody, DyLight® 488
  • Host Rabbit
  • Target Species Fish, Hamster, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate DyLight® 488
  • Catalog # PA522695
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:200-1:1000)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522695 375.00
Check your price
  • HIF3A Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116952
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7; PAS domain-containing protein 7; Class E basic helix-loop-helix protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116952 375.00
Check your price
  • HIF3A Antibody, DyLight® 550
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 550
  • Catalog # PA522751
  • Size 100 µL
Applications
Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7; PAS domain-containing protein 7; Class E basic helix-loop-helix protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522751 375.00
Check your price
  • HIF3A Antibody, DyLight® 488
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 488
  • Catalog # PA522749
  • Size 100 µL
Applications
Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7; PAS domain-containing protein 7; Class E basic helix-loop-helix protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522749 375.00
Check your price
  • HIF3A Antibody, Biotin
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # PA522747
  • Size 100 µL
Applications
Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7; PAS domain-containing protein 7; Class E basic helix-loop-helix protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522747 375.00
Check your price
  • HIF3A Antibody, HRP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate HRP
  • Catalog # PA522750
  • Size 100 µL
Applications
Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7; PAS domain-containing protein 7; Class E basic helix-loop-helix protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522750 375.00
Check your price
  • HIF3A Antibody, DyLight® 650
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 650
  • Catalog # PA522715
  • Size 100 µL
Applications
Western Blot (1:100-1:200)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522715 375.00
Check your price
  • HIF3A Antibody, DyLight® 550
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 550
  • Catalog # PA522718
  • Size 100 µL
Applications
Western Blot (1:100-1:200)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522718 375.00
Check your price
  • HIF3A Antibody, DyLight® 488
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 488
  • Catalog # PA522716
  • Size 100 µL
Applications
Western Blot (1:100-1:200)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522716 375.00
Check your price
  • HIF3A Antibody, Biotin
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # PA522714
  • Size 100 µL
Applications
Western Blot (1:100-1:200)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522714 375.00
Check your price
  • HIF3A Antibody, HRP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate HRP
  • Catalog # PA522717
  • Size 100 µL
Applications
Western Blot (1:100-1:200)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522717 375.00
Check your price