1 - 30 of 30 products displayed
  • HRH1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533843
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (4 µg/ml)
Alternate names:
HRH1, H1 histamine receptor, H1 receptor, Histamine 1 receptor, Histamine H1 receptor, H1-R, H1R, Histamine H(1) receptor, Histamine receptor H1, HisH1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533843 349.00
Check your price
  • HRH1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533842
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (11 µg/ml)
Alternate names:
HRH1, H1 histamine receptor, H1 receptor, Histamine 1 receptor, Histamine H1 receptor, H1-R, H1R, Histamine H(1) receptor, Histamine receptor H1, HisH1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533842 349.00
Check your price
  • Phospho-HRH1 (Ser398) Polyclonal Antibody for IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537597
  • Size 100 µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200)
Alternate names:
HRH1, H1 histamine receptor, H1 receptor, Histamine 1 receptor, Histamine H1 receptor, H1-R, H1R, Histamine H(1) receptor, Histamine receptor H1, HisH1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537597 309.00
Check your price
  • HRH4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533850
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (12-24 µg/ml)
Alternate names:
SP9144; Pfi-013; GPRv53; G-protein coupled receptor 105; AXOR35
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533850 349.00
Check your price
  • HRH4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533852
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (14 µg/ml)
Alternate names:
SP9144; Pfi-013; GPRv53; G-protein coupled receptor 105; AXOR35
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533852 349.00
Check your price
  • HRH4 Polyclonal Antibody for ICC, IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533851
  • Size 50 ug
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (5-10 ug/ml)
Alternate names:
SP9144; Pfi-013; GPRv53; G-protein coupled receptor 105; AXOR35
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533851 349.00
Check your price
  • HRH4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533849
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (3.6 µg/ml)
Alternate names:
SP9144; Pfi-013; GPRv53; G-protein coupled receptor 105; AXOR35
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533849 349.00
Check your price
  • HRH3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533846
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (7-15 µg/ml)
Alternate names:
G-protein coupled receptor 97
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533846 349.00
Check your price
  • HRH2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533844
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (38 µg/ml)
Alternate names:
Gastric receptor I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533844 349.00
Check your price
  • HRH2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533845
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (18 µg/ml)
Alternate names:
Gastric receptor I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533845 349.00
Check your price
  • HRH3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533847
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (2-8 µg/ml), Western Blot (Assay Dependent)
Alternate names:
G-protein coupled receptor 97
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533847 349.00
Check your price
  • HRH3 Polyclonal Antibody for ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533848
  • Size 50 ug
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (18-36 µg/ml)
Alternate names:
G-protein coupled receptor 97
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533848 349.00
Check your price
  • HRH1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527817
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
H1-R, hisH1, H1R, HH1R, histamine H(1) receptor, histamine H1 receptor, histamine receptor, subclass H1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527817 349.00
Check your price
  • HRH1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518680
  • Size 100 ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
histamine receptor H1; H1-R; HRH1; hisH1 ; histamine receptor, subclass H1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518680 349.00
Check your price
  • HRH1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OPA115039
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
HRH1
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA115039 399.00
Check your price
  • Endothelin A Receptor Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530589
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Endothelin A receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530589 349.00
Check your price
  • DRD4 Polyclonal Antibody for Western Blot, IF and IHC (F)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528756
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Dopamine D4 receptor; D(2C) dopamine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528756 349.00
Check your price
  • TAAR1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523141
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
RP11-295F4.9, TA1, TAR1, TRAR1, taR-1, trace amine receptor 1, trace amine-associated receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523141 349.00
Check your price
  • HRH4 Polyclonal Antibody for IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124553
  • Size 25 µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent)
Alternate names:
SP9144; Pfi-013; GPRv53; G-protein coupled receptor 105; AXOR35
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124553 600.00
Check your price
  • HRH4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521313
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
SP9144; Pfi-013; GPRv53; G-protein coupled receptor 105; AXOR35
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521313 349.00
Check your price
  • DRD4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533483
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
Dopamine D4 receptor; D(2C) dopamine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533483 349.00
Check your price
  • DRD4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533484
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (13 µg/ml)
Alternate names:
Dopamine D4 receptor; D(2C) dopamine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533484 349.00
Check your price
  • GABRA1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527985
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
GABA(A) receptor subunit alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527985 349.00
Check your price
  • GABRB1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528167
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
GABA(A) receptor subunit beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528167 349.00
Check your price
  • HRH3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521312
  • Size 100 µL
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
G-protein coupled receptor 97
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521312 349.00
Check your price
  • DRD3 Polyclonal Antibody for IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532737
  • Size 50 ug
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (20 µg/ml)
Alternate names:
Dopamine D3 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532737 349.00
Check your price
  • DRD3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533482
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (16-32 µg/ml)
Alternate names:
Dopamine D3 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533482 349.00
Check your price
  • HRH2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124510
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
Gastric receptor I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124510 399.00
Check your price
  • HRH2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521907
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Gastric receptor I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521907 349.00
Check your price
  • GABRA1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA184892
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
GABA(A) receptor subunit alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184892 550.00
Check your price