1 - 30 of 30 products displayed
  • 6x-His Tag Monoclonal Antibody, FITC conjugate for IF and Flow
  • Host Mouse
  • Target Species Tag
  • Conjugate FITC
  • Catalog # MA181891
  • Size 100 µg
Applications
Flow Cytometry (1/10), Immunofluorescence (Assay dependent)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181891 309.00
Check your price
  • Penta-His Tag Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # P21315
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
pentahistidine, Histidine 5X, 5Xhis
Catalog # Size Price (AUD) Your Price (AUD) Qty
P21315 674.00
Check your price
  • HDC Polyclonal Antibody for IF, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Dog, Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA136090
  • Size 50 µL
Applications
Immunofluorescence (1:1000-1:1500), Immunohistochemistry (Paraffin, Frozen) (Assay dependent.)
Alternate names:
MGC163399
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136090 900.00
Check your price
  • 6x-His Tag Monoclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate HRP
  • Catalog # MA180218
  • Size 100 µg
Applications
ELISA (1:20-1:1000), Western Blot (1/1000 -1/5,000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180218 449.00
Check your price
  • FHIT Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532418
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Fragile histidine triad protein; Dinucleosidetriphosphatase; Diadenosine 5',5'''-P1,P3-triphosphate hydrolase; AP3A hydrolase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532418 275.00
Check your price
  • NME2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA515344
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
nm23-M2; P18; Histidine protein kinase NDKB; nm23-H2; C-myc purine-binding transcription factor PUF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515344 349.00
Check your price
  • 6x-His Tag Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # 710286
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1-3 ug/ml)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
710286 303.00
Check your price
  • HAL Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542754
  • Size 100 µl
Applications
Immunohistochemistry (1.25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Histidine ammonia-lyase, HIS, HSTD, histidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542754 309.00
Check your price
  • HAL Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542755
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1.25 µg/ml)
Alternate names:
Histidine ammonia-lyase, HIS, HSTD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542755 309.00
Check your price
  • HARS Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514510
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Histidyl-tRNA synthetase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514510 349.00
Check your price
  • FHIT Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137690
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Fragile histidine triad protein; Dinucleosidetriphosphatase; Diadenosine 5',5'''-P1,P3-triphosphate hydrolase; AP3A hydrolase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137690 375.00
Check your price
  • NME2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527488
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
nm23-M2; P18; Histidine protein kinase NDKB; nm23-H2; C-myc purine-binding transcription factor PUF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527488 349.00
Check your price
  • NME2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530602
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
nm23-M2; P18; Histidine protein kinase NDKB; nm23-H2; C-myc purine-binding transcription factor PUF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530602 349.00
Check your price
  • HisG Epitope Tag Monoclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate HRP
  • Catalog # R94125
  • Size 50 µl
Applications
ELISA (Assay Dependent), Western Blot (1:1000-1:5000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag, His G, 6xHis-Gly, 6xHis-Glycine
Catalog # Size Price (AUD) Your Price (AUD) Qty
R94125 226.00
Check your price
  • HisG Epitope Tag Monoclonal Antibody, AP conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate AP
  • Catalog # R94225
  • Size 125 µl
Applications
Western Blot (1:2000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag, His G, 6xHis-Gly, 6xHis-Glycine
Catalog # Size Price (AUD) Your Price (AUD) Qty
R94225 226.00
Check your price
  • HisG Epitope Tag Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # R94025
  • Size 50 µl
Applications
ELISA (Assay Dependent), Immunocytochemistry (2-4µg/ml), Immunofluorescence (2-4 µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (1:500-1:2000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag, His G, 6xHis-Gly, 6xHis-Glycine
Catalog # Size Price (AUD) Your Price (AUD) Qty
R94025 184.00
Check your price
  • 6x-His Tag Monoclonal Antibody, Alexa Fluor® 647 conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate Alexa Fluor® 647
  • Catalog # MA121315A647
  • Size 50 µl
Applications
Western Blot (1:500)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315A647 225.00
Check your price
  • 6x-His Tag Monoclonal Antibody, Alexa Fluor® 555 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate Alexa Fluor® 555
  • Catalog # MA121315A555
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (Assay-Dependent)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315A555 225.00
Check your price
  • 6x-His Tag Monoclonal Antibody, Alexa Fluor® 488 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate Alexa Fluor® 488
  • Catalog # MA121315A488
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (1:500)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315A488 225.00
Check your price
  • 6x-His Tag Monoclonal Antibody, DyLight® 680 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight® 680
  • Catalog # MA121315D680
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (1:500-1:1000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315D680 159.00
Check your price
  • 6x-His Tag Monoclonal Antibody, DyLight® 550 conjugate for IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight® 550
  • Catalog # MA121315D550
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315D550 159.00
Check your price
  • 6x-His Tag Monoclonal Antibody, DyLight® 488 conjugate for IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight® 488
  • Catalog # MA121315D488
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315D488 159.00
Check your price
  • 6x-His Tag Monoclonal Antibody, DyLight® 650 conjugate for IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight® 650
  • Catalog # MA121315D650
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315D650 159.00
Check your price
  • 6x-His Tag Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Biotin
  • Catalog # MA121315BTIN
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:500-1:2000), Immunofluorescence (1:500-1:2000), Immunoprecipitation (Assay Dependent), Western Blot (1:500-1000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315BTIN 159.00
Check your price
  • 6x-His Tag Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate HRP
  • Catalog # MA121315HRP
  • Size 50 µL
Applications
Western Blot (1:500-1:2000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315HRP 169.00
Check your price
  • 6x-His Tag Monoclonal Antibody, Alexa Fluor® 647 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate Alexa Fluor® 647
  • Catalog # MA1135A647
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (Assay-Dependent)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1135A647 219.00
Check your price
  • 6x-His Tag Monoclonal Antibody, Alexa Fluor® 555 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate Alexa Fluor® 555
  • Catalog # MA1135A555
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (Assay-Dependent)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1135A555 225.00
Check your price
  • 6x-His Tag Monoclonal Antibody, Alexa Fluor® 488 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate Alexa Fluor® 488
  • Catalog # MA1135A488
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (Assay-Dependent)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1135A488 219.00
Check your price
  • 6x-His Tag Polyclonal Antibody, HRP conjugate for Western Blot, ICC and ELISA
  • Host Goat
  • Target Species Tag
  • Conjugate HRP
  • Catalog # PA123024
  • Size 100 µg
Applications
ELISA (1:10,000-1:20,000), Immunocytochemistry (1:200-1:500), Western Blot (1:1000-1:5000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123024 550.00
Check your price
  • 6x-His Tag Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow, IP and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA14806
  • Size 200 µg
Applications
ELISA (0.1 to 0.4 µg/ml), Flow Cytometry (5 to 10 µl/10^6 cells), Immunofluorescence (1 to 4 µg/ml), Immunohistochemistry (Frozen) (1 to 4 µg/ml), Immunoprecipitation (4 ug/ml), Western Blot (1 to 4 µg/ml)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA14806 425.00
Check your price