1 - 30 of 30 products displayed
  • Tri-Methyl-Histone H3 (Lys27) Monoclonal Antibody for Western Blot, IF, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA511198
  • Size 100µL
Applications
ChIP assay (1:50), Immunofluorescence (1:1600), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511198 349.00
Check your price
  • Phospho-Histone H3 (Ser10) Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515220
  • Size 100µL
Applications
Flow Cytometry (1:25), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Frozen) (1:100), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515220 309.00
Check your price
  • Acetyl-Histone H3 (Lys9) Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow, IP and ChIP
  • Host Rabbit
  • Target Species C. elegans, Human, Mouse, Non-human primate, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA511195
  • Size 100µL
Applications
ChIP assay (1:50), Flow Cytometry (1:200), Immunocytochemistry (1:400), Immunofluorescence (1:400), Immunohistochemistry (Paraffin) (1:800), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511195 309.00
Check your price
  • Acetyl-Histone H3 (Lys9, Lys14) Polyclonal Antibody for Western Blot, IF, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species C. elegans, Human, Mouse, Non-human primate, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA516194
  • Size 100µL
Applications
ChIP assay (1:50), Immunofluorescence (1:5000), Immunohistochemistry (Paraffin) (1:800), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516194 349.00
Check your price
  • Di-Methyl-Histone H3 (Lys36) Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA514867
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:25), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514867 309.00
Check your price
  • Di-Methyl-Histone H3 (Lys4) Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA514977
  • Size 100µL
Applications
ChIP assay (1:50), Immunocytochemistry (1:800), Immunofluorescence (1:800), Immunohistochemistry (Paraffin) (1:1500), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514977 309.00
Check your price
  • Tri-Methyl-Histone H3 (Lys36) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517109
  • Size 100µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517109 309.00
Check your price
  • Phospho-Histone H3 (Thr11) Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA517360
  • Size 100µL
Applications
Flow Cytometry (1:25), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517360 309.00
Check your price
  • Acetyl-Histone H3 (Lys18) Polyclonal Antibody for Western Blot, IHC (P) and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517801
  • Size 100µL
Applications
ChIP assay (1-3µl), Immunohistochemistry (Paraffin) (1:800), Western Blot (1:2000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517801 309.00
Check your price
  • Methyl-Histone H3 (Lys4) Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517418
  • Size 100µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517418 309.00
Check your price
  • Phospho-Histone H3 (Thr3) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517502
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517502 309.00
Check your price
  • Histone H3 Polyclonal Antibody for Western Blot and ChIP
  • Host Rabbit
  • Target Species Bovine, Chicken, Fruit fly, Horse, Human, Mouse, Non-human primate, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA516183
  • Size 100µL
Applications
ChIP assay (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516183 309.00
Check your price
  • Tri-Methyl-Histone H3 (Lys4) Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA517420
  • Size 100µL
Applications
ChIP assay (1:100), Immunocytochemistry (1:4000), Immunofluorescence (1:4000), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517420 309.00
Check your price
  • Histone H3 Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Fruit fly, Hamster, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA515150
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515150 275.00
Check your price
  • Tri-Methyl-Histone H3 (Lys4) Monoclonal Antibody for Western Blot, IF, IHC, IHC (P) and ChIP
  • Host Rabbit
  • Target Species Fruit fly, Human, Mouse, Non-human primate, Rat, Xenopus, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA511199
  • Size 100µL
Applications
ChIP assay (1:50), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511199 309.00
Check your price
  • Phospho-Histone H3 (Ser28) Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Bovine, Chicken, Fruit fly, Hamster, Human, Mouse, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA517318
  • Size 100µL
Applications
Flow Cytometry (1:200), Immunocytochemistry (1:400), Immunofluorescence (1:400), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517318 309.00
Check your price
  • Histone H3.3 Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Bovine, Fruit fly, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA517697
  • Size 100µL
Applications
Immunofluorescence (1:100), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517697 275.00
Check your price
  • Di-Methyl-Histone H3 (Lys9) Polyclonal Antibody for Western Blot, IF, IHC, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Fruit fly, Human, Mouse, Non-human primate, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # PA516195
  • Size 100µL
Applications
ChIP assay (1:25), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Assay-Dependent), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:25), Western Blot (Assay-Dependent)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516195 309.00
Check your price
  • Phospho-Histone H3 (Ser10) Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and Flow
  • Host Rabbit
  • Target Species Fruit fly, Human, Mouse, Non-human primate, Rat, Xenopus, Yeast
  • Conjugate Unconjugated
  • Catalog # PA517869
  • Size 100µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Frozen) (1:400), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517869 309.00
Check your price
  • Histone H2A Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Non-human primate, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA517600
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone H2A.X; XH2AZ; H2A.Z1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517600 275.00
Check your price
  • Di-Methyl-Histone H3 (Lys27) Monoclonal Antibody for Western Blot, IF and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA511197
  • Size 100µL
Applications
ChIP assay (1:25), Immunofluorescence (1:1600), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511197 375.00
Check your price
  • Histone H2A.X Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141004
  • Size 100µg
Applications
ELISA (1:1000), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141004 309.00
Check your price
  • Histone H2A.X Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA528778
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528778 349.00
Check your price
  • Histone H3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116941
  • Size 100µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116941 375.00
Check your price
  • Histone H3.3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Chicken, Fruit fly, Horse, Human, Marsupial, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523288
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (0.25-0.5 µg/ml)
Alternate names:
Histone H3, H3.3A, H3.3B, H3F3, H3F3A, H3F3B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523288 349.00
Check your price
  • Histone H2A.X Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523450
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (4-7 µg/ml)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523450 349.00
Check your price
  • Histone H2A.X Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523465
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523465 349.00
Check your price
  • BRAF35 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA146226
  • Size 100µg
Applications
ELISA (1:100-1:2000), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (1:10-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Structural DNA-binding protein BRAF35; Sox-like transcriptional factor; HMG domain-containing protein HMGX2; HMG domain-containing protein 2; HMG box-containing protein 20B; BRCA2-associated factor 35
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146226 309.00
Check your price
  • CREB Monoclonal Antibody for Western Blot, IF, ICC, Flow and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701120
  • Size 100µg
Applications
ChIP assay (1 ul), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:500-1:5000), Immunofluorescence (1:500-1:5000), Western Blot (1:500-1:5000)
Alternate names:
Active transcription factor CREB, cAMP responsive element binding protein 1, CREB1, MGC9284, Transactivator protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
701120 309.00
Check your price
  • 53BP1 Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate DyLight® 650
  • Catalog # PA522758
  • Size 100µl
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Western Blot (1:200-1:10,000)
Alternate names:
53BP1, p202, p53-binding protein 1, p53BP1, tumor protein 53-binding protein, 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522758 375.00
Check your price