1 - 30 of 30 products displayed
  • HDAC9 Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523346
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
MEF2-interacting transcription repressor MITR; Histone deacetylase-related protein; Histone deacetylase 7B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523346 349.00
Check your price
  • HDAC4 Polyclonal Antibody for Western Blot, IHC (P) and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1863
  • Size 100 µg
Applications
ChIP assay (1-3 ul), Immunohistochemistry (Paraffin) (1:20), Western Blot (1:1,000)
Alternate names:
Histone Deacetylase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1863 349.00
Check your price
  • Phospho-HDAC4 (Ser632) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538135
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA, histone deacetylase A, DKFZp779K1053, HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9, HDAC9B, HDAC9FL, HDRP, histone deacetylase 4/5-related protein, histone deacetylase 7, Histone deacetylase 7B, Histone deacetylase 9, Histone deacetylase-related protein, KIAA0744, MEF-2 interacti, HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR, MEF-2 interacting transcription repressor (MITR) protein, histone deacetylase 4/5-related protein, histone deacetylase 7B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538135 309.00
Check your price
  • Phospho-HDAC4/HDAC5/HDAC9 (Ser246, Ser259, Ser220) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537835
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Antigen NY-CO-9; MEF2-interacting transcription repressor MITR; Histone deacetylase-related protein; Histone deacetylase 7B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537835 309.00
Check your price
  • HDAC4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511236
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Histone deacetylase 4; HD4; KIAA0288
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511236 349.00
Check your price
  • Phospho-HDAC5/HDAC9 (Ser259, Ser220) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540215
  • Size 100 µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Antigen NY-CO-9; MEF2-interacting transcription repressor MITR; Histone deacetylase-related protein; Histone deacetylase 7B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540215 309.00
Check your price
  • HDAC9 Polyclonal Antibody for Western Blot, IHC (P) and ChIP
  • Host Rabbit
  • Target Species Guinea pig, Human, Mouse, Sheep
  • Conjugate Unconjugated
  • Catalog # PA542247
  • Size 100 µl
Applications
ChIP assay (Assay-Dependent), Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (1.25 µg/ml)
Alternate names:
MEF2-interacting transcription repressor MITR; Histone deacetylase-related protein; Histone deacetylase 7B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542247 309.00
Check your price
  • HDAC9 Polyclonal Antibody for Western Blot, IF, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511245
  • Size 400 µL
Applications
ChIP assay (1-3 ul), Immunofluorescence (1:1000), Immunohistochemistry (1:50-1:100), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
MEF2-interacting transcription repressor MITR; Histone deacetylase-related protein; Histone deacetylase 7B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511245 349.00
Check your price
  • HDAC9 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511246
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
MEF2-interacting transcription repressor MITR; Histone deacetylase-related protein; Histone deacetylase 7B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511246 349.00
Check your price
  • HDAC4 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529103
  • Size 100 µL
Applications
ChIP assay (Assay-Dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA, histone deacetylase A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529103 349.00
Check your price
  • HDAC4 Synthetic Peptide for BLOCK and Ctrl
  • Host N/A
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PEP095
  • Size 50 ug
Applications
Blocking Assay (Assay Dependent), Control (Assay Dependent)
Alternate names:
AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA, histone deacetylase A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP095 109.00
Check your price
  • HDAC4 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A16688
  • Size 100 µL
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA, histone deacetylase A
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16688 309.00
Check your price
  • Phospho-HDAC4/HDAC5/HDAC7 (Ser246, Ser259, Ser155) Monoclonal Antibody for Western Blot, IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514776
  • Size 100 µL
Applications
ChIP assay (1-3 ul), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone deacetylase 7A; Antigen NY-CO-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514776 309.00
Check your price
  • HDAC4 Polyclonal Antibody for Western Blot, IP and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141092
  • Size 100 µg
Applications
ChIP assay (Assay Dependent.), Immunoprecipitation (Assay-Dependent), Western Blot (0.5-2 µg/ml)
Alternate names:
HA6116, HD4, HDAC-A, HDACA, KIAA0288
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141092 375.00
Check your price
  • HDAC4 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515580
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
HD4; HDACA; HA6116; HDAC-A; KIAA0288; HDAC4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515580 349.00
Check your price
  • Phospho-HDAC7 (Ser448) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523384
  • Size 50 ug
Applications
Western Blot (0.1-0.5 µg/ml)
Alternate names:
Histone deacetylase 7A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523384 349.00
Check your price
  • Phospho-HDAC7 (Ser155) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523202
  • Size 50 ug
Applications
Western Blot (0.1-1 µg/ml)
Alternate names:
Histone deacetylase 7A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523202 349.00
Check your price
  • Phospho-HDAC7 (Ser318) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Non-human primate, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA523203
  • Size 50 ug
Applications
Western Blot (0.1-1 µg/ml)
Alternate names:
Histone deacetylase 7A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523203 349.00
Check your price
  • HDAC2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532444
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:25)
Alternate names:
HD2, RPD3, YAF1, YY1-associated factor 1, transcriptional regulator homolog RPD3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532444 375.00
Check your price
  • HDAC10 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123496
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
RP3-402G11.7, DKFZp761B039, MGC149722
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123496 399.00
Check your price
  • HDAC1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532443
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
GON-10, HD1, RPD3, RPD3L1, reduced potassium dependency, yeast homolog-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532443 275.00
Check your price
  • HDAC10 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA146293
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
RP3-402G11.7, DKFZp761B039, MGC149722
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146293 309.00
Check your price
  • HDAC1 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IP and ChIP
  • Host Rabbit
  • Target Species Dog, Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1860
  • Size 100 µg
Applications
ChIP assay (Assay dependent), Immunocytochemistry (2.0 µg/mL), Immunofluorescence (Assay dependent), Immunohistochemistry (1:200), Immunohistochemistry (Frozen) (0.2 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Histone Deacetylase 1, DKFZp686H12203, GON-10, HD1, RPD3, RPD3L1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1860 375.00
Check your price
  • HDAC6 Polyclonal Antibody for Western Blot, IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141056
  • Size 100 µg
Applications
ChIP assay (1:10-1:500), Immunoprecipitation (Assay-Dependent), Western Blot (2-5 µg/ml)
Alternate names:
Histone deacetylase mHDA2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141056 375.00
Check your price
  • HDAC7 Polyclonal Antibody for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Rabbit
  • Target Species Mouse, Human
  • Conjugate
  • Catalog # PA523131
  • Size 100ug
Applications
ChIP assay (1-3 ul), Immunocytochemistry (5-10 µg/ml), Immunofluorescence (5-10 µg/ml), Immunohistochemistry (Paraffin) (2-5 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Histone deacetylase 7A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523131 349.00
Check your price
  • HDAC1 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA120354
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (2-4 µg/ml)
Alternate names:
Histone Deacetylase 1, DKFZp686H12203, GON-10, HD1, RPD3, RPD3L1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120354 475.00
Check your price
  • HDAC1 Polyclonal Antibody for IF, ICC, ChIP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 491025
  • Size 50 µg
Applications
ChIP assay (2.4 ug), ELISA (1:100 to 1:300), Immunocytochemistry (1:200), Immunofluorescence (1:200)
Alternate names:
Histone deacetylase 1, HD1, HDAC1, RPD3L1
Catalog # Size Price (AUD) Your Price (AUD) Qty
491025 304.00
Check your price
  • HDAC1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and ChIP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA518071
  • Size 100 µL
Applications
ChIP assay (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:500-1:4000)
Alternate names:
Histone deacetylase 1; HD1; RPD3L1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518071 425.00
Check your price
  • HDAC7 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511242
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Histone deacetylase 7A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511242 349.00
Check your price
  • Phospho-HDAC8 (Ser39) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536648
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Histone deacetylase 8, HD8, HDAC8, HDACL1, CDA07
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536648 349.00
Check your price