1 - 3 of 3 products displayed
  • Histone H1.5 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519487
  • Size 100µg
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (5 µg/ml), Immunohistochemistry (Paraffin) (1:800), Western Blot (0.5 µg/ml)
Alternate names:
Histone H1s-3; Histone H1b; Histone H1a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519487 349.00
Check your price
  • Histone H1.5 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525155
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Histone H1s-3; Histone H1b; Histone H1a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525155 349.00
Check your price
  • Phospho-Histone H1 (Thr17) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540288
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Histone H1s-3; Histone H1b; Histone H1a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540288 309.00
Check your price