1 - 30 of 30 products displayed
  • Histone H2A Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Non-human primate, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA517600
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone H2A.X; XH2AZ; H2A.Z1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517600 275.00
Check your price
  • Histone H2A Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA535893
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
Histone H2A/p; Histone H2A/m; Histone H2A/a; Histone H2A.2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535893 309.00
Check your price
  • Acetyl-Histone H2A (Lys5) Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517763
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone H2A/p; Histone H2A/m; Histone H2A/a; Histone H2A.2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517763 309.00
Check your price
  • Phospho-Histone H2A (Ser129) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540149
  • Size 100 µl
Applications
Western Blot (1:1000-1:2000)
Alternate names:
Histone H2A/r; Histone H2A/p
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540149 309.00
Check your price
  • Acetyl-Histone H2A (Lys5) Polyclonal Antibody for Western Blot, ChIP, ChIP-seq and ARRAY
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540102
  • Size 50 µg
Applications
ChIP assay (0.5-1 µg), ChIP-sequencing (Assay dependant), Peptide Array (1:5000), Western Blot (1:1000)
Alternate names:
Histone H2A/p
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540102 309.00
Check your price
  • Histone H2A Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # 710045
  • Size 100 µg
Applications
Western Blot (4-5 ug/ml)
Alternate names:
Histone H2A/p; Histone H2A/s; Histone H2A/e; Histone H2A/m; Histone H2A/a; Histone H2A.2; Histone H2A/g; Histone H2A.3
Catalog # Size Price (AUD) Your Price (AUD) Qty
710045 309.00
Check your price
  • Histone H2A Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # 700158
  • Size 100 µg
Applications
Western Blot (0.1-2 ug/ml)
Alternate names:
Histone H2A/p; Histone H2A/s; Histone H2A/e; Histone H2A/m; Histone H2A/a; Histone H2A.2; Histone H2A/g; Histone H2A.3
Catalog # Size Price (AUD) Your Price (AUD) Qty
700158 309.00
Check your price
  • Histone H2A.X Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141004
  • Size 100 µg
Applications
ELISA (1:1000), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141004 309.00
Check your price
  • Acetyl-Histone H2A (Lys5) Polyclonal Antibody for Western Blot, IF, ICC, Flow and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 720070
  • Size 100 µg
Applications
ChIP assay (5 ug/ml), Flow Cytometry (2-4 ug/million cells), Immunocytochemistry (0.5 ug/ml), Immunofluorescence (0.5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Histone H2A/p
Catalog # Size Price (AUD) Your Price (AUD) Qty
720070 309.00
Check your price
  • Acetyl-Histone H3 (Lys9) Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow, IP and ChIP
  • Host Rabbit
  • Target Species C. elegans, Human, Mouse, Non-human primate, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA511195
  • Size 100 µL
Applications
ChIP assay (1:50), Flow Cytometry (1:200), Immunocytochemistry (1:400), Immunofluorescence (1:400), Immunohistochemistry (Paraffin) (1:800), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511195 309.00
Check your price
  • Histone H3 Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Fruit fly, Hamster, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA515150
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515150 275.00
Check your price
  • Histone H2A.X Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523450
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (4-7 µg/ml)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523450 349.00
Check your price
  • Histone H2A.X Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523465
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523465 349.00
Check your price
  • Histone H3 Polyclonal Antibody for Western Blot, IF, ICC, Flow and ChIP
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Fruit fly, Horse, Human, Mouse, Non-human primate, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA516183
  • Size 100 µL
Applications
ChIP assay (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516183 309.00
Check your price
  • Histone H3.3 Polyclonal Antibody for Western Blot, IF, ICC, Flow and ChIP
  • Host Rabbit
  • Target Species Bovine, Fruit fly, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA517697
  • Size 100 µL
Applications
Immunofluorescence (1:100), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517697 275.00
Check your price
  • Histone H2A.X Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA528778
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528778 349.00
Check your price
  • Histone H2A.X Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528785
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528785 349.00
Check your price
  • Acetyl-Histone H3 (Lys9, Lys14) Polyclonal Antibody for Western Blot, IF, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species C. elegans, Human, Mouse, Non-human primate, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA516194
  • Size 100 µL
Applications
ChIP assay (1:50), Immunofluorescence (1:5000), Immunohistochemistry (Paraffin) (1:800), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516194 349.00
Check your price
  • Phospho-Histone H2A.X (Thr120) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA540152
  • Size 100 µl
Applications
Western Blot (1:1000-1:2000)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540152 309.00
Check your price
  • Phospho-Histone H2A.X (Tyr142) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540153
  • Size 100 µl
Applications
Western Blot (1:1000-1:2000)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540153 309.00
Check your price
  • Histone H2A.Z Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA517336
  • Size 100 µL
Applications
Immunocytochemistry (1:400), Immunofluorescence (1:200), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
XH2AZ; H2A.Z1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517336 275.00
Check your price
  • Histone H2A.Z Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521923
  • Size 100 µL
Applications
ChIP assay (Assay-dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:500-1:1000), Western Blot (1:500-1:3000)
Alternate names:
H2A.z, H2A/z, H2AZ, H2AZ histone, histone H2A.Z
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521923 349.00
Check your price
  • Histone H2A.Z Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521928
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
H2A.z, H2A/z, H2AZ, H2AZ histone, histone H2A.Z
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521928 349.00
Check your price
  • Histone H2A.X Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA184856
  • Size 50µg
Applications
Immunofluorescence (1:400-1:800), Western Blot (1:5,000-1:25,000)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184856 389.00
Check your price
  • HIST1H2AG Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525266
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Histone H2A/p
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525266 349.00
Check your price
  • HIST1H2AG Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524822
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Histone H2A/p
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524822 349.00
Check your price
  • HIST2H2AA3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA543692
  • Size 100 µl
Applications
Western Blot (2.5 µg/ml)
Alternate names:
Histone H2A.2; Histone H2A/o
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543692 309.00
Check your price
  • HIST2H2AC Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA543691
  • Size 100 µl
Applications
Western Blot (2.5 µg/ml)
Alternate names:
H2a-613B; Histone H2A/q; Histone H2A-GL101
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543691 309.00
Check your price
  • Tri-Methyl-Histone H3 (Lys27) Monoclonal Antibody for Western Blot, IF, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA511198
  • Size 100 µL
Applications
ChIP assay (1:50), Immunofluorescence (1:1600), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511198 349.00
Check your price
  • Phospho-Histone H3 (Ser10) Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515220
  • Size 100 µL
Applications
Flow Cytometry (1:25), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Frozen) (1:100), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515220 309.00
Check your price