1 - 8 of 8 products displayed
  • HLA-A2 Monoclonal Antibody for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180117
  • Size 200µg
Applications
Flow Cytometry (10 ug/ml), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180117 375.00
Check your price
  • HLA-A2 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA516587
  • Size 100ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516587 425.00
Check your price
  • HLA-A2 Monoclonal Antibody, Alexa Fluor® 488 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 488
  • Catalog # MA518162
  • Size 100tests
Applications
Flow Cytometry (4ulx100ul whole blood or 10^6 cells)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518162 275.00
Check your price
  • HLA-A2 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA110345
  • Size 100Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110345 275.00
Check your price
  • HLA-A2 Monoclonal Antibody for Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110344
  • Size 100ug
Applications
Flow Cytometry (5-10 ug/ml), Immunoprecipitation (Assay dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110344 225.00
Check your price
  • HLA-A2 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA180303
  • Size 100Tests
Applications
Flow Cytometry (Neat)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180303 499.00
Check your price
  • HLA-A2 Monoclonal Antibody for IHC (F), Flow, IP and FN
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516586
  • Size 500ug
Applications
Flow Cytometry (10 ug/ml), Functional Assay (5 -10 ug/ml), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516586 499.00
Check your price
  • WHSC2 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA517199
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Wolf-Hirschhorn syndrome candidate 2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517199 309.00
Check your price